Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 98744756-98745256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS1B
Entrez Gene ID
56899
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
UniGene
Hs.506458
Gene Chromosome Location
Chr.12: 98729904-99984727 on Build GRCh38
Transcripts
NM_001204065.1 NM_001204066.1 NM_001204067.1 NM_001204068.1 NM_001204069.1 NM_001204070.1 NM_001204079.1 NM_001204080.1 NM_001204081.1 NM_020140.3 NM_152788.4 NM_181670.3 XM_005269028.4 XM_005269029.4 XM_005269032.3 XM_006719504.3 XM_006719507.3 XM_006719512.3 XM_006719514.3 XM_011538571.2 XM_017019651.1 XM_017019652.1 XM_017019653.1 XM_017019654.1 XM_017019656.1 XM_017019657.1 XM_017019658.1 XM_017019659.1 XM_017019660.1 XM_017019661.1 XM_017019663.1 XM_017019664.1

SNP Detail

SNP ID Chromosome Location
rs1028019439 Chr.12: 98744949-98744949 on Build GRCh38
rs555187322 Chr.12: 98745029-98745029 on Build GRCh38
rs775901630 Chr.12: 98744964-98744964 on Build GRCh38
rs57332865 Chr.12: 98744990-98744990 on Build GRCh38
rs373673986 Chr.12: 98744915-98744915 on Build GRCh38
rs139460220 Chr.12: 98744879-98744879 on Build GRCh38
rs1021334712 Chr.12: 98745056-98745056 on Build GRCh38
rs968362990 Chr.12: 98745077-98745077 on Build GRCh38
rs956218611 Chr.12: 98745018-98745018 on Build GRCh38
rs938632729 Chr.12: 98745193-98745193 on Build GRCh38
rs569601706 Chr.12: 98744867-98744867 on Build GRCh38
rs887331254 Chr.12: 98744946-98744946 on Build GRCh38
rs927294350 Chr.12: 98745180-98745180 on Build GRCh38
rs896332944 Chr.12: 98744845-98744845 on Build GRCh38
rs953695659 Chr.12: 98744983-98744983 on Build GRCh38
rs942766835 Chr.12: 98744836-98744836 on Build GRCh38
rs1003065041 Chr.12: 98744969-98744969 on Build GRCh38
rs980574649 Chr.12: 98745151-98745151 on Build GRCh38
rs112521698 Chr.12: 98744869-98744869 on Build GRCh38
rs958513783 Chr.12: 98744981-98744981 on Build GRCh38
rs767541336 Chr.12: 98745118-98745118 on Build GRCh38
rs939915439 Chr.12: 98745221-98745221 on Build GRCh38
rs532519350 Chr.12: 98744808-98744808 on Build GRCh38
rs185612716 Chr.12: 98744789-98744789 on Build GRCh38
rs1040196404 Chr.12: 98745216-98745216 on Build GRCh38
rs1036252791 Chr.12: 98744971-98744971 on Build GRCh38
rs1004014874 Chr.12: 98744868-98744868 on Build GRCh38
rs764658752 Chr.12: 98745204-98745204 on Build GRCh38
rs947521748 Chr.12: 98745107-98745107 on Build GRCh38
rs914724166 Chr.12: 98745067-98745067 on Build GRCh38
rs898498490 Chr.12: 98744848-98744848 on Build GRCh38
rs59438243 Chr.12: 98745087-98745087 on Build GRCh38
rs896364076 Chr.12: 98745214-98745214 on Build GRCh38
rs551028786 Chr.12: 98744854-98744854 on Build GRCh38
rs977027107 Chr.12: 98745080-98745080 on Build GRCh38
rs954610094 Chr.12: 98745102-98745102 on Build GRCh38
rs71305588 Chr.12: 98744808-98744809 on Build GRCh38
rs867714621 Chr.12: 98744956-98744956 on Build GRCh38
rs1047075457 Chr.12: 98745226-98745227 on Build GRCh38
rs568588207 Chr.12: 98745209-98745209 on Build GRCh38
rs988956492 Chr.12: 98745019-98745019 on Build GRCh38
rs570388701 Chr.12: 98745122-98745122 on Build GRCh38
rs942839651 Chr.12: 98745202-98745202 on Build GRCh38
rs924550098 Chr.12: 98745094-98745094 on Build GRCh38
rs912672087 Chr.12: 98744828-98744828 on Build GRCh38
rs34366930 Chr.12: 98744795-98744796 on Build GRCh38
rs552229582 Chr.12: 98744991-98744991 on Build GRCh38
rs572849686 Chr.12: 98745222-98745222 on Build GRCh38
rs931318263 Chr.12: 98745226-98745226 on Build GRCh38

View Full Product Details