Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101394800-101395316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL1
Entrez Gene ID
400
Gene Name
ADP ribosylation factor like GTPase 1
Gene Aliases
ARFL1
UniGene
Hs.372616
Gene Chromosome Location
Chr.12: 101393120-101407820 on Build GRCh38
Transcripts
NM_001177.5 NM_001301068.1

SNP Detail

SNP ID Chromosome Location
rs575394810 Chr.12: 101395205-101395205 on Build GRCh38
rs1001658759 Chr.12: 101394835-101394835 on Build GRCh38
rs911367543 Chr.12: 101395087-101395087 on Build GRCh38
rs866150139 Chr.12: 101395007-101395007 on Build GRCh38
rs894268453 Chr.12: 101395184-101395184 on Build GRCh38
rs1024260059 Chr.12: 101395195-101395195 on Build GRCh38
rs1018488905 Chr.12: 101395053-101395053 on Build GRCh38
rs905743026 Chr.12: 101395264-101395264 on Build GRCh38
rs760289951 Chr.12: 101394883-101394883 on Build GRCh38
rs958378762 Chr.12: 101395270-101395274 on Build GRCh38
rs759886743 Chr.12: 101395141-101395141 on Build GRCh38
rs867502225 Chr.12: 101395265-101395265 on Build GRCh38
rs555614468 Chr.12: 101394952-101394952 on Build GRCh38
rs999548809 Chr.12: 101395254-101395254 on Build GRCh38
rs777042815 Chr.12: 101395119-101395119 on Build GRCh38
rs903097475 Chr.12: 101395240-101395240 on Build GRCh38
rs535505073 Chr.12: 101394897-101394897 on Build GRCh38
rs894396294 Chr.12: 101394953-101394953 on Build GRCh38
rs976934847 Chr.12: 101395089-101395089 on Build GRCh38
rs574074211 Chr.12: 101395274-101395274 on Build GRCh38
rs1005837468 Chr.12: 101394850-101394850 on Build GRCh38
rs888196562 Chr.12: 101394847-101394848 on Build GRCh38
rs369873218 Chr.12: 101394853-101394853 on Build GRCh38
rs180803521 Chr.12: 101395223-101395223 on Build GRCh38
rs893230377 Chr.12: 101395275-101395275 on Build GRCh38
rs955633976 Chr.12: 101395029-101395029 on Build GRCh38
rs55944830 Chr.12: 101394839-101394839 on Build GRCh38
rs942975348 Chr.12: 101395088-101395088 on Build GRCh38
rs138304077 Chr.12: 101395030-101395030 on Build GRCh38
rs569764256 Chr.12: 101394864-101394864 on Build GRCh38
rs964205596 Chr.12: 101395054-101395054 on Build GRCh38
rs190997739 Chr.12: 101395101-101395101 on Build GRCh38
rs753161722 Chr.12: 101395170-101395170 on Build GRCh38
rs558078298 Chr.12: 101395160-101395160 on Build GRCh38
rs1034169092 Chr.12: 101394932-101394932 on Build GRCh38
rs1014156118 Chr.12: 101394964-101394964 on Build GRCh38
rs764305292 Chr.12: 101395218-101395218 on Build GRCh38
rs929679469 Chr.12: 101395109-101395109 on Build GRCh38
rs990080863 Chr.12: 101395036-101395036 on Build GRCh38
rs974230314 Chr.12: 101395078-101395078 on Build GRCh38
rs537102409 Chr.12: 101395190-101395190 on Build GRCh38
rs772803067 Chr.12: 101394882-101394882 on Build GRCh38
rs1045613935 Chr.12: 101395261-101395261 on Build GRCh38
rs1020275515 Chr.12: 101395010-101395010 on Build GRCh38
rs1056053020 Chr.12: 101395174-101395174 on Build GRCh38
rs1024982019 Chr.12: 101394984-101394984 on Build GRCh38
rs958128179 Chr.12: 101395035-101395035 on Build GRCh38

View Full Product Details