Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101395292-101395793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL1
Entrez Gene ID
400
Gene Name
ADP ribosylation factor like GTPase 1
Gene Aliases
ARFL1
UniGene
Hs.372616
Gene Chromosome Location
Chr.12: 101393120-101407820 on Build GRCh38
Transcripts
NM_001177.5 NM_001301068.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
933959 c.516-1G>T p.? Chr.12: 101395671-101395671 on Build GRCh38
3810785 c.546A>C p.*182Y Chr.12: 101395640-101395640 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748105215 Chr.12: 101395718-101395718 on Build GRCh38
rs374031068 Chr.12: 101395529-101395529 on Build GRCh38
rs78828803 Chr.12: 101395427-101395427 on Build GRCh38
rs983151212 Chr.12: 101395407-101395407 on Build GRCh38
rs143825195 Chr.12: 101395545-101395545 on Build GRCh38
rs1803152 Chr.12: 101395542-101395542 on Build GRCh38
rs779293997 Chr.12: 101395594-101395594 on Build GRCh38
rs995753728 Chr.12: 101395692-101395692 on Build GRCh38
rs778851795 Chr.12: 101395641-101395641 on Build GRCh38
rs201219157 Chr.12: 101395637-101395637 on Build GRCh38
rs981033224 Chr.12: 101395487-101395487 on Build GRCh38
rs1029772142 Chr.12: 101395715-101395715 on Build GRCh38
rs891124075 Chr.12: 101395324-101395324 on Build GRCh38
rs1004628309 Chr.12: 101395753-101395753 on Build GRCh38
rs915655256 Chr.12: 101395481-101395484 on Build GRCh38
rs1061351 Chr.12: 101395455-101395455 on Build GRCh38
rs528616190 Chr.12: 101395327-101395327 on Build GRCh38
rs772884998 Chr.12: 101395701-101395701 on Build GRCh38
rs754717821 Chr.12: 101395332-101395332 on Build GRCh38
rs769377805 Chr.12: 101395680-101395680 on Build GRCh38
rs746184587 Chr.12: 101395597-101395597 on Build GRCh38
rs558117025 Chr.12: 101395610-101395610 on Build GRCh38
rs1008524441 Chr.12: 101395335-101395335 on Build GRCh38
rs900100900 Chr.12: 101395654-101395654 on Build GRCh38
rs1027129439 Chr.12: 101395368-101395368 on Build GRCh38
rs1052885066 Chr.12: 101395557-101395557 on Build GRCh38
rs964291777 Chr.12: 101395356-101395356 on Build GRCh38
rs190602389 Chr.12: 101395384-101395384 on Build GRCh38
rs183080651 Chr.12: 101395359-101395359 on Build GRCh38
rs949718387 Chr.12: 101395470-101395470 on Build GRCh38
rs1027593650 Chr.12: 101395727-101395727 on Build GRCh38
rs761389991 Chr.12: 101395644-101395644 on Build GRCh38
rs1054719947 Chr.12: 101395591-101395591 on Build GRCh38
rs903128453 Chr.12: 101395538-101395538 on Build GRCh38
rs148098547 Chr.12: 101395324-101395325 on Build GRCh38
rs780940661 Chr.12: 101395432-101395436 on Build GRCh38
rs150388630 Chr.12: 101395621-101395621 on Build GRCh38
rs951650034 Chr.12: 101395728-101395728 on Build GRCh38
rs759251451 Chr.12: 101395593-101395594 on Build GRCh38
rs190860736 Chr.12: 101395349-101395349 on Build GRCh38
rs752248214 Chr.12: 101395687-101395687 on Build GRCh38
rs951571508 Chr.12: 101395742-101395742 on Build GRCh38
rs781323341 Chr.12: 101395580-101395580 on Build GRCh38
rs769264006 Chr.12: 101395633-101395633 on Build GRCh38
rs927227540 Chr.12: 101395526-101395526 on Build GRCh38
rs891178400 Chr.12: 101395623-101395623 on Build GRCh38
rs535206532 Chr.12: 101395507-101395507 on Build GRCh38
rs534799307 Chr.12: 101395608-101395608 on Build GRCh38
rs772593664 Chr.12: 101395605-101395605 on Build GRCh38
rs1014628588 Chr.12: 101395755-101395755 on Build GRCh38
rs559533125 Chr.12: 101395323-101395323 on Build GRCh38
rs764391253 Chr.12: 101395564-101395564 on Build GRCh38
rs187207395 Chr.12: 101395372-101395372 on Build GRCh38
rs1055514837 Chr.12: 101395409-101395409 on Build GRCh38
rs549050067 Chr.12: 101395532-101395532 on Build GRCh38

View Full Product Details