Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 103988905-103989419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLT8D2
Entrez Gene ID
83468
Gene Name
glycosyltransferase 8 domain containing 2
Gene Aliases
-
UniGene
Hs.631650
Gene Chromosome Location
Chr.12: 103988983-104064177 on Build GRCh38
Transcripts
NM_001316967.1 NM_031302.4 XM_011538793.2 XM_017019999.1

SNP Detail

SNP ID Chromosome Location
rs947872623 Chr.12: 103989298-103989298 on Build GRCh38
rs766412486 Chr.12: 103989306-103989306 on Build GRCh38
rs997578636 Chr.12: 103989121-103989121 on Build GRCh38
rs34064725 Chr.12: 103989320-103989321 on Build GRCh38
rs141969591 Chr.12: 103989290-103989290 on Build GRCh38
rs929210631 Chr.12: 103989021-103989021 on Build GRCh38
rs892537744 Chr.12: 103989113-103989113 on Build GRCh38
rs556762110 Chr.12: 103989250-103989250 on Build GRCh38
rs766190040 Chr.12: 103989368-103989368 on Build GRCh38
rs1056299549 Chr.12: 103988947-103988947 on Build GRCh38
rs181748546 Chr.12: 103989227-103989227 on Build GRCh38
rs1000868475 Chr.12: 103989044-103989044 on Build GRCh38
rs568889806 Chr.12: 103989284-103989284 on Build GRCh38
rs371318251 Chr.12: 103989360-103989360 on Build GRCh38
rs556214401 Chr.12: 103989244-103989244 on Build GRCh38
rs918287335 Chr.12: 103989254-103989254 on Build GRCh38
rs911317466 Chr.12: 103988951-103988951 on Build GRCh38
rs941887352 Chr.12: 103989083-103989083 on Build GRCh38
rs973759106 Chr.12: 103989253-103989253 on Build GRCh38
rs762174344 Chr.12: 103989041-103989041 on Build GRCh38
rs953519236 Chr.12: 103989160-103989160 on Build GRCh38
rs547204196 Chr.12: 103988985-103988985 on Build GRCh38
rs1052898415 Chr.12: 103989106-103989106 on Build GRCh38
rs999722820 Chr.12: 103989278-103989278 on Build GRCh38
rs896345854 Chr.12: 103988954-103988954 on Build GRCh38
rs556130977 Chr.12: 103989335-103989335 on Build GRCh38
rs185728185 Chr.12: 103988967-103988967 on Build GRCh38
rs1047716644 Chr.12: 103989043-103989043 on Build GRCh38
rs1030391180 Chr.12: 103989127-103989127 on Build GRCh38
rs751300162 Chr.12: 103989377-103989377 on Build GRCh38
rs527516972 Chr.12: 103988972-103988972 on Build GRCh38
rs951094292 Chr.12: 103989257-103989257 on Build GRCh38
rs923148928 Chr.12: 103989016-103989016 on Build GRCh38
rs767661523 Chr.12: 103989120-103989120 on Build GRCh38
rs567990597 Chr.12: 103988970-103988970 on Build GRCh38
rs903769340 Chr.12: 103989017-103989017 on Build GRCh38
rs188629782 Chr.12: 103989218-103989218 on Build GRCh38

View Full Product Details