Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 103989254-103989783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLT8D2
Entrez Gene ID
83468
Gene Name
glycosyltransferase 8 domain containing 2
Gene Aliases
-
UniGene
Hs.631650
Gene Chromosome Location
Chr.12: 103988983-104064177 on Build GRCh38
Transcripts
NM_001316967.1 NM_031302.4 XM_011538793.2 XM_017019999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6479904 c.1009C>T p.P337S Chr.12: 103989449-103989449 on Build GRCh38
2064434 c.952C>T p.H318Y Chr.12: 103989506-103989506 on Build GRCh38
934210 c.889C>T p.P297S Chr.12: 103989569-103989569 on Build GRCh38
4189086 c.1021G>T p.G341W Chr.12: 103989437-103989437 on Build GRCh38
3721938 c.1018G>A p.A340T Chr.12: 103989440-103989440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779114174 Chr.12: 103989423-103989423 on Build GRCh38
rs772719598 Chr.12: 103989623-103989623 on Build GRCh38
rs889202474 Chr.12: 103989532-103989532 on Build GRCh38
rs947872623 Chr.12: 103989298-103989298 on Build GRCh38
rs766412486 Chr.12: 103989306-103989306 on Build GRCh38
rs747344462 Chr.12: 103989465-103989465 on Build GRCh38
rs1052368450 Chr.12: 103989504-103989504 on Build GRCh38
rs202178249 Chr.12: 103989466-103989466 on Build GRCh38
rs754883873 Chr.12: 103989405-103989405 on Build GRCh38
rs916300836 Chr.12: 103989661-103989661 on Build GRCh38
rs539171225 Chr.12: 103989690-103989690 on Build GRCh38
rs772666198 Chr.12: 103989495-103989495 on Build GRCh38
rs949162749 Chr.12: 103989678-103989678 on Build GRCh38
rs969575473 Chr.12: 103989747-103989747 on Build GRCh38
rs568889806 Chr.12: 103989284-103989284 on Build GRCh38
rs1051764332 Chr.12: 103989531-103989531 on Build GRCh38
rs371318251 Chr.12: 103989360-103989360 on Build GRCh38
rs762506232 Chr.12: 103989626-103989626 on Build GRCh38
rs751157772 Chr.12: 103989551-103989551 on Build GRCh38
rs980616779 Chr.12: 103989750-103989750 on Build GRCh38
rs925132100 Chr.12: 103989759-103989759 on Build GRCh38
rs994181791 Chr.12: 103989668-103989668 on Build GRCh38
rs925213424 Chr.12: 103989447-103989447 on Build GRCh38
rs746116688 Chr.12: 103989427-103989427 on Build GRCh38
rs779167284 Chr.12: 103989581-103989581 on Build GRCh38
rs759442554 Chr.12: 103989552-103989552 on Build GRCh38
rs762596787 Chr.12: 103989499-103989499 on Build GRCh38
rs772386158 Chr.12: 103989435-103989435 on Build GRCh38
rs999722820 Chr.12: 103989278-103989278 on Build GRCh38
rs983356145 Chr.12: 103989494-103989494 on Build GRCh38
rs556130977 Chr.12: 103989335-103989335 on Build GRCh38
rs781359673 Chr.12: 103989595-103989595 on Build GRCh38
rs963651136 Chr.12: 103989659-103989659 on Build GRCh38
rs755805314 Chr.12: 103989522-103989526 on Build GRCh38
rs997485235 Chr.12: 103989521-103989521 on Build GRCh38
rs762281315 Chr.12: 103989481-103989481 on Build GRCh38
rs200062838 Chr.12: 103989536-103989536 on Build GRCh38
rs780384630 Chr.12: 103989583-103989583 on Build GRCh38
rs991825667 Chr.12: 103989696-103989696 on Build GRCh38
rs767185474 Chr.12: 103989553-103989553 on Build GRCh38
rs1026587169 Chr.12: 103989679-103989679 on Build GRCh38
rs902172289 Chr.12: 103989709-103989709 on Build GRCh38
rs976739693 Chr.12: 103989445-103989445 on Build GRCh38
rs34064725 Chr.12: 103989320-103989321 on Build GRCh38
rs752437757 Chr.12: 103989474-103989476 on Build GRCh38
rs141969591 Chr.12: 103989290-103989290 on Build GRCh38
rs766190040 Chr.12: 103989368-103989368 on Build GRCh38
rs750464748 Chr.12: 103989582-103989582 on Build GRCh38
rs374206216 Chr.12: 103989564-103989564 on Build GRCh38
rs140349477 Chr.12: 103989474-103989474 on Build GRCh38
rs765776236 Chr.12: 103989473-103989473 on Build GRCh38
rs757453205 Chr.12: 103989414-103989414 on Build GRCh38
rs572794992 Chr.12: 103989601-103989601 on Build GRCh38
rs752730912 Chr.12: 103989561-103989561 on Build GRCh38
rs377345089 Chr.12: 103989592-103989592 on Build GRCh38
rs777516869 Chr.12: 103989630-103989630 on Build GRCh38
rs999169889 Chr.12: 103989715-103989715 on Build GRCh38
rs781247503 Chr.12: 103989406-103989406 on Build GRCh38
rs892442798 Chr.12: 103989520-103989520 on Build GRCh38
rs765998052 Chr.12: 103989539-103989539 on Build GRCh38
rs751300162 Chr.12: 103989377-103989377 on Build GRCh38
rs965852641 Chr.12: 103989430-103989430 on Build GRCh38
rs149680129 Chr.12: 103989604-103989604 on Build GRCh38
rs748030157 Chr.12: 103989408-103989408 on Build GRCh38
rs775724282 Chr.12: 103989456-103989456 on Build GRCh38
rs917668557 Chr.12: 103989736-103989736 on Build GRCh38
rs747195968 Chr.12: 103989589-103989589 on Build GRCh38
rs769312571 Chr.12: 103989483-103989483 on Build GRCh38

View Full Product Details