Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 103996643-103997155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLT8D2
Entrez Gene ID
83468
Gene Name
glycosyltransferase 8 domain containing 2
Gene Aliases
-
UniGene
Hs.631650
Gene Chromosome Location
Chr.12: 103988983-104064177 on Build GRCh38
Transcripts
NM_001316967.1 NM_031302.4 XM_011538793.2 XM_017019999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1322044 c.536C>T p.A179V Chr.12: 103996799-103996799 on Build GRCh38
5374007 c.496C>T p.Q166* Chr.12: 103996839-103996839 on Build GRCh38
1358314 c.589G>A p.V197M Chr.12: 103996746-103996746 on Build GRCh38
2150943 c.532G>A p.A178T Chr.12: 103996803-103996803 on Build GRCh38
4553507 c.592G>A p.G198R Chr.12: 103996743-103996743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536108955 Chr.12: 103996721-103996721 on Build GRCh38
rs777460443 Chr.12: 103996803-103996803 on Build GRCh38
rs561589585 Chr.12: 103997085-103997085 on Build GRCh38
rs997422207 Chr.12: 103996898-103996898 on Build GRCh38
rs376728289 Chr.12: 103996869-103996869 on Build GRCh38
rs913957563 Chr.12: 103996675-103996675 on Build GRCh38
rs143889059 Chr.12: 103996825-103996825 on Build GRCh38
rs776076472 Chr.12: 103997003-103997003 on Build GRCh38
rs149453281 Chr.12: 103996823-103996823 on Build GRCh38
rs763454796 Chr.12: 103996854-103996854 on Build GRCh38
rs368329923 Chr.12: 103996819-103996819 on Build GRCh38
rs766942055 Chr.12: 103996876-103996876 on Build GRCh38
rs1020924239 Chr.12: 103997050-103997050 on Build GRCh38
rs556930573 Chr.12: 103996678-103996678 on Build GRCh38
rs377436632 Chr.12: 103996693-103996693 on Build GRCh38
rs141060407 Chr.12: 103996820-103996820 on Build GRCh38
rs766848963 Chr.12: 103996866-103996866 on Build GRCh38
rs990566599 Chr.12: 103997090-103997090 on Build GRCh38
rs576824387 Chr.12: 103996700-103996700 on Build GRCh38
rs765529546 Chr.12: 103996737-103996737 on Build GRCh38
rs893901658 Chr.12: 103996842-103996842 on Build GRCh38
rs973606119 Chr.12: 103997119-103997119 on Build GRCh38
rs770392521 Chr.12: 103996984-103996984 on Build GRCh38
rs563317685 Chr.12: 103996956-103996956 on Build GRCh38
rs141452676 Chr.12: 103996742-103996742 on Build GRCh38
rs548041984 Chr.12: 103997078-103997078 on Build GRCh38
rs199878047 Chr.12: 103996780-103996780 on Build GRCh38
rs916226807 Chr.12: 103997116-103997116 on Build GRCh38
rs983577466 Chr.12: 103996724-103996724 on Build GRCh38
rs773288133 Chr.12: 103996849-103996849 on Build GRCh38
rs111374835 Chr.12: 103996674-103996674 on Build GRCh38
rs373952967 Chr.12: 103996802-103996802 on Build GRCh38
rs941660822 Chr.12: 103996743-103996743 on Build GRCh38
rs762476373 Chr.12: 103996736-103996736 on Build GRCh38
rs79327130 Chr.12: 103997087-103997087 on Build GRCh38
rs746760272 Chr.12: 103996861-103996863 on Build GRCh38
rs910645800 Chr.12: 103997089-103997089 on Build GRCh38
rs908979919 Chr.12: 103996732-103996732 on Build GRCh38
rs3817602 Chr.12: 103996747-103996747 on Build GRCh38
rs762078725 Chr.12: 103996735-103996735 on Build GRCh38
rs1032803475 Chr.12: 103996838-103996838 on Build GRCh38
rs757022179 Chr.12: 103996811-103996811 on Build GRCh38
rs547205216 Chr.12: 103997096-103997096 on Build GRCh38
rs920827403 Chr.12: 103997120-103997120 on Build GRCh38
rs376059234 Chr.12: 103996893-103996893 on Build GRCh38
rs752211940 Chr.12: 103996881-103996881 on Build GRCh38
rs755743531 Chr.12: 103996885-103996885 on Build GRCh38
rs577983940 Chr.12: 103996903-103996903 on Build GRCh38
rs150432657 Chr.12: 103996746-103996746 on Build GRCh38
rs988533375 Chr.12: 103997035-103997035 on Build GRCh38
rs147324980 Chr.12: 103996801-103996801 on Build GRCh38
rs768899800 Chr.12: 103996890-103996890 on Build GRCh38
rs767045215 Chr.12: 103996759-103996759 on Build GRCh38
rs943507689 Chr.12: 103997095-103997095 on Build GRCh38
rs929547451 Chr.12: 103997123-103997123 on Build GRCh38
rs570511772 Chr.12: 103997118-103997118 on Build GRCh38
rs1025149245 Chr.12: 103996913-103996913 on Build GRCh38
rs7133444 Chr.12: 103996779-103996779 on Build GRCh38
rs113971087 Chr.12: 103996870-103996870 on Build GRCh38
rs957972230 Chr.12: 103997030-103997030 on Build GRCh38
rs768673698 Chr.12: 103996822-103996822 on Build GRCh38
rs543424156 Chr.12: 103996939-103996939 on Build GRCh38
rs894846877 Chr.12: 103996798-103996798 on Build GRCh38
rs531399775 Chr.12: 103996804-103996804 on Build GRCh38
rs148324304 Chr.12: 103996958-103996958 on Build GRCh38
rs753455691 Chr.12: 103996894-103996894 on Build GRCh38
rs369770376 Chr.12: 103996707-103996707 on Build GRCh38
rs768689085 Chr.12: 103996701-103996701 on Build GRCh38
rs949083267 Chr.12: 103997117-103997120 on Build GRCh38
rs143994495 Chr.12: 103996799-103996799 on Build GRCh38

View Full Product Details