Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 106238211-106238724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKAP4
Entrez Gene ID
10970
Gene Name
cytoskeleton associated protein 4
Gene Aliases
CLIMP-63, ERGIC-63, p63
UniGene
Hs.74368
Gene Chromosome Location
Chr.12: 106237881-106247935 on Build GRCh38
Transcripts
NM_006825.3

SNP Detail

SNP ID Chromosome Location
rs1003274884 Chr.12: 106238648-106238649 on Build GRCh38
rs539772064 Chr.12: 106238341-106238341 on Build GRCh38
rs1051229294 Chr.12: 106238570-106238571 on Build GRCh38
rs1021084080 Chr.12: 106238422-106238422 on Build GRCh38
rs889948115 Chr.12: 106238575-106238575 on Build GRCh38
rs112124678 Chr.12: 106238308-106238308 on Build GRCh38
rs902083193 Chr.12: 106238322-106238322 on Build GRCh38
rs181473538 Chr.12: 106238313-106238313 on Build GRCh38
rs71452903 Chr.12: 106238430-106238430 on Build GRCh38
rs1051088126 Chr.12: 106238588-106238588 on Build GRCh38
rs918638802 Chr.12: 106238551-106238551 on Build GRCh38
rs139021046 Chr.12: 106238360-106238360 on Build GRCh38
rs941321663 Chr.12: 106238544-106238544 on Build GRCh38
rs1007079070 Chr.12: 106238393-106238394 on Build GRCh38
rs984517449 Chr.12: 106238366-106238366 on Build GRCh38
rs1019655244 Chr.12: 106238319-106238319 on Build GRCh38
rs71442016 Chr.12: 106238676-106238676 on Build GRCh38
rs1034886702 Chr.12: 106238649-106238651 on Build GRCh38
rs75086692 Chr.12: 106238675-106238676 on Build GRCh38
rs991792439 Chr.12: 106238440-106238440 on Build GRCh38
rs962755653 Chr.12: 106238663-106238667 on Build GRCh38
rs552751682 Chr.12: 106238435-106238435 on Build GRCh38
rs71468294 Chr.12: 106238428-106238428 on Build GRCh38
rs150994370 Chr.12: 106238244-106238244 on Build GRCh38
rs981070720 Chr.12: 106238419-106238419 on Build GRCh38
rs138744965 Chr.12: 106238587-106238587 on Build GRCh38
rs1034837336 Chr.12: 106238460-106238460 on Build GRCh38
rs914424819 Chr.12: 106238467-106238467 on Build GRCh38
rs770718055 Chr.12: 106238663-106238663 on Build GRCh38
rs63264962 Chr.12: 106238600-106238601 on Build GRCh38
rs115024037 Chr.12: 106238504-106238504 on Build GRCh38
rs924022517 Chr.12: 106238500-106238500 on Build GRCh38
rs528558789 Chr.12: 106238698-106238698 on Build GRCh38
rs7132695 Chr.12: 106238658-106238658 on Build GRCh38
rs759804666 Chr.12: 106238662-106238664 on Build GRCh38
rs372592438 Chr.12: 106238660-106238660 on Build GRCh38
rs563720547 Chr.12: 106238624-106238624 on Build GRCh38
rs769643643 Chr.12: 106238301-106238301 on Build GRCh38
rs1037492426 Chr.12: 106238329-106238329 on Build GRCh38
rs570576157 Chr.12: 106238663-106238666 on Build GRCh38
rs981911104 Chr.12: 106238453-106238453 on Build GRCh38
rs961690118 Chr.12: 106238396-106238396 on Build GRCh38
rs771800291 Chr.12: 106238451-106238451 on Build GRCh38
rs544105127 Chr.12: 106238622-106238622 on Build GRCh38
rs1010962702 Chr.12: 106238593-106238593 on Build GRCh38
rs748265205 Chr.12: 106238340-106238340 on Build GRCh38
rs529415790 Chr.12: 106238239-106238239 on Build GRCh38
rs774823882 Chr.12: 106238596-106238597 on Build GRCh38
rs1010663610 Chr.12: 106238420-106238420 on Build GRCh38
rs966550106 Chr.12: 106238434-106238434 on Build GRCh38
rs937255876 Chr.12: 106238439-106238439 on Build GRCh38
rs972860319 Chr.12: 106238548-106238548 on Build GRCh38
rs117471598 Chr.12: 106238257-106238257 on Build GRCh38
rs7303700 Chr.12: 106238663-106238663 on Build GRCh38
rs1014592357 Chr.12: 106238385-106238385 on Build GRCh38
rs71452904 Chr.12: 106238519-106238519 on Build GRCh38
rs938006241 Chr.12: 106238277-106238277 on Build GRCh38
rs144908227 Chr.12: 106238272-106238272 on Build GRCh38
rs11112910 Chr.12: 106238373-106238373 on Build GRCh38
rs952188550 Chr.12: 106238363-106238363 on Build GRCh38
rs202044819 Chr.12: 106238471-106238471 on Build GRCh38
rs572602784 Chr.12: 106238472-106238472 on Build GRCh38
rs145732575 Chr.12: 106238581-106238581 on Build GRCh38
rs1040360540 Chr.12: 106238571-106238571 on Build GRCh38
rs7112 Chr.12: 106238353-106238353 on Build GRCh38
rs893474643 Chr.12: 106238592-106238592 on Build GRCh38
rs541520325 Chr.12: 106238567-106238567 on Build GRCh38
rs553980888 Chr.12: 106238446-106238448 on Build GRCh38
rs187153693 Chr.12: 106238629-106238629 on Build GRCh38
rs140881125 Chr.12: 106238273-106238273 on Build GRCh38
rs536361719 Chr.12: 106238377-106238377 on Build GRCh38
rs10574 Chr.12: 106238261-106238261 on Build GRCh38
rs772930346 Chr.12: 106238565-106238566 on Build GRCh38
rs890066586 Chr.12: 106238390-106238390 on Build GRCh38
rs1017309352 Chr.12: 106238401-106238401 on Build GRCh38
rs547876788 Chr.12: 106238300-106238300 on Build GRCh38
rs909894521 Chr.12: 106238515-106238515 on Build GRCh38
rs569924071 Chr.12: 106238633-106238633 on Build GRCh38

View Full Product Details