Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 106972181-106972688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM263
Entrez Gene ID
90488
Gene Name
transmembrane protein 263
Gene Aliases
C12orf23
UniGene
Hs.257664
Gene Chromosome Location
Chr.12: 106955535-106974037 on Build GRCh38
Transcripts
NM_001319661.1 NM_001319662.1 NM_001319663.1 NM_001319664.1 NM_001319666.1 NM_152261.3 XM_017020198.1

SNP Detail

SNP ID Chromosome Location
rs541760330 Chr.12: 106972465-106972465 on Build GRCh38
rs902613331 Chr.12: 106972343-106972343 on Build GRCh38
rs1003467348 Chr.12: 106972478-106972478 on Build GRCh38
rs915765937 Chr.12: 106972301-106972306 on Build GRCh38
rs550525568 Chr.12: 106972424-106972424 on Build GRCh38
rs959427006 Chr.12: 106972501-106972501 on Build GRCh38
rs564630200 Chr.12: 106972531-106972531 on Build GRCh38
rs758638480 Chr.12: 106972602-106972602 on Build GRCh38
rs1021228532 Chr.12: 106972437-106972437 on Build GRCh38
rs965978840 Chr.12: 106972363-106972363 on Build GRCh38
rs939627925 Chr.12: 106972246-106972246 on Build GRCh38
rs578220520 Chr.12: 106972588-106972588 on Build GRCh38
rs1034988466 Chr.12: 106972479-106972479 on Build GRCh38
rs907605329 Chr.12: 106972441-106972441 on Build GRCh38
rs895342700 Chr.12: 106972318-106972318 on Build GRCh38
rs1055291089 Chr.12: 106972260-106972260 on Build GRCh38
rs769225545 Chr.12: 106972525-106972525 on Build GRCh38
rs984600928 Chr.12: 106972436-106972436 on Build GRCh38
rs192474126 Chr.12: 106972372-106972372 on Build GRCh38
rs555761822 Chr.12: 106972308-106972308 on Build GRCh38
rs1051501959 Chr.12: 106972351-106972353 on Build GRCh38
rs1055744740 Chr.12: 106972650-106972650 on Build GRCh38
rs1022356383 Chr.12: 106972339-106972339 on Build GRCh38
rs1018979525 Chr.12: 106972358-106972358 on Build GRCh38
rs915718591 Chr.12: 106972539-106972539 on Build GRCh38
rs928323989 Chr.12: 106972546-106972546 on Build GRCh38
rs527320242 Chr.12: 106972381-106972381 on Build GRCh38
rs999503337 Chr.12: 106972353-106972353 on Build GRCh38
rs918900587 Chr.12: 106972330-106972330 on Build GRCh38
rs775474068 Chr.12: 106972584-106972585 on Build GRCh38
rs755969924 Chr.12: 106972244-106972244 on Build GRCh38
rs963266685 Chr.12: 106972618-106972618 on Build GRCh38
rs938107412 Chr.12: 106972238-106972238 on Build GRCh38
rs940465731 Chr.12: 106972522-106972522 on Build GRCh38
rs893985327 Chr.12: 106972389-106972389 on Build GRCh38
rs1011559717 Chr.12: 106972415-106972416 on Build GRCh38
rs748415617 Chr.12: 106972422-106972422 on Build GRCh38
rs967108571 Chr.12: 106972451-106972452 on Build GRCh38
rs779789597 Chr.12: 106972418-106972418 on Build GRCh38
rs982141393 Chr.12: 106972210-106972210 on Build GRCh38
rs1011075646 Chr.12: 106972328-106972328 on Build GRCh38
rs879198626 Chr.12: 106972386-106972386 on Build GRCh38
rs12299494 Chr.12: 106972221-106972221 on Build GRCh38
rs940330700 Chr.12: 106972206-106972206 on Build GRCh38
rs568844348 Chr.12: 106972227-106972227 on Build GRCh38
rs12320317 Chr.12: 106972659-106972659 on Build GRCh38

View Full Product Details