Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108647355-108647865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO1C
Entrez Gene ID
23603
Gene Name
coronin 1C
Gene Aliases
HCRNN4
UniGene
Hs.330384
Gene Chromosome Location
Chr.12: 108645109-108731550 on Build GRCh38
Transcripts
NM_001105237.2 NM_001276471.1 NM_014325.3 XM_011538125.1 XM_017019121.1 XM_017019122.1 XM_017019123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3687933 c.1382G>A p.R461H Chr.12: 108647446-108647446 on Build GRCh38
3810899 c.1372C>G p.Q458E Chr.12: 108647456-108647456 on Build GRCh38
6634233 c.1422C>A p.A474A Chr.12: 108647406-108647406 on Build GRCh38
3455685 c.1383T>C p.R461R Chr.12: 108647445-108647445 on Build GRCh38
25175 c.1324G>A p.D442N Chr.12: 108647504-108647504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375229600 Chr.12: 108647701-108647701 on Build GRCh38
rs771648298 Chr.12: 108647541-108647541 on Build GRCh38
rs1051641491 Chr.12: 108647695-108647695 on Build GRCh38
rs767300014 Chr.12: 108647390-108647390 on Build GRCh38
rs764073423 Chr.12: 108647551-108647551 on Build GRCh38
rs893075235 Chr.12: 108647727-108647727 on Build GRCh38
rs548307242 Chr.12: 108647626-108647626 on Build GRCh38
rs780319369 Chr.12: 108647419-108647419 on Build GRCh38
rs1022442973 Chr.12: 108647737-108647737 on Build GRCh38
rs972780257 Chr.12: 108647669-108647669 on Build GRCh38
rs889648907 Chr.12: 108647446-108647446 on Build GRCh38
rs775279256 Chr.12: 108647723-108647723 on Build GRCh38
rs151306281 Chr.12: 108647513-108647513 on Build GRCh38
rs956881936 Chr.12: 108647705-108647705 on Build GRCh38
rs774905941 Chr.12: 108647544-108647544 on Build GRCh38
rs538734067 Chr.12: 108647771-108647771 on Build GRCh38
rs541171484 Chr.12: 108647588-108647588 on Build GRCh38
rs201986662 Chr.12: 108647412-108647412 on Build GRCh38
rs918557120 Chr.12: 108647674-108647674 on Build GRCh38
rs117660611 Chr.12: 108647749-108647749 on Build GRCh38
rs531112642 Chr.12: 108647686-108647686 on Build GRCh38
rs373745426 Chr.12: 108647568-108647568 on Build GRCh38
rs1023299799 Chr.12: 108647673-108647673 on Build GRCh38
rs746944357 Chr.12: 108647418-108647418 on Build GRCh38
rs1020346232 Chr.12: 108647592-108647592 on Build GRCh38
rs1009758700 Chr.12: 108647706-108647706 on Build GRCh38
rs780890146 Chr.12: 108647499-108647499 on Build GRCh38
rs1034083983 Chr.12: 108647616-108647616 on Build GRCh38
rs748719946 Chr.12: 108647467-108647467 on Build GRCh38
rs781501120 Chr.12: 108647461-108647461 on Build GRCh38
rs377569941 Chr.12: 108647484-108647484 on Build GRCh38
rs910142623 Chr.12: 108647629-108647629 on Build GRCh38
rs1003832196 Chr.12: 108647699-108647699 on Build GRCh38
rs755094723 Chr.12: 108647452-108647452 on Build GRCh38
rs986177597 Chr.12: 108647788-108647788 on Build GRCh38
rs758925057 Chr.12: 108647532-108647532 on Build GRCh38
rs774161804 Chr.12: 108647527-108647527 on Build GRCh38
rs149788602 Chr.12: 108647620-108647620 on Build GRCh38
rs986144684 Chr.12: 108647421-108647421 on Build GRCh38
rs1003063245 Chr.12: 108647609-108647609 on Build GRCh38
rs576276914 Chr.12: 108647386-108647386 on Build GRCh38
rs763848395 Chr.12: 108647394-108647394 on Build GRCh38
rs139157556 Chr.12: 108647550-108647550 on Build GRCh38
rs982731542 Chr.12: 108647381-108647381 on Build GRCh38
rs957084197 Chr.12: 108647392-108647392 on Build GRCh38
rs141599477 Chr.12: 108647599-108647600 on Build GRCh38
rs530069070 Chr.12: 108647625-108647625 on Build GRCh38
rs778259532 Chr.12: 108647482-108647482 on Build GRCh38
rs971184288 Chr.12: 108647605-108647605 on Build GRCh38
rs1009906635 Chr.12: 108647590-108647590 on Build GRCh38
rs117503985 Chr.12: 108647651-108647651 on Build GRCh38
rs376884358 Chr.12: 108647681-108647681 on Build GRCh38
rs888207900 Chr.12: 108647631-108647631 on Build GRCh38
rs777569084 Chr.12: 108647397-108647397 on Build GRCh38
rs76470389 Chr.12: 108647481-108647481 on Build GRCh38
rs553938120 Chr.12: 108647837-108647837 on Build GRCh38
rs751523459 Chr.12: 108647423-108647423 on Build GRCh38
rs770766039 Chr.12: 108647383-108647383 on Build GRCh38
rs764528013 Chr.12: 108647591-108647591 on Build GRCh38

View Full Product Details