Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109880118-109880614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLTP
Entrez Gene ID
51228
Gene Name
glycolipid transfer protein
Gene Aliases
-
UniGene
Hs.381256
Gene Chromosome Location
Chr.12: 109850943-109880488 on Build GRCh38
Transcripts
NM_016433.3 XM_011538434.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1298893 c.54C>G p.I18M Chr.12: 109880321-109880321 on Build GRCh38
4038599 c.19C>T p.H7Y Chr.12: 109880356-109880356 on Build GRCh38
7086444 c.102C>T p.F34F Chr.12: 109880273-109880273 on Build GRCh38
4645824 c.7C>A p.L3M Chr.12: 109880368-109880368 on Build GRCh38
7088286 c.72C>T p.L24L Chr.12: 109880303-109880303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048358838 Chr.12: 109880186-109880186 on Build GRCh38
rs779537357 Chr.12: 109880236-109880236 on Build GRCh38
rs998813726 Chr.12: 109880430-109880430 on Build GRCh38
rs765906968 Chr.12: 109880355-109880355 on Build GRCh38
rs143113542 Chr.12: 109880255-109880255 on Build GRCh38
rs375674800 Chr.12: 109880344-109880344 on Build GRCh38
rs780642735 Chr.12: 109880309-109880309 on Build GRCh38
rs1002670674 Chr.12: 109880172-109880172 on Build GRCh38
rs776568557 Chr.12: 109880250-109880250 on Build GRCh38
rs758807029 Chr.12: 109880374-109880374 on Build GRCh38
rs1046617298 Chr.12: 109880148-109880148 on Build GRCh38
rs370224148 Chr.12: 109880359-109880359 on Build GRCh38
rs888526160 Chr.12: 109880377-109880377 on Build GRCh38
rs561849029 Chr.12: 109880154-109880154 on Build GRCh38
rs901429967 Chr.12: 109880406-109880406 on Build GRCh38
rs374681174 Chr.12: 109880261-109880261 on Build GRCh38
rs888475457 Chr.12: 109880211-109880211 on Build GRCh38
rs758682528 Chr.12: 109880287-109880287 on Build GRCh38
rs970638660 Chr.12: 109880508-109880508 on Build GRCh38
rs965614392 Chr.12: 109880235-109880235 on Build GRCh38
rs767820677 Chr.12: 109880258-109880258 on Build GRCh38
rs916666552 Chr.12: 109880205-109880205 on Build GRCh38
rs780801883 Chr.12: 109880392-109880392 on Build GRCh38
rs982463349 Chr.12: 109880512-109880512 on Build GRCh38
rs745651942 Chr.12: 109880423-109880423 on Build GRCh38
rs746588820 Chr.12: 109880296-109880296 on Build GRCh38
rs889973597 Chr.12: 109880443-109880443 on Build GRCh38
rs745667233 Chr.12: 109880400-109880432 on Build GRCh38
rs868407641 Chr.12: 109880511-109880511 on Build GRCh38
rs747697981 Chr.12: 109880393-109880393 on Build GRCh38
rs1037047185 Chr.12: 109880587-109880587 on Build GRCh38
rs949261209 Chr.12: 109880142-109880142 on Build GRCh38
rs749174248 Chr.12: 109880312-109880312 on Build GRCh38
rs754525285 Chr.12: 109880306-109880306 on Build GRCh38
rs779811031 Chr.12: 109880288-109880288 on Build GRCh38
rs764202926 Chr.12: 109880267-109880267 on Build GRCh38
rs1010577260 Chr.12: 109880264-109880264 on Build GRCh38
rs199600346 Chr.12: 109880339-109880339 on Build GRCh38
rs983970191 Chr.12: 109880537-109880537 on Build GRCh38
rs749112289 Chr.12: 109880407-109880407 on Build GRCh38
rs754540722 Chr.12: 109880387-109880387 on Build GRCh38
rs769773157 Chr.12: 109880311-109880311 on Build GRCh38
rs569511418 Chr.12: 109880166-109880166 on Build GRCh38
rs745535608 Chr.12: 109880225-109880225 on Build GRCh38
rs769036371 Chr.12: 109880426-109880426 on Build GRCh38
rs773966724 Chr.12: 109880417-109880417 on Build GRCh38
rs906365419 Chr.12: 109880244-109880244 on Build GRCh38
rs778854058 Chr.12: 109880313-109880317 on Build GRCh38
rs761951056 Chr.12: 109880350-109880350 on Build GRCh38
rs755617575 Chr.12: 109880400-109880400 on Build GRCh38
rs748109225 Chr.12: 109880310-109880310 on Build GRCh38
rs375791569 Chr.12: 109880349-109880349 on Build GRCh38
rs909968377 Chr.12: 109880551-109880551 on Build GRCh38
rs751259266 Chr.12: 109880386-109880386 on Build GRCh38
rs746593825 Chr.12: 109880242-109880242 on Build GRCh38
rs71458309 Chr.12: 109880456-109880456 on Build GRCh38
rs539843013 Chr.12: 109880149-109880149 on Build GRCh38
rs937878090 Chr.12: 109880530-109880530 on Build GRCh38
rs760820940 Chr.12: 109880265-109880265 on Build GRCh38
rs770572843 Chr.12: 109880332-109880332 on Build GRCh38
rs1046243015 Chr.12: 109880325-109880325 on Build GRCh38
rs940050679 Chr.12: 109880562-109880562 on Build GRCh38
rs556224155 Chr.12: 109880446-109880446 on Build GRCh38
rs770790097 Chr.12: 109880414-109880414 on Build GRCh38
rs138072824 Chr.12: 109880224-109880224 on Build GRCh38
rs765813437 Chr.12: 109880275-109880275 on Build GRCh38
rs866777297 Chr.12: 109880370-109880370 on Build GRCh38
rs367798721 Chr.12: 109880273-109880273 on Build GRCh38
rs926549369 Chr.12: 109880524-109880524 on Build GRCh38
rs533516142 Chr.12: 109880283-109880283 on Build GRCh38
rs868143439 Chr.12: 109880502-109880502 on Build GRCh38
rs1036993239 Chr.12: 109880394-109880394 on Build GRCh38
rs545922353 Chr.12: 109880210-109880210 on Build GRCh38
rs775784984 Chr.12: 109880336-109880336 on Build GRCh38
rs957746584 Chr.12: 109880495-109880495 on Build GRCh38
rs771803771 Chr.12: 109880227-109880227 on Build GRCh38
rs987775413 Chr.12: 109880498-109880498 on Build GRCh38
rs777746270 Chr.12: 109880405-109880405 on Build GRCh38
rs949270997 Chr.12: 109880321-109880321 on Build GRCh38
rs552094505 Chr.12: 109880435-109880435 on Build GRCh38
rs1048410356 Chr.12: 109880185-109880185 on Build GRCh38
rs1015889112 Chr.12: 109880229-109880229 on Build GRCh38
rs760766596 Chr.12: 109880343-109880343 on Build GRCh38
rs201843232 Chr.12: 109880378-109880378 on Build GRCh38
rs893688525 Chr.12: 109880263-109880263 on Build GRCh38
rs757699018 Chr.12: 109880223-109880223 on Build GRCh38
rs761675478 Chr.12: 109880254-109880254 on Build GRCh38

View Full Product Details