Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132773047-132773559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA3
Entrez Gene ID
2802
Gene Name
golgin A3
Gene Aliases
GCP170, MEA-2
UniGene
Hs.507333
Gene Chromosome Location
Chr.12: 132768909-132829131 on Build GRCh38
Transcripts
NM_005895.3 XM_005266164.4 XM_005266167.4 XM_006719736.3 XM_006719737.3 XM_011534793.2 XM_011534794.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
692173 c.4480G>C p.E1494Q Chr.12: 132773122-132773122 on Build GRCh38
3811550 c.4350C>A p.A1450A Chr.12: 132773252-132773252 on Build GRCh38
4040421 c.4440C>T p.R1480R Chr.12: 132773162-132773162 on Build GRCh38
1476358 c.4444G>A p.D1482N Chr.12: 132773158-132773158 on Build GRCh38
4627014 c.4381A>G p.T1461A Chr.12: 132773221-132773221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765635809 Chr.12: 132773255-132773255 on Build GRCh38
rs1045691181 Chr.12: 132773489-132773489 on Build GRCh38
rs1010530493 Chr.12: 132773393-132773393 on Build GRCh38
rs775039337 Chr.12: 132773302-132773304 on Build GRCh38
rs553555087 Chr.12: 132773187-132773187 on Build GRCh38
rs894319050 Chr.12: 132773475-132773475 on Build GRCh38
rs779662104 Chr.12: 132773352-132773352 on Build GRCh38
rs900278523 Chr.12: 132773335-132773335 on Build GRCh38
rs958353274 Chr.12: 132773162-132773162 on Build GRCh38
rs368889038 Chr.12: 132773085-132773085 on Build GRCh38
rs374060454 Chr.12: 132773184-132773184 on Build GRCh38
rs141251873 Chr.12: 132773171-132773171 on Build GRCh38
rs780575322 Chr.12: 132773104-132773104 on Build GRCh38
rs1005423702 Chr.12: 132773129-132773129 on Build GRCh38
rs771597649 Chr.12: 132773258-132773260 on Build GRCh38
rs759272409 Chr.12: 132773332-132773332 on Build GRCh38
rs551771941 Chr.12: 132773152-132773152 on Build GRCh38
rs938020653 Chr.12: 132773091-132773091 on Build GRCh38
rs747991076 Chr.12: 132773317-132773317 on Build GRCh38
rs757377416 Chr.12: 132773302-132773302 on Build GRCh38
rs966276657 Chr.12: 132773536-132773536 on Build GRCh38
rs76846220 Chr.12: 132773469-132773469 on Build GRCh38
rs1032788132 Chr.12: 132773374-132773374 on Build GRCh38
rs976363252 Chr.12: 132773423-132773423 on Build GRCh38
rs1032694966 Chr.12: 132773504-132773504 on Build GRCh38
rs762891677 Chr.12: 132773081-132773081 on Build GRCh38
rs771356896 Chr.12: 132773406-132773406 on Build GRCh38
rs760366618 Chr.12: 132773242-132773242 on Build GRCh38
rs532038952 Chr.12: 132773133-132773133 on Build GRCh38
rs777921874 Chr.12: 132773183-132773183 on Build GRCh38
rs966660888 Chr.12: 132773411-132773411 on Build GRCh38
rs766104172 Chr.12: 132773245-132773245 on Build GRCh38
rs957506191 Chr.12: 132773384-132773384 on Build GRCh38
rs749061772 Chr.12: 132773225-132773225 on Build GRCh38
rs752558734 Chr.12: 132773103-132773103 on Build GRCh38
rs113568898 Chr.12: 132773170-132773170 on Build GRCh38
rs568807811 Chr.12: 132773153-132773153 on Build GRCh38
rs75423335 Chr.12: 132773325-132773325 on Build GRCh38
rs772802956 Chr.12: 132773240-132773240 on Build GRCh38
rs774397166 Chr.12: 132773161-132773161 on Build GRCh38
rs774152620 Chr.12: 132773231-132773231 on Build GRCh38
rs999010111 Chr.12: 132773366-132773366 on Build GRCh38
rs995883806 Chr.12: 132773214-132773214 on Build GRCh38
rs771429316 Chr.12: 132773239-132773239 on Build GRCh38
rs919248430 Chr.12: 132773430-132773430 on Build GRCh38
rs75050779 Chr.12: 132773479-132773479 on Build GRCh38
rs767054682 Chr.12: 132773092-132773092 on Build GRCh38
rs1035047129 Chr.12: 132773415-132773415 on Build GRCh38
rs190685447 Chr.12: 132773348-132773348 on Build GRCh38
rs892825784 Chr.12: 132773351-132773351 on Build GRCh38
rs201715887 Chr.12: 132773247-132773247 on Build GRCh38
rs909326290 Chr.12: 132773458-132773458 on Build GRCh38
rs370075864 Chr.12: 132773159-132773159 on Build GRCh38
rs758966043 Chr.12: 132773283-132773283 on Build GRCh38
rs751606757 Chr.12: 132773301-132773301 on Build GRCh38
rs557621368 Chr.12: 132773167-132773167 on Build GRCh38
rs748124922 Chr.12: 132773235-132773235 on Build GRCh38
rs575572787 Chr.12: 132773392-132773392 on Build GRCh38
rs765823558 Chr.12: 132773177-132773177 on Build GRCh38
rs138174089 Chr.12: 132773404-132773404 on Build GRCh38
rs778053983 Chr.12: 132773403-132773403 on Build GRCh38
rs929356222 Chr.12: 132773433-132773433 on Build GRCh38
rs971258008 Chr.12: 132773246-132773246 on Build GRCh38
rs888273119 Chr.12: 132773271-132773271 on Build GRCh38
rs865885098 Chr.12: 132773291-132773291 on Build GRCh38
rs754289604 Chr.12: 132773173-132773173 on Build GRCh38
rs763323388 Chr.12: 132773222-132773224 on Build GRCh38
rs984103876 Chr.12: 132773461-132773461 on Build GRCh38
rs144951708 Chr.12: 132773254-132773254 on Build GRCh38
rs376755932 Chr.12: 132773079-132773079 on Build GRCh38
rs902832450 Chr.12: 132773492-132773492 on Build GRCh38
rs986272616 Chr.12: 132773396-132773396 on Build GRCh38
rs756705119 Chr.12: 132773363-132773363 on Build GRCh38
rs756433590 Chr.12: 132773100-132773100 on Build GRCh38
rs764038125 Chr.12: 132773292-132773292 on Build GRCh38
rs781738756 Chr.12: 132773304-132773304 on Build GRCh38
rs748553141 Chr.12: 132773376-132773376 on Build GRCh38
rs753086431 Chr.12: 132773272-132773272 on Build GRCh38
rs957079559 Chr.12: 132773507-132773507 on Build GRCh38
rs1049122945 Chr.12: 132773454-132773454 on Build GRCh38
rs200698266 Chr.12: 132773074-132773074 on Build GRCh38
rs200447509 Chr.12: 132773163-132773163 on Build GRCh38
rs776806026 Chr.12: 132773407-132773407 on Build GRCh38
rs1055525582 Chr.12: 132773472-132773472 on Build GRCh38
rs372947141 Chr.12: 132773086-132773086 on Build GRCh38
rs759668837 Chr.12: 132773307-132773308 on Build GRCh38
rs779601354 Chr.12: 132773130-132773130 on Build GRCh38
rs754701774 Chr.12: 132773309-132773309 on Build GRCh38
rs776351702 Chr.12: 132773331-132773331 on Build GRCh38
rs769521039 Chr.12: 132773158-132773158 on Build GRCh38
rs1018150550 Chr.12: 132773533-132773533 on Build GRCh38
rs773642496 Chr.12: 132773126-132773126 on Build GRCh38
rs746501549 Chr.12: 132773323-132773323 on Build GRCh38
rs772081477 Chr.12: 132773318-132773318 on Build GRCh38
rs755567339 Chr.12: 132773115-132773115 on Build GRCh38
rs529674657 Chr.12: 132773226-132773226 on Build GRCh38
rs556090921 Chr.12: 132773217-132773217 on Build GRCh38
rs202114902 Chr.12: 132773178-132773178 on Build GRCh38
rs3741484 Chr.12: 132773307-132773307 on Build GRCh38
rs1007322519 Chr.12: 132773526-132773526 on Build GRCh38
rs1032650700 Chr.12: 132773350-132773350 on Build GRCh38
rs755547704 Chr.12: 132773308-132773308 on Build GRCh38
rs990803590 Chr.12: 132773303-132773303 on Build GRCh38
rs376982029 Chr.12: 132773114-132773114 on Build GRCh38
rs768552052 Chr.12: 132773160-132773160 on Build GRCh38
rs944993851 Chr.12: 132773180-132773180 on Build GRCh38
rs540049033 Chr.12: 132773522-132773522 on Build GRCh38
rs886575779 Chr.12: 132773316-132773316 on Build GRCh38
rs920569225 Chr.12: 132773434-132773434 on Build GRCh38
rs546328865 Chr.12: 132773078-132773078 on Build GRCh38
rs959475886 Chr.12: 132773421-132773421 on Build GRCh38
rs370553580 Chr.12: 132773179-132773179 on Build GRCh38
rs894361801 Chr.12: 132773313-132773313 on Build GRCh38
rs765469060 Chr.12: 132773341-132773341 on Build GRCh38
rs750148652 Chr.12: 132773098-132773098 on Build GRCh38
rs755439620 Chr.12: 132773174-132773174 on Build GRCh38
rs747258920 Chr.12: 132773157-132773157 on Build GRCh38
rs761216719 Chr.12: 132773166-132773166 on Build GRCh38
rs781431131 Chr.12: 132773190-132773190 on Build GRCh38
rs773066814 Chr.12: 132773322-132773322 on Build GRCh38
rs775668250 Chr.12: 132773500-132773500 on Build GRCh38

View Full Product Details