Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 24318490-24318995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF9
Entrez Gene ID
338872
Gene Name
C1q and tumor necrosis factor related protein 9
Gene Aliases
AQL1, C1QTNF9A, CTRP9
UniGene
Hs.362854
Gene Chromosome Location
Chr.13: 24307166-24322531 on Build GRCh38
Transcripts
NM_001303137.1 NM_001303138.1 NM_178540.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6479465 c.167-1G>T p.? Chr.13: 24318817-24318817 on Build GRCh38
3467802 c.221G>A p.G74E Chr.13: 24318872-24318872 on Build GRCh38
6875932 c.196A>G p.K66E Chr.13: 24318847-24318847 on Build GRCh38
469260 c.221G>T p.G74V Chr.13: 24318872-24318872 on Build GRCh38
1513919 c.227G>A p.R76Q Chr.13: 24318878-24318878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749817437 Chr.13: 24318794-24318794 on Build GRCh38
rs9580937 Chr.13: 24318585-24318585 on Build GRCh38
rs868823665 Chr.13: 24318616-24318616 on Build GRCh38
rs150781719 Chr.13: 24318829-24318829 on Build GRCh38
rs775479111 Chr.13: 24318901-24318901 on Build GRCh38
rs751174829 Chr.13: 24318833-24318833 on Build GRCh38
rs772248572 Chr.13: 24318803-24318803 on Build GRCh38
rs537026232 Chr.13: 24318711-24318711 on Build GRCh38
rs549791788 Chr.13: 24318921-24318921 on Build GRCh38
rs3979308 Chr.13: 24318735-24318736 on Build GRCh38
rs761867840 Chr.13: 24318802-24318802 on Build GRCh38
rs746288436 Chr.13: 24318787-24318787 on Build GRCh38
rs773114158 Chr.13: 24318884-24318884 on Build GRCh38
rs1031612647 Chr.13: 24318942-24318942 on Build GRCh38
rs762832591 Chr.13: 24318906-24318906 on Build GRCh38
rs3864972 Chr.13: 24318682-24318682 on Build GRCh38
rs552085577 Chr.13: 24318625-24318625 on Build GRCh38
rs149288885 Chr.13: 24318830-24318830 on Build GRCh38
rs756913251 Chr.13: 24318767-24318767 on Build GRCh38
rs529734583 Chr.13: 24318586-24318586 on Build GRCh38
rs866193421 Chr.13: 24318617-24318617 on Build GRCh38
rs780428044 Chr.13: 24318857-24318857 on Build GRCh38
rs779386798 Chr.13: 24318878-24318878 on Build GRCh38
rs755673923 Chr.13: 24318849-24318849 on Build GRCh38
rs759260616 Chr.13: 24318881-24318882 on Build GRCh38
rs574466038 Chr.13: 24318678-24318678 on Build GRCh38
rs748534982 Chr.13: 24318881-24318881 on Build GRCh38
rs2148236 Chr.13: 24318524-24318524 on Build GRCh38
rs534575877 Chr.13: 24318631-24318631 on Build GRCh38
rs941164539 Chr.13: 24318811-24318811 on Build GRCh38
rs760985100 Chr.13: 24318885-24318885 on Build GRCh38
rs185437549 Chr.13: 24318643-24318643 on Build GRCh38
rs764196906 Chr.13: 24318916-24318916 on Build GRCh38
rs573399680 Chr.13: 24318812-24318812 on Build GRCh38
rs546281490 Chr.13: 24318801-24318801 on Build GRCh38
rs780226763 Chr.13: 24318793-24318793 on Build GRCh38
rs369453137 Chr.13: 24318888-24318888 on Build GRCh38
rs749630211 Chr.13: 24318858-24318858 on Build GRCh38
rs181829504 Chr.13: 24318804-24318804 on Build GRCh38
rs752497059 Chr.13: 24318823-24318823 on Build GRCh38
rs769112380 Chr.13: 24318799-24318799 on Build GRCh38
rs997580486 Chr.13: 24318900-24318900 on Build GRCh38
rs9580939 Chr.13: 24318703-24318703 on Build GRCh38
rs980781461 Chr.13: 24318695-24318695 on Build GRCh38
rs9580938 Chr.13: 24318630-24318630 on Build GRCh38
rs375254759 Chr.13: 24318791-24318791 on Build GRCh38
rs144539792 Chr.13: 24318842-24318842 on Build GRCh38
rs939319205 Chr.13: 24318712-24318712 on Build GRCh38
rs765098049 Chr.13: 24318813-24318813 on Build GRCh38
rs569749993 Chr.13: 24318595-24318595 on Build GRCh38
rs761599171 Chr.13: 24318922-24318922 on Build GRCh38
rs370601883 Chr.13: 24318629-24318629 on Build GRCh38
rs146063067 Chr.13: 24318588-24318588 on Build GRCh38
rs567115914 Chr.13: 24318579-24318579 on Build GRCh38
rs142688497 Chr.13: 24318877-24318877 on Build GRCh38
rs955929526 Chr.13: 24318607-24318607 on Build GRCh38
rs556970693 Chr.13: 24318737-24318737 on Build GRCh38
rs77318507 Chr.13: 24318624-24318624 on Build GRCh38
rs188339290 Chr.13: 24318780-24318780 on Build GRCh38
rs968110242 Chr.13: 24318641-24318641 on Build GRCh38
rs915359478 Chr.13: 24318639-24318639 on Build GRCh38
rs1031979905 Chr.13: 24318606-24318606 on Build GRCh38
rs997463442 Chr.13: 24318596-24318596 on Build GRCh38
rs756789682 Chr.13: 24318837-24318837 on Build GRCh38
rs78731266 Chr.13: 24318843-24318843 on Build GRCh38
rs549828793 Chr.13: 24318592-24318592 on Build GRCh38

View Full Product Details