Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30735367-30735875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX5AP
Entrez Gene ID
241
Gene Name
arachidonate 5-lipoxygenase activating protein
Gene Aliases
FLAP
UniGene
Hs.507658
Gene Chromosome Location
Chr.13: 30713478-30764428 on Build GRCh38
Transcripts
NM_001204406.1 NM_001629.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2070848 c.58G>A p.V20M Chr.13: 30735663-30735663 on Build GRCh38
2070849 c.70+2T>C p.? Chr.13: 30735677-30735677 on Build GRCh38
6553875 c.70G>T p.G24* Chr.13: 30735675-30735675 on Build GRCh38
7321383 c.57C>T p.S19S Chr.13: 30735662-30735662 on Build GRCh38
4479795 c.64C>T p.Q22* Chr.13: 30735669-30735669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201360548 Chr.13: 30735713-30735713 on Build GRCh38
rs11542984 Chr.13: 30735647-30735647 on Build GRCh38
rs1018242116 Chr.13: 30735473-30735473 on Build GRCh38
rs78247371 Chr.13: 30735791-30735791 on Build GRCh38
rs200150324 Chr.13: 30735491-30735491 on Build GRCh38
rs767566357 Chr.13: 30735572-30735572 on Build GRCh38
rs976784450 Chr.13: 30735499-30735499 on Build GRCh38
rs771186169 Chr.13: 30735539-30735539 on Build GRCh38
rs965380234 Chr.13: 30735767-30735767 on Build GRCh38
rs187127207 Chr.13: 30735779-30735779 on Build GRCh38
rs887971906 Chr.13: 30735437-30735437 on Build GRCh38
rs1049113284 Chr.13: 30735710-30735710 on Build GRCh38
rs572943218 Chr.13: 30735469-30735469 on Build GRCh38
rs200634958 Chr.13: 30735704-30735704 on Build GRCh38
rs775067752 Chr.13: 30735436-30735441 on Build GRCh38
rs766297631 Chr.13: 30735686-30735686 on Build GRCh38
rs887878875 Chr.13: 30735725-30735725 on Build GRCh38
rs966324098 Chr.13: 30735494-30735494 on Build GRCh38
rs112066079 Chr.13: 30735424-30735424 on Build GRCh38
rs374960174 Chr.13: 30735521-30735521 on Build GRCh38
rs770152662 Chr.13: 30735648-30735648 on Build GRCh38
rs935804062 Chr.13: 30735635-30735635 on Build GRCh38
rs967111169 Chr.13: 30735527-30735527 on Build GRCh38
rs775787793 Chr.13: 30735658-30735658 on Build GRCh38
rs931056338 Chr.13: 30735795-30735795 on Build GRCh38
rs757726001 Chr.13: 30735723-30735723 on Build GRCh38
rs200167676 Chr.13: 30735685-30735685 on Build GRCh38
rs1039204637 Chr.13: 30735770-30735770 on Build GRCh38
rs775732668 Chr.13: 30735564-30735564 on Build GRCh38
rs61105798 Chr.13: 30735459-30735459 on Build GRCh38
rs1007747289 Chr.13: 30735459-30735459 on Build GRCh38
rs199916092 Chr.13: 30735641-30735641 on Build GRCh38
rs368028891 Chr.13: 30735547-30735547 on Build GRCh38
rs150121845 Chr.13: 30735611-30735611 on Build GRCh38
rs78236471 Chr.13: 30735566-30735566 on Build GRCh38
rs61947401 Chr.13: 30735667-30735667 on Build GRCh38
rs199498867 Chr.13: 30735483-30735483 on Build GRCh38
rs776014652 Chr.13: 30735622-30735622 on Build GRCh38
rs368925794 Chr.13: 30735625-30735625 on Build GRCh38
rs754612224 Chr.13: 30735600-30735600 on Build GRCh38
rs988441126 Chr.13: 30735705-30735705 on Build GRCh38
rs776963148 Chr.13: 30735541-30735541 on Build GRCh38
rs773275143 Chr.13: 30735574-30735574 on Build GRCh38
rs747451651 Chr.13: 30735644-30735644 on Build GRCh38
rs565035189 Chr.13: 30735571-30735571 on Build GRCh38
rs768839702 Chr.13: 30735663-30735663 on Build GRCh38
rs537023446 Chr.13: 30735435-30735440 on Build GRCh38
rs201514687 Chr.13: 30735489-30735489 on Build GRCh38
rs746047394 Chr.13: 30735548-30735548 on Build GRCh38
rs1032729055 Chr.13: 30735839-30735839 on Build GRCh38
rs371498736 Chr.13: 30735721-30735721 on Build GRCh38
rs764640364 Chr.13: 30735822-30735822 on Build GRCh38
rs150273767 Chr.13: 30735662-30735662 on Build GRCh38
rs528561957 Chr.13: 30735771-30735771 on Build GRCh38
rs747442077 Chr.13: 30735523-30735523 on Build GRCh38
rs764814001 Chr.13: 30735609-30735609 on Build GRCh38
rs17245407 Chr.13: 30735582-30735582 on Build GRCh38
rs76698472 Chr.13: 30735439-30735439 on Build GRCh38
rs200553849 Chr.13: 30735484-30735484 on Build GRCh38
rs544491297 Chr.13: 30735584-30735584 on Build GRCh38
rs114001301 Chr.13: 30735703-30735703 on Build GRCh38
rs759403271 Chr.13: 30735567-30735567 on Build GRCh38
rs1008103835 Chr.13: 30735728-30735728 on Build GRCh38
rs760495680 Chr.13: 30735668-30735668 on Build GRCh38
rs749701446 Chr.13: 30735440-30735441 on Build GRCh38
rs878880111 Chr.13: 30735698-30735698 on Build GRCh38
rs776348087 Chr.13: 30735435-30735441 on Build GRCh38
rs4769055 Chr.13: 30735693-30735693 on Build GRCh38
rs950036727 Chr.13: 30735680-30735680 on Build GRCh38
rs757903139 Chr.13: 30735630-30735630 on Build GRCh38
rs778311127 Chr.13: 30735636-30735636 on Build GRCh38
rs1045702879 Chr.13: 30735687-30735687 on Build GRCh38
rs922128732 Chr.13: 30735440-30735440 on Build GRCh38
rs1029371704 Chr.13: 30735508-30735508 on Build GRCh38
rs781681329 Chr.13: 30735727-30735727 on Build GRCh38
rs59918130 Chr.13: 30735454-30735459 on Build GRCh38
rs1049216826 Chr.13: 30735488-30735488 on Build GRCh38
rs967058992 Chr.13: 30735801-30735801 on Build GRCh38
rs757681724 Chr.13: 30735645-30735645 on Build GRCh38
rs770017088 Chr.13: 30735552-30735552 on Build GRCh38
rs763565016 Chr.13: 30735715-30735715 on Build GRCh38

View Full Product Details