Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30755866-30756375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX5AP
Entrez Gene ID
241
Gene Name
arachidonate 5-lipoxygenase activating protein
Gene Aliases
FLAP
UniGene
Hs.507658
Gene Chromosome Location
Chr.13: 30713478-30764428 on Build GRCh38
Transcripts
NM_001204406.1 NM_001629.3 XM_017020522.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3955524 c.309A>T p.G103G Chr.13: 30756011-30756011 on Build GRCh38
6463150 c.294T>G p.F98L Chr.13: 30755996-30755996 on Build GRCh38
1182585 c.275T>G p.F92C Chr.13: 30755977-30755977 on Build GRCh38
3737446 c.299G>A p.G100D Chr.13: 30756001-30756001 on Build GRCh38
1493106 c.297C>T p.V99V Chr.13: 30755999-30755999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036271594 Chr.13: 30756205-30756205 on Build GRCh38
rs539558967 Chr.13: 30756165-30756165 on Build GRCh38
rs771766962 Chr.13: 30755944-30755944 on Build GRCh38
rs772996992 Chr.13: 30755953-30755953 on Build GRCh38
rs865790666 Chr.13: 30756242-30756242 on Build GRCh38
rs780022871 Chr.13: 30755909-30755909 on Build GRCh38
rs200702316 Chr.13: 30756024-30756024 on Build GRCh38
rs201756102 Chr.13: 30755978-30755978 on Build GRCh38
rs760124781 Chr.13: 30756019-30756019 on Build GRCh38
rs147288214 Chr.13: 30756327-30756327 on Build GRCh38
rs867960774 Chr.13: 30756287-30756287 on Build GRCh38
rs988393834 Chr.13: 30755911-30755911 on Build GRCh38
rs763163243 Chr.13: 30756034-30756034 on Build GRCh38
rs990805616 Chr.13: 30756298-30756298 on Build GRCh38
rs536667172 Chr.13: 30756196-30756196 on Build GRCh38
rs1029023570 Chr.13: 30756241-30756241 on Build GRCh38
rs915314110 Chr.13: 30755920-30755920 on Build GRCh38
rs199776317 Chr.13: 30755940-30755940 on Build GRCh38
rs889474042 Chr.13: 30756135-30756135 on Build GRCh38
rs968352868 Chr.13: 30756297-30756297 on Build GRCh38
rs917082408 Chr.13: 30756347-30756347 on Build GRCh38
rs978452437 Chr.13: 30756310-30756310 on Build GRCh38
rs199890484 Chr.13: 30756059-30756059 on Build GRCh38
rs754765456 Chr.13: 30755936-30755936 on Build GRCh38
rs759671113 Chr.13: 30756335-30756335 on Build GRCh38
rs202205227 Chr.13: 30756009-30756009 on Build GRCh38
rs898227740 Chr.13: 30756200-30756200 on Build GRCh38
rs770543301 Chr.13: 30755976-30755976 on Build GRCh38
rs1057285845 Chr.13: 30756344-30756344 on Build GRCh38
rs144098425 Chr.13: 30755907-30755907 on Build GRCh38
rs912865300 Chr.13: 30755954-30755954 on Build GRCh38
rs574346713 Chr.13: 30756314-30756314 on Build GRCh38
rs994821578 Chr.13: 30756211-30756211 on Build GRCh38
rs568100159 Chr.13: 30756137-30756137 on Build GRCh38
rs201858343 Chr.13: 30756025-30756025 on Build GRCh38
rs754965545 Chr.13: 30756077-30756077 on Build GRCh38
rs12721458 Chr.13: 30756147-30756147 on Build GRCh38
rs550287447 Chr.13: 30756291-30756291 on Build GRCh38
rs4468448 Chr.13: 30756179-30756179 on Build GRCh38
rs1004241280 Chr.13: 30756285-30756285 on Build GRCh38
rs746577187 Chr.13: 30755960-30755960 on Build GRCh38
rs566207275 Chr.13: 30756156-30756156 on Build GRCh38
rs757323805 Chr.13: 30755908-30755908 on Build GRCh38
rs978166084 Chr.13: 30756345-30756345 on Build GRCh38
rs957098915 Chr.13: 30756271-30756271 on Build GRCh38
rs775940117 Chr.13: 30756029-30756029 on Build GRCh38
rs200311897 Chr.13: 30755904-30755904 on Build GRCh38
rs750462335 Chr.13: 30756076-30756076 on Build GRCh38
rs774511865 Chr.13: 30756304-30756304 on Build GRCh38
rs200532730 Chr.13: 30755947-30755947 on Build GRCh38
rs1042208470 Chr.13: 30756069-30756069 on Build GRCh38
rs899170410 Chr.13: 30756208-30756208 on Build GRCh38
rs747845467 Chr.13: 30755941-30755941 on Build GRCh38
rs17245589 Chr.13: 30756257-30756257 on Build GRCh38
rs994967904 Chr.13: 30756214-30756214 on Build GRCh38
rs576725278 Chr.13: 30756245-30756245 on Build GRCh38
rs750857122 Chr.13: 30756142-30756142 on Build GRCh38
rs547572854 Chr.13: 30756154-30756154 on Build GRCh38
rs1026406870 Chr.13: 30756244-30756244 on Build GRCh38
rs933930871 Chr.13: 30756107-30756107 on Build GRCh38
rs370359839 Chr.13: 30756109-30756109 on Build GRCh38
rs770172373 Chr.13: 30756055-30756055 on Build GRCh38
rs767549630 Chr.13: 30756060-30756060 on Build GRCh38
rs777318426 Chr.13: 30755983-30755983 on Build GRCh38
rs4075252 Chr.13: 30756233-30756233 on Build GRCh38
rs764364129 Chr.13: 30756045-30756045 on Build GRCh38
rs747752581 Chr.13: 30756183-30756183 on Build GRCh38
rs778796663 Chr.13: 30755939-30755939 on Build GRCh38
rs201614405 Chr.13: 30755922-30755922 on Build GRCh38
rs12721460 Chr.13: 30756124-30756124 on Build GRCh38
rs200075156 Chr.13: 30756022-30756022 on Build GRCh38

View Full Product Details