Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40932941-40933475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELF1
Entrez Gene ID
1997
Gene Name
E74 like ETS transcription factor 1
Gene Aliases
-
UniGene
Hs.135646
Gene Chromosome Location
Chr.13: 40931919-41061414 on Build GRCh38
Transcripts
NM_001145353.1 NM_172373.3 XM_005266276.1 XM_005266277.3 XM_011534950.1 XM_011534951.2 XM_017020409.1 XM_017020410.1 XM_017020411.1 XM_017020412.1 XM_017020413.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
122816 c.1811C>G p.S604C Chr.13: 40933474-40933474 on Build GRCh38
4680725 c.1817T>A p.V606E Chr.13: 40933468-40933468 on Build GRCh38
3417590 c.1834G>A p.E612K Chr.13: 40933451-40933451 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756814760 Chr.13: 40933152-40933162 on Build GRCh38
rs531424503 Chr.13: 40933421-40933421 on Build GRCh38
rs147878914 Chr.13: 40933451-40933451 on Build GRCh38
rs867050001 Chr.13: 40933442-40933442 on Build GRCh38
rs916065017 Chr.13: 40933226-40933226 on Build GRCh38
rs183456530 Chr.13: 40933228-40933228 on Build GRCh38
rs983374531 Chr.13: 40933368-40933369 on Build GRCh38
rs925043118 Chr.13: 40933395-40933395 on Build GRCh38
rs140656224 Chr.13: 40933449-40933449 on Build GRCh38
rs960292615 Chr.13: 40933164-40933164 on Build GRCh38
rs190075209 Chr.13: 40933029-40933029 on Build GRCh38
rs141938160 Chr.13: 40933122-40933122 on Build GRCh38
rs888968483 Chr.13: 40933061-40933061 on Build GRCh38
rs944732345 Chr.13: 40933034-40933034 on Build GRCh38
rs1052070375 Chr.13: 40933438-40933438 on Build GRCh38
rs555326637 Chr.13: 40933022-40933022 on Build GRCh38
rs761186414 Chr.13: 40933024-40933024 on Build GRCh38
rs980540081 Chr.13: 40933229-40933229 on Build GRCh38
rs936304799 Chr.13: 40933399-40933399 on Build GRCh38
rs1007639283 Chr.13: 40933080-40933080 on Build GRCh38
rs963339136 Chr.13: 40933092-40933092 on Build GRCh38
rs115657038 Chr.13: 40933443-40933443 on Build GRCh38
rs1031701563 Chr.13: 40933166-40933166 on Build GRCh38
rs1027509330 Chr.13: 40933245-40933245 on Build GRCh38
rs540906731 Chr.13: 40933085-40933085 on Build GRCh38
rs576770565 Chr.13: 40933149-40933149 on Build GRCh38
rs747430985 Chr.13: 40933356-40933359 on Build GRCh38
rs904072918 Chr.13: 40933134-40933138 on Build GRCh38
rs932142008 Chr.13: 40933114-40933114 on Build GRCh38
rs749050855 Chr.13: 40933391-40933394 on Build GRCh38
rs776243721 Chr.13: 40933386-40933386 on Build GRCh38
rs769555380 Chr.13: 40933437-40933437 on Build GRCh38
rs750580222 Chr.13: 40933377-40933377 on Build GRCh38
rs759534401 Chr.13: 40933416-40933416 on Build GRCh38
rs75810168 Chr.13: 40933225-40933225 on Build GRCh38
rs1051589905 Chr.13: 40933038-40933038 on Build GRCh38
rs1016794482 Chr.13: 40933091-40933091 on Build GRCh38
rs907882918 Chr.13: 40933032-40933035 on Build GRCh38
rs368129847 Chr.13: 40933139-40933143 on Build GRCh38
rs892387336 Chr.13: 40932998-40932998 on Build GRCh38
rs900622987 Chr.13: 40933097-40933097 on Build GRCh38
rs763822922 Chr.13: 40933031-40933031 on Build GRCh38
rs555153796 Chr.13: 40932973-40932973 on Build GRCh38
rs951914605 Chr.13: 40933327-40933327 on Build GRCh38
rs891444075 Chr.13: 40933191-40933191 on Build GRCh38
rs144467943 Chr.13: 40933450-40933450 on Build GRCh38

View Full Product Details