Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 42285900-42286409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP11
Entrez Gene ID
11215
Gene Name
A-kinase anchoring protein 11
Gene Aliases
AKAP-11, AKAP220, PPP1R44, PRKA11
UniGene
Hs.105105
Gene Chromosome Location
Chr.13: 42271470-42323267 on Build GRCh38
Transcripts
NM_016248.3 XM_005266247.1 XM_005266248.3 XM_005266249.3 XM_005266250.3 XM_011534903.2 XM_011534904.1 XM_011534905.1 XM_011534906.2 XM_017020381.1 XM_017020382.1 XM_017020383.1

SNP Detail

SNP ID Chromosome Location
rs544208884 Chr.13: 42286323-42286323 on Build GRCh38
rs188937716 Chr.13: 42286223-42286223 on Build GRCh38
rs1004902774 Chr.13: 42286204-42286204 on Build GRCh38
rs751594294 Chr.13: 42286188-42286188 on Build GRCh38
rs953836913 Chr.13: 42285945-42285945 on Build GRCh38
rs1025116444 Chr.13: 42286302-42286302 on Build GRCh38
rs778734966 Chr.13: 42286300-42286300 on Build GRCh38
rs1045920644 Chr.13: 42286160-42286160 on Build GRCh38
rs963004131 Chr.13: 42285950-42285950 on Build GRCh38
rs1052381979 Chr.13: 42286075-42286075 on Build GRCh38
rs905939198 Chr.13: 42286161-42286161 on Build GRCh38
rs750835442 Chr.13: 42286383-42286383 on Build GRCh38
rs936585359 Chr.13: 42286074-42286074 on Build GRCh38
rs898681565 Chr.13: 42286164-42286164 on Build GRCh38
rs958147002 Chr.13: 42286346-42286346 on Build GRCh38
rs995555273 Chr.13: 42285968-42285968 on Build GRCh38
rs763158492 Chr.13: 42286345-42286345 on Build GRCh38
rs570922168 Chr.13: 42286007-42286007 on Build GRCh38
rs925169858 Chr.13: 42286072-42286072 on Build GRCh38
rs764280252 Chr.13: 42286303-42286311 on Build GRCh38
rs1039889937 Chr.13: 42286162-42286162 on Build GRCh38
rs891216780 Chr.13: 42286133-42286133 on Build GRCh38
rs374378036 Chr.13: 42286305-42286305 on Build GRCh38
rs925587732 Chr.13: 42285999-42285999 on Build GRCh38
rs150281985 Chr.13: 42286038-42286038 on Build GRCh38
rs1051143704 Chr.13: 42286124-42286124 on Build GRCh38
rs560459823 Chr.13: 42286160-42286161 on Build GRCh38
rs546650281 Chr.13: 42286158-42286158 on Build GRCh38
rs946072210 Chr.13: 42286100-42286100 on Build GRCh38
rs754118738 Chr.13: 42286337-42286338 on Build GRCh38
rs773122535 Chr.13: 42286321-42286321 on Build GRCh38
rs775683791 Chr.13: 42286343-42286343 on Build GRCh38
rs913230606 Chr.13: 42286089-42286089 on Build GRCh38
rs986809960 Chr.13: 42286380-42286380 on Build GRCh38
rs764614672 Chr.13: 42286349-42286349 on Build GRCh38
rs150549828 Chr.13: 42286375-42286375 on Build GRCh38
rs1025244786 Chr.13: 42285986-42285986 on Build GRCh38
rs980644446 Chr.13: 42286064-42286064 on Build GRCh38
rs561013659 Chr.13: 42286333-42286333 on Build GRCh38
rs111907616 Chr.13: 42286099-42286099 on Build GRCh38
rs1025886501 Chr.13: 42286207-42286207 on Build GRCh38
rs887756069 Chr.13: 42286191-42286191 on Build GRCh38
rs41288307 Chr.13: 42286246-42286246 on Build GRCh38
rs917705186 Chr.13: 42286016-42286016 on Build GRCh38
rs530157653 Chr.13: 42286361-42286361 on Build GRCh38
rs771764292 Chr.13: 42286309-42286309 on Build GRCh38
rs1015915550 Chr.13: 42286256-42286256 on Build GRCh38
rs1029532476 Chr.13: 42286319-42286319 on Build GRCh38
rs988056838 Chr.13: 42286356-42286356 on Build GRCh38
rs991861544 Chr.13: 42286013-42286013 on Build GRCh38
rs377672654 Chr.13: 42286354-42286354 on Build GRCh38
rs980954260 Chr.13: 42286336-42286336 on Build GRCh38
rs985084714 Chr.13: 42285967-42285968 on Build GRCh38
rs945993917 Chr.13: 42286373-42286373 on Build GRCh38
rs749339314 Chr.13: 42286229-42286229 on Build GRCh38
rs1013213596 Chr.13: 42286247-42286247 on Build GRCh38
rs368488446 Chr.13: 42286168-42286169 on Build GRCh38

View Full Product Details