Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45200735-45201249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2F2
Entrez Gene ID
2963
Gene Name
general transcription factor IIF subunit 2
Gene Aliases
BTF4, RAP30, TF2F2, TFIIF
UniGene
Hs.654582
Gene Chromosome Location
Chr.13: 45119830-45284105 on Build GRCh38
Transcripts
NM_004128.2 XM_011535052.2 XM_017020551.1 NM_198404.2

Gene Symbol
KCTD4
Entrez Gene ID
386618
Gene Name
potassium channel tetramerization domain containing 4
Gene Aliases
bA321C24.3
UniGene
Hs.23406
Gene Chromosome Location
Chr.13: 45192853-45201040 on Build GRCh38
Transcripts
NM_004128.2 XM_011535052.2 XM_017020551.1 NM_198404.2

SNP Detail

SNP ID Chromosome Location
rs531098213 Chr.13: 45200978-45200978 on Build GRCh38
rs781508737 Chr.13: 45200885-45200885 on Build GRCh38
rs187620567 Chr.13: 45200923-45200923 on Build GRCh38
rs555994208 Chr.13: 45200841-45200841 on Build GRCh38
rs534437287 Chr.13: 45200838-45200838 on Build GRCh38
rs779812964 Chr.13: 45201092-45201092 on Build GRCh38
rs1036305481 Chr.13: 45200803-45200807 on Build GRCh38
rs574477659 Chr.13: 45200897-45200897 on Build GRCh38
rs551286676 Chr.13: 45201105-45201105 on Build GRCh38
rs757503867 Chr.13: 45200853-45200854 on Build GRCh38
rs899427762 Chr.13: 45200772-45200772 on Build GRCh38
rs546435059 Chr.13: 45201200-45201200 on Build GRCh38
rs575148832 Chr.13: 45200971-45200971 on Build GRCh38
rs909020668 Chr.13: 45201048-45201048 on Build GRCh38
rs897495814 Chr.13: 45200809-45200809 on Build GRCh38
rs953272369 Chr.13: 45201061-45201061 on Build GRCh38
rs941915875 Chr.13: 45201104-45201104 on Build GRCh38
rs1032909713 Chr.13: 45200857-45200857 on Build GRCh38
rs1056347411 Chr.13: 45200826-45200826 on Build GRCh38
rs1029963822 Chr.13: 45200840-45200840 on Build GRCh38
rs920280450 Chr.13: 45201219-45201219 on Build GRCh38
rs140545162 Chr.13: 45201202-45201202 on Build GRCh38
rs911651015 Chr.13: 45201122-45201122 on Build GRCh38
rs77657704 Chr.13: 45200984-45200984 on Build GRCh38
rs528359105 Chr.13: 45201178-45201178 on Build GRCh38
rs1006539141 Chr.13: 45200800-45200800 on Build GRCh38
rs563998138 Chr.13: 45201034-45201034 on Build GRCh38
rs888808927 Chr.13: 45201188-45201188 on Build GRCh38
rs1024430529 Chr.13: 45200876-45200876 on Build GRCh38
rs763643148 Chr.13: 45201100-45201100 on Build GRCh38
rs1029231996 Chr.13: 45201032-45201032 on Build GRCh38
rs958252422 Chr.13: 45200860-45200860 on Build GRCh38
rs529400016 Chr.13: 45201220-45201220 on Build GRCh38
rs930802846 Chr.13: 45200793-45200793 on Build GRCh38
rs753252898 Chr.13: 45200918-45200918 on Build GRCh38
rs745388820 Chr.13: 45200921-45200921 on Build GRCh38
rs1014743885 Chr.13: 45200870-45200870 on Build GRCh38
rs903154988 Chr.13: 45200877-45200877 on Build GRCh38
rs997392133 Chr.13: 45201121-45201121 on Build GRCh38
rs182085758 Chr.13: 45200920-45200920 on Build GRCh38
rs987624708 Chr.13: 45201107-45201107 on Build GRCh38
rs997174492 Chr.13: 45200828-45200828 on Build GRCh38

View Full Product Details