Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45464785-45465312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG3
Entrez Gene ID
83548
Gene Name
component of oligomeric golgi complex 3
Gene Aliases
SEC34
UniGene
Hs.507948
Gene Chromosome Location
Chr.13: 45464895-45536698 on Build GRCh38
Transcripts
NM_031431.3 XM_011535266.1 XM_011535267.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
947647 c.153G>T p.E51D Chr.13: 45465189-45465189 on Build GRCh38
5842571 c.166C>T p.P56S Chr.13: 45465202-45465202 on Build GRCh38
5842570 c.148G>A p.A50T Chr.13: 45465184-45465184 on Build GRCh38
5648466 c.113G>A p.R38K Chr.13: 45465149-45465149 on Build GRCh38
1300214 c.172G>A p.E58K Chr.13: 45465208-45465208 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs542249909 Chr.13: 45464968-45464968 on Build GRCh38
rs757274309 Chr.13: 45465116-45465116 on Build GRCh38
rs981366149 Chr.13: 45464949-45464949 on Build GRCh38
rs755933277 Chr.13: 45465145-45465145 on Build GRCh38
rs34678243 Chr.13: 45465018-45465018 on Build GRCh38
rs748184991 Chr.13: 45465027-45465027 on Build GRCh38
rs772040389 Chr.13: 45465162-45465162 on Build GRCh38
rs559024391 Chr.13: 45465233-45465233 on Build GRCh38
rs774381979 Chr.13: 45465002-45465002 on Build GRCh38
rs1032823181 Chr.13: 45464891-45464891 on Build GRCh38
rs974350205 Chr.13: 45464980-45464980 on Build GRCh38
rs941797147 Chr.13: 45465022-45465022 on Build GRCh38
rs988111111 Chr.13: 45464930-45464930 on Build GRCh38
rs144983754 Chr.13: 45465206-45465206 on Build GRCh38
rs773183513 Chr.13: 45465236-45465239 on Build GRCh38
rs1001013085 Chr.13: 45464821-45464821 on Build GRCh38
rs374186273 Chr.13: 45465232-45465232 on Build GRCh38
rs1006280553 Chr.13: 45464808-45464808 on Build GRCh38
rs770894032 Chr.13: 45464989-45464989 on Build GRCh38
rs373457132 Chr.13: 45465131-45465131 on Build GRCh38
rs141222488 Chr.13: 45465155-45465155 on Build GRCh38
rs766640714 Chr.13: 45465047-45465047 on Build GRCh38
rs557353471 Chr.13: 45464988-45464988 on Build GRCh38
rs945910145 Chr.13: 45464942-45464942 on Build GRCh38
rs777687273 Chr.13: 45465260-45465260 on Build GRCh38
rs752423674 Chr.13: 45465097-45465097 on Build GRCh38
rs760744868 Chr.13: 45465041-45465041 on Build GRCh38
rs902813181 Chr.13: 45465048-45465048 on Build GRCh38
rs376444139 Chr.13: 45465213-45465213 on Build GRCh38
rs919760306 Chr.13: 45464984-45464984 on Build GRCh38
rs779891760 Chr.13: 45465252-45465252 on Build GRCh38
rs758696792 Chr.13: 45465257-45465257 on Build GRCh38
rs755840663 Chr.13: 45465249-45465249 on Build GRCh38
rs749511290 Chr.13: 45464992-45464992 on Build GRCh38
rs961264961 Chr.13: 45464969-45464969 on Build GRCh38
rs777312080 Chr.13: 45465197-45465197 on Build GRCh38
rs377320049 Chr.13: 45465030-45465030 on Build GRCh38
rs574959391 Chr.13: 45464945-45464945 on Build GRCh38
rs991700953 Chr.13: 45465286-45465286 on Build GRCh38
rs368743474 Chr.13: 45464842-45464842 on Build GRCh38
rs11619408 Chr.13: 45465272-45465272 on Build GRCh38
rs111818704 Chr.13: 45465091-45465091 on Build GRCh38
rs761623416 Chr.13: 45465240-45465240 on Build GRCh38
rs917639695 Chr.13: 45465006-45465006 on Build GRCh38
rs781104021 Chr.13: 45464996-45464996 on Build GRCh38
rs535153560 Chr.13: 45464872-45464872 on Build GRCh38
rs543312923 Chr.13: 45465178-45465178 on Build GRCh38
rs768914260 Chr.13: 45465001-45465001 on Build GRCh38
rs538009116 Chr.13: 45465068-45465069 on Build GRCh38
rs767448586 Chr.13: 45465246-45465246 on Build GRCh38
rs983421657 Chr.13: 45464903-45464903 on Build GRCh38
rs953679499 Chr.13: 45464878-45464879 on Build GRCh38
rs576050376 Chr.13: 45464997-45464997 on Build GRCh38
rs200180168 Chr.13: 45465071-45465071 on Build GRCh38
rs553207670 Chr.13: 45464928-45464928 on Build GRCh38
rs759235670 Chr.13: 45465077-45465077 on Build GRCh38
rs564733357 Chr.13: 45465082-45465082 on Build GRCh38
rs908487501 Chr.13: 45464932-45464932 on Build GRCh38
rs773525363 Chr.13: 45465036-45465036 on Build GRCh38
rs35739237 Chr.13: 45465282-45465283 on Build GRCh38
rs763660050 Chr.13: 45465114-45465114 on Build GRCh38
rs958177593 Chr.13: 45464921-45464921 on Build GRCh38
rs913925566 Chr.13: 45464936-45464936 on Build GRCh38
rs762490827 Chr.13: 45465099-45465099 on Build GRCh38
rs986356560 Chr.13: 45464886-45464886 on Build GRCh38
rs747074738 Chr.13: 45465262-45465262 on Build GRCh38
rs750382407 Chr.13: 45465248-45465248 on Build GRCh38
rs529699854 Chr.13: 45465236-45465236 on Build GRCh38
rs747451750 Chr.13: 45465192-45465192 on Build GRCh38
rs900671515 Chr.13: 45465136-45465136 on Build GRCh38
rs996283834 Chr.13: 45464861-45464861 on Build GRCh38
rs759935145 Chr.13: 45465199-45465199 on Build GRCh38
rs771511545 Chr.13: 45465196-45465196 on Build GRCh38
rs1030792849 Chr.13: 45464867-45464867 on Build GRCh38
rs780283026 Chr.13: 45464865-45464865 on Build GRCh38
rs371280736 Chr.13: 45465230-45465230 on Build GRCh38
rs558763575 Chr.13: 45464845-45464845 on Build GRCh38
rs997741572 Chr.13: 45464866-45464866 on Build GRCh38
rs1042019094 Chr.13: 45465042-45465042 on Build GRCh38
rs958342295 Chr.13: 45465106-45465106 on Build GRCh38
rs751444143 Chr.13: 45465115-45465115 on Build GRCh38
rs1030432576 Chr.13: 45465161-45465161 on Build GRCh38
rs764947212 Chr.13: 45465085-45465085 on Build GRCh38
rs570532158 Chr.13: 45464851-45464851 on Build GRCh38
rs777968918 Chr.13: 45465169-45465169 on Build GRCh38
rs528692223 Chr.13: 45465143-45465143 on Build GRCh38
rs1054413377 Chr.13: 45465119-45465119 on Build GRCh38
rs12585128 Chr.13: 45464820-45464820 on Build GRCh38
rs762230625 Chr.13: 45465043-45465066 on Build GRCh38
rs751128641 Chr.13: 45465237-45465237 on Build GRCh38
rs890530074 Chr.13: 45465207-45465207 on Build GRCh38
rs965191017 Chr.13: 45464853-45464853 on Build GRCh38
rs997687835 Chr.13: 45465065-45465065 on Build GRCh38

View Full Product Details