Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45579841-45580346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH6B
Entrez Gene ID
220081
Gene Name
glutamate rich 6B
Gene Aliases
FAM194B
UniGene
Hs.668747
Gene Chromosome Location
Chr.13: 45541203-45615739 on Build GRCh38
Transcripts
NM_182542.2 XM_011534965.2 XM_017020418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5777192 c.961+3G>A p.? Chr.13: 45579930-45579930 on Build GRCh38
1367026 c.950_951insAA p.E318fs*18 Chr.13: 45579943-45579944 on Build GRCh38
1367027 c.950delA p.K317fs*18 Chr.13: 45579944-45579944 on Build GRCh38
3813878 c.920-1G>C p.? Chr.13: 45579975-45579975 on Build GRCh38
4455916 c.954A>G p.E318E Chr.13: 45579940-45579940 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs548563948 Chr.13: 45579937-45579937 on Build GRCh38
rs970621720 Chr.13: 45579868-45579868 on Build GRCh38
rs925136615 Chr.13: 45580263-45580263 on Build GRCh38
rs145351378 Chr.13: 45580295-45580295 on Build GRCh38
rs370748043 Chr.13: 45579998-45580000 on Build GRCh38
rs534390154 Chr.13: 45580283-45580283 on Build GRCh38
rs894909509 Chr.13: 45580130-45580130 on Build GRCh38
rs771750644 Chr.13: 45580010-45580010 on Build GRCh38
rs976725210 Chr.13: 45580160-45580160 on Build GRCh38
rs757735177 Chr.13: 45579957-45579958 on Build GRCh38
rs766076518 Chr.13: 45580237-45580237 on Build GRCh38
rs921265927 Chr.13: 45580161-45580161 on Build GRCh38
rs990603715 Chr.13: 45579984-45579984 on Build GRCh38
rs140020807 Chr.13: 45580006-45580006 on Build GRCh38
rs965466514 Chr.13: 45580150-45580150 on Build GRCh38
rs1048595161 Chr.13: 45580122-45580122 on Build GRCh38
rs1002873689 Chr.13: 45580003-45580003 on Build GRCh38
rs763736466 Chr.13: 45580155-45580155 on Build GRCh38
rs999437846 Chr.13: 45580179-45580179 on Build GRCh38
rs530230002 Chr.13: 45580166-45580166 on Build GRCh38
rs765220425 Chr.13: 45579943-45579943 on Build GRCh38
rs986485235 Chr.13: 45580176-45580176 on Build GRCh38
rs944274470 Chr.13: 45580064-45580064 on Build GRCh38
rs746820539 Chr.13: 45580009-45580009 on Build GRCh38
rs754232917 Chr.13: 45579943-45579944 on Build GRCh38
rs760237923 Chr.13: 45580132-45580132 on Build GRCh38
rs753570957 Chr.13: 45580246-45580246 on Build GRCh38
rs752726152 Chr.13: 45579954-45579954 on Build GRCh38
rs912759046 Chr.13: 45580047-45580047 on Build GRCh38
rs764884012 Chr.13: 45580302-45580302 on Build GRCh38
rs985604288 Chr.13: 45580014-45580014 on Build GRCh38
rs1037325879 Chr.13: 45580070-45580070 on Build GRCh38
rs537501844 Chr.13: 45580106-45580106 on Build GRCh38
rs773450959 Chr.13: 45580067-45580069 on Build GRCh38
rs918240018 Chr.13: 45580098-45580098 on Build GRCh38
rs953027315 Chr.13: 45580013-45580013 on Build GRCh38
rs375257773 Chr.13: 45579875-45579875 on Build GRCh38
rs999468719 Chr.13: 45579882-45579882 on Build GRCh38
rs1046062139 Chr.13: 45579991-45579991 on Build GRCh38
rs985532252 Chr.13: 45580310-45580310 on Build GRCh38
rs375384371 Chr.13: 45579921-45579921 on Build GRCh38
rs571716510 Chr.13: 45580267-45580267 on Build GRCh38
rs904866584 Chr.13: 45580305-45580305 on Build GRCh38
rs750238376 Chr.13: 45580304-45580304 on Build GRCh38
rs868278813 Chr.13: 45579942-45579943 on Build GRCh38
rs758234414 Chr.13: 45579988-45579988 on Build GRCh38
rs532048982 Chr.13: 45580041-45580041 on Build GRCh38
rs34262705 Chr.13: 45579994-45579996 on Build GRCh38
rs761867147 Chr.13: 45580272-45580272 on Build GRCh38
rs11147979 Chr.13: 45580121-45580121 on Build GRCh38
rs547104026 Chr.13: 45580069-45580069 on Build GRCh38
rs531672332 Chr.13: 45579865-45579865 on Build GRCh38
rs548807800 Chr.13: 45580173-45580173 on Build GRCh38
rs914662937 Chr.13: 45579986-45579986 on Build GRCh38
rs1046246867 Chr.13: 45580158-45580158 on Build GRCh38
rs36104923 Chr.13: 45579871-45579872 on Build GRCh38

View Full Product Details