Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45580524-45581035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH6B
Entrez Gene ID
220081
Gene Name
glutamate rich 6B
Gene Aliases
FAM194B
UniGene
Hs.668747
Gene Chromosome Location
Chr.13: 45541203-45615739 on Build GRCh38
Transcripts
NM_182542.2 XM_011534965.2 XM_017020418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3999096 c.908T>C p.L303P Chr.13: 45580614-45580614 on Build GRCh38
5733119 c.902C>T p.T301M Chr.13: 45580620-45580620 on Build GRCh38
947664 c.896C>T p.T299I Chr.13: 45580626-45580626 on Build GRCh38
947665 c.878C>G p.S293* Chr.13: 45580644-45580644 on Build GRCh38
5913486 c.917A>T p.E306V Chr.13: 45580605-45580605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11618506 Chr.13: 45580614-45580614 on Build GRCh38
rs543331852 Chr.13: 45580562-45580562 on Build GRCh38
rs372374780 Chr.13: 45580720-45580720 on Build GRCh38
rs768676117 Chr.13: 45580607-45580607 on Build GRCh38
rs1009437741 Chr.13: 45580953-45580953 on Build GRCh38
rs749381750 Chr.13: 45580585-45580585 on Build GRCh38
rs947711990 Chr.13: 45580787-45580787 on Build GRCh38
rs909556189 Chr.13: 45580808-45580808 on Build GRCh38
rs576388246 Chr.13: 45580758-45580758 on Build GRCh38
rs1033034950 Chr.13: 45580712-45580712 on Build GRCh38
rs1037167987 Chr.13: 45580634-45580634 on Build GRCh38
rs182532598 Chr.13: 45580650-45580650 on Build GRCh38
rs970575021 Chr.13: 45580771-45580771 on Build GRCh38
rs1028847931 Chr.13: 45580682-45580682 on Build GRCh38
rs889847458 Chr.13: 45580903-45580903 on Build GRCh38
rs548475736 Chr.13: 45580779-45580779 on Build GRCh38
rs368539765 Chr.13: 45580632-45580632 on Build GRCh38
rs764690120 Chr.13: 45580697-45580697 on Build GRCh38
rs758476781 Chr.13: 45580579-45580580 on Build GRCh38
rs988027836 Chr.13: 45580746-45580746 on Build GRCh38
rs1023898731 Chr.13: 45580952-45580952 on Build GRCh38
rs893652380 Chr.13: 45580818-45580818 on Build GRCh38
rs536904900 Chr.13: 45580994-45580994 on Build GRCh38
rs1020737939 Chr.13: 45580752-45580752 on Build GRCh38
rs191440137 Chr.13: 45580877-45580877 on Build GRCh38
rs761044187 Chr.13: 45580649-45580649 on Build GRCh38
rs994684697 Chr.13: 45580922-45580922 on Build GRCh38
rs563929022 Chr.13: 45580786-45580786 on Build GRCh38
rs1054353691 Chr.13: 45580923-45580923 on Build GRCh38
rs372925977 Chr.13: 45580691-45580691 on Build GRCh38
rs149206878 Chr.13: 45580883-45580883 on Build GRCh38
rs1000290518 Chr.13: 45580904-45580904 on Build GRCh38
rs772600831 Chr.13: 45580620-45580620 on Build GRCh38
rs186493801 Chr.13: 45580803-45580803 on Build GRCh38
rs552915622 Chr.13: 45580806-45580806 on Build GRCh38
rs118125794 Chr.13: 45580692-45580692 on Build GRCh38
rs781268611 Chr.13: 45580602-45580602 on Build GRCh38
rs901374294 Chr.13: 45580642-45580642 on Build GRCh38
rs889821779 Chr.13: 45580575-45580575 on Build GRCh38
rs889778603 Chr.13: 45580685-45580685 on Build GRCh38
rs940246759 Chr.13: 45580848-45580848 on Build GRCh38
rs184625062 Chr.13: 45580882-45580882 on Build GRCh38
rs970522681 Chr.13: 45580985-45580985 on Build GRCh38
rs745937144 Chr.13: 45580573-45580573 on Build GRCh38
rs1011267261 Chr.13: 45580551-45580551 on Build GRCh38
rs946652078 Chr.13: 45580823-45580823 on Build GRCh38
rs1003763984 Chr.13: 45580991-45580991 on Build GRCh38
rs1050335781 Chr.13: 45580866-45580866 on Build GRCh38
rs755023372 Chr.13: 45580567-45580567 on Build GRCh38
rs957912325 Chr.13: 45580744-45580744 on Build GRCh38
rs776217367 Chr.13: 45580667-45580667 on Build GRCh38
rs7998038 Chr.13: 45580934-45580934 on Build GRCh38
rs1025810518 Chr.13: 45580930-45580930 on Build GRCh38
rs1052260943 Chr.13: 45580810-45580810 on Build GRCh38
rs780143661 Chr.13: 45580584-45580584 on Build GRCh38
rs545145359 Chr.13: 45580774-45580774 on Build GRCh38
rs576200240 Chr.13: 45580968-45580968 on Build GRCh38
rs927714079 Chr.13: 45580804-45580804 on Build GRCh38
rs375497408 Chr.13: 45580619-45580619 on Build GRCh38
rs375672149 Chr.13: 45580931-45580931 on Build GRCh38
rs568445226 Chr.13: 45580889-45580889 on Build GRCh38
rs7334362 Chr.13: 45580964-45580964 on Build GRCh38
rs981783876 Chr.13: 45581004-45581004 on Build GRCh38

View Full Product Details