Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 52460811-52461320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKAP2
Entrez Gene ID
26586
Gene Name
cytoskeleton associated protein 2
Gene Aliases
LB1, TMAP, se20-10
UniGene
Hs.444028
Gene Chromosome Location
Chr.13: 52455360-52476631 on Build GRCh38
Transcripts
NM_001098525.2 NM_001286686.1 NM_001286687.1 NM_018204.4 XM_005266344.4 XM_011535043.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4575982 c.412A>G p.S138G Chr.13: 52461238-52461238 on Build GRCh38
7361366 c.160G>T p.D54Y Chr.13: 52460903-52460903 on Build GRCh38
6629520 c.220A>G p.K74E Chr.13: 52460963-52460963 on Build GRCh38
3885429 c.209T>C p.V70A Chr.13: 52460952-52460952 on Build GRCh38
4617473 c.381G>A p.P127P Chr.13: 52461207-52461207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201217629 Chr.13: 52461250-52461250 on Build GRCh38
rs373545539 Chr.13: 52461064-52461064 on Build GRCh38
rs376367571 Chr.13: 52461093-52461093 on Build GRCh38
rs761382347 Chr.13: 52460938-52460938 on Build GRCh38
rs376601366 Chr.13: 52461034-52461034 on Build GRCh38
rs769763485 Chr.13: 52460999-52460999 on Build GRCh38
rs764723599 Chr.13: 52461066-52461066 on Build GRCh38
rs539183420 Chr.13: 52461074-52461074 on Build GRCh38
rs778665507 Chr.13: 52461287-52461287 on Build GRCh38
rs747228732 Chr.13: 52460852-52460852 on Build GRCh38
rs767832969 Chr.13: 52461084-52461084 on Build GRCh38
rs770694033 Chr.13: 52460860-52460860 on Build GRCh38
rs763445264 Chr.13: 52461207-52461207 on Build GRCh38
rs979583979 Chr.13: 52461122-52461122 on Build GRCh38
rs753167885 Chr.13: 52460958-52460958 on Build GRCh38
rs546281774 Chr.13: 52461092-52461092 on Build GRCh38
rs372390966 Chr.13: 52461264-52461264 on Build GRCh38
rs764480140 Chr.13: 52461225-52461225 on Build GRCh38
rs758464956 Chr.13: 52460967-52460967 on Build GRCh38
rs149189169 Chr.13: 52461120-52461120 on Build GRCh38
rs1014596543 Chr.13: 52460875-52460875 on Build GRCh38
rs779723818 Chr.13: 52461121-52461121 on Build GRCh38
rs775612292 Chr.13: 52460904-52460904 on Build GRCh38
rs749162412 Chr.13: 52461137-52461137 on Build GRCh38
rs749182773 Chr.13: 52461015-52461015 on Build GRCh38
rs138583090 Chr.13: 52461205-52461205 on Build GRCh38
rs771681941 Chr.13: 52461039-52461039 on Build GRCh38
rs770006252 Chr.13: 52461131-52461132 on Build GRCh38
rs369526042 Chr.13: 52461055-52461055 on Build GRCh38
rs757410851 Chr.13: 52460980-52460980 on Build GRCh38
rs748578783 Chr.13: 52460963-52460967 on Build GRCh38
rs750611939 Chr.13: 52461262-52461262 on Build GRCh38
rs769855079 Chr.13: 52460894-52460894 on Build GRCh38
rs759934056 Chr.13: 52460943-52460943 on Build GRCh38
rs778359504 Chr.13: 52461173-52461174 on Build GRCh38
rs925451353 Chr.13: 52461129-52461129 on Build GRCh38
rs765710419 Chr.13: 52460946-52460946 on Build GRCh38
rs766687999 Chr.13: 52461272-52461272 on Build GRCh38
rs750701904 Chr.13: 52461101-52461101 on Build GRCh38
rs781151799 Chr.13: 52460893-52460893 on Build GRCh38
rs774270806 Chr.13: 52461283-52461283 on Build GRCh38
rs36022164 Chr.13: 52461155-52461155 on Build GRCh38
rs754195972 Chr.13: 52460942-52460942 on Build GRCh38
rs780095413 Chr.13: 52461001-52461001 on Build GRCh38
rs917946986 Chr.13: 52461227-52461227 on Build GRCh38
rs746670021 Chr.13: 52461191-52461191 on Build GRCh38
rs781630367 Chr.13: 52460989-52460989 on Build GRCh38
rs777774781 Chr.13: 52461161-52461161 on Build GRCh38
rs772813318 Chr.13: 52461180-52461180 on Build GRCh38
rs776001339 Chr.13: 52461058-52461058 on Build GRCh38
rs995266683 Chr.13: 52461270-52461270 on Build GRCh38
rs566498056 Chr.13: 52461054-52461054 on Build GRCh38
rs773719463 Chr.13: 52461028-52461028 on Build GRCh38
rs1045671475 Chr.13: 52460905-52460905 on Build GRCh38
rs751748472 Chr.13: 52460973-52460973 on Build GRCh38
rs771592179 Chr.13: 52461163-52461163 on Build GRCh38
rs774976799 Chr.13: 52461230-52461230 on Build GRCh38
rs768078904 Chr.13: 52460930-52460930 on Build GRCh38
rs775705785 Chr.13: 52461206-52461206 on Build GRCh38
rs370214081 Chr.13: 52461251-52461251 on Build GRCh38
rs778948021 Chr.13: 52461142-52461142 on Build GRCh38
rs768152052 Chr.13: 52461022-52461022 on Build GRCh38
rs905763228 Chr.13: 52461278-52461278 on Build GRCh38
rs777721907 Chr.13: 52460971-52460971 on Build GRCh38
rs772899454 Chr.13: 52461040-52461040 on Build GRCh38
rs767214221 Chr.13: 52460940-52460940 on Build GRCh38
rs762472862 Chr.13: 52460920-52460920 on Build GRCh38
rs780678371 Chr.13: 52460876-52460879 on Build GRCh38
rs1016719329 Chr.13: 52460970-52460970 on Build GRCh38
rs753544014 Chr.13: 52461281-52461281 on Build GRCh38
rs758498329 Chr.13: 52461141-52461141 on Build GRCh38
rs113062030 Chr.13: 52460996-52460996 on Build GRCh38
rs757457224 Chr.13: 52461082-52461082 on Build GRCh38

View Full Product Details