Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 52460974-52461480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKAP2
Entrez Gene ID
26586
Gene Name
cytoskeleton associated protein 2
Gene Aliases
LB1, TMAP, se20-10
UniGene
Hs.444028
Gene Chromosome Location
Chr.13: 52455360-52476631 on Build GRCh38
Transcripts
NM_001098525.2 NM_001286686.1 NM_001286687.1 NM_018204.4 XM_005266344.4 XM_011535043.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4500608 c.566C>T p.S189L Chr.13: 52461392-52461392 on Build GRCh38
4575982 c.412A>G p.S138G Chr.13: 52461238-52461238 on Build GRCh38
5547479 c.578_579delCT p.L194fs*5 Chr.13: 52461404-52461405 on Build GRCh38
7426755 c.592G>C p.D198H Chr.13: 52461418-52461418 on Build GRCh38
4617473 c.381G>A p.P127P Chr.13: 52461207-52461207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762419406 Chr.13: 52461334-52461334 on Build GRCh38
rs201217629 Chr.13: 52461250-52461250 on Build GRCh38
rs373545539 Chr.13: 52461064-52461064 on Build GRCh38
rs749598540 Chr.13: 52461314-52461314 on Build GRCh38
rs376367571 Chr.13: 52461093-52461093 on Build GRCh38
rs376601366 Chr.13: 52461034-52461034 on Build GRCh38
rs769763485 Chr.13: 52460999-52460999 on Build GRCh38
rs764723599 Chr.13: 52461066-52461066 on Build GRCh38
rs539183420 Chr.13: 52461074-52461074 on Build GRCh38
rs751040419 Chr.13: 52461401-52461401 on Build GRCh38
rs778665507 Chr.13: 52461287-52461287 on Build GRCh38
rs767832969 Chr.13: 52461084-52461084 on Build GRCh38
rs376545198 Chr.13: 52461337-52461337 on Build GRCh38
rs763445264 Chr.13: 52461207-52461207 on Build GRCh38
rs979583979 Chr.13: 52461122-52461122 on Build GRCh38
rs748586558 Chr.13: 52461438-52461438 on Build GRCh38
rs546281774 Chr.13: 52461092-52461092 on Build GRCh38
rs372390966 Chr.13: 52461264-52461264 on Build GRCh38
rs764480140 Chr.13: 52461225-52461225 on Build GRCh38
rs149189169 Chr.13: 52461120-52461120 on Build GRCh38
rs538606264 Chr.13: 52461320-52461320 on Build GRCh38
rs149254206 Chr.13: 52461343-52461343 on Build GRCh38
rs147763631 Chr.13: 52461408-52461408 on Build GRCh38
rs779723818 Chr.13: 52461121-52461121 on Build GRCh38
rs749162412 Chr.13: 52461137-52461137 on Build GRCh38
rs777280524 Chr.13: 52461298-52461298 on Build GRCh38
rs754111083 Chr.13: 52461357-52461357 on Build GRCh38
rs749182773 Chr.13: 52461015-52461015 on Build GRCh38
rs752574199 Chr.13: 52461373-52461373 on Build GRCh38
rs771681941 Chr.13: 52461039-52461039 on Build GRCh38
rs138583090 Chr.13: 52461205-52461205 on Build GRCh38
rs369526042 Chr.13: 52461055-52461055 on Build GRCh38
rs770006252 Chr.13: 52461131-52461132 on Build GRCh38
rs746580471 Chr.13: 52461299-52461299 on Build GRCh38
rs750611939 Chr.13: 52461262-52461262 on Build GRCh38
rs202229967 Chr.13: 52461388-52461388 on Build GRCh38
rs778359504 Chr.13: 52461173-52461174 on Build GRCh38
rs925451353 Chr.13: 52461129-52461129 on Build GRCh38
rs756842890 Chr.13: 52461407-52461407 on Build GRCh38
rs766687999 Chr.13: 52461272-52461272 on Build GRCh38
rs750701904 Chr.13: 52461101-52461101 on Build GRCh38
rs776389223 Chr.13: 52461306-52461306 on Build GRCh38
rs774270806 Chr.13: 52461283-52461283 on Build GRCh38
rs36022164 Chr.13: 52461155-52461155 on Build GRCh38
rs780095413 Chr.13: 52461001-52461001 on Build GRCh38
rs917946986 Chr.13: 52461227-52461227 on Build GRCh38
rs757837450 Chr.13: 52461297-52461297 on Build GRCh38
rs779404173 Chr.13: 52461431-52461431 on Build GRCh38
rs746670021 Chr.13: 52461191-52461191 on Build GRCh38
rs777774781 Chr.13: 52461161-52461161 on Build GRCh38
rs772813318 Chr.13: 52461180-52461180 on Build GRCh38
rs776001339 Chr.13: 52461058-52461058 on Build GRCh38
rs995266683 Chr.13: 52461270-52461270 on Build GRCh38
rs748115410 Chr.13: 52461295-52461295 on Build GRCh38
rs566498056 Chr.13: 52461054-52461054 on Build GRCh38
rs774650555 Chr.13: 52461322-52461322 on Build GRCh38
rs773719463 Chr.13: 52461028-52461028 on Build GRCh38
rs370906992 Chr.13: 52461364-52461364 on Build GRCh38
rs771426842 Chr.13: 52461445-52461446 on Build GRCh38
rs994329688 Chr.13: 52461331-52461331 on Build GRCh38
rs768891857 Chr.13: 52461393-52461393 on Build GRCh38
rs771592179 Chr.13: 52461163-52461163 on Build GRCh38
rs774976799 Chr.13: 52461230-52461230 on Build GRCh38
rs775705785 Chr.13: 52461206-52461206 on Build GRCh38
rs370214081 Chr.13: 52461251-52461251 on Build GRCh38
rs778948021 Chr.13: 52461142-52461142 on Build GRCh38
rs758319952 Chr.13: 52461384-52461384 on Build GRCh38
rs749732460 Chr.13: 52461301-52461304 on Build GRCh38
rs558016936 Chr.13: 52461349-52461349 on Build GRCh38
rs184495852 Chr.13: 52461362-52461362 on Build GRCh38
rs768152052 Chr.13: 52461022-52461022 on Build GRCh38
rs905763228 Chr.13: 52461278-52461278 on Build GRCh38
rs772899454 Chr.13: 52461040-52461040 on Build GRCh38
rs759212034 Chr.13: 52461360-52461360 on Build GRCh38
rs979613692 Chr.13: 52461457-52461457 on Build GRCh38
rs1007831465 Chr.13: 52461376-52461376 on Build GRCh38
rs770568360 Chr.13: 52461301-52461301 on Build GRCh38
rs768841255 Chr.13: 52461423-52461423 on Build GRCh38
rs772673417 Chr.13: 52461453-52461453 on Build GRCh38
rs753544014 Chr.13: 52461281-52461281 on Build GRCh38
rs758498329 Chr.13: 52461141-52461141 on Build GRCh38
rs757457224 Chr.13: 52461082-52461082 on Build GRCh38

View Full Product Details