Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 72734838-72735334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BORA
Entrez Gene ID
79866
Gene Name
bora, aurora kinase A activator
Gene Aliases
C13orf34
UniGene
Hs.643464
Gene Chromosome Location
Chr.13: 72727739-72756198 on Build GRCh38
Transcripts
NM_001286746.1 NM_001286747.1 NM_024808.3 XM_006719868.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
469549 c.280G>C p.D94H Chr.13: 72734979-72734979 on Build GRCh38
6477807 c.273T>C p.D91D Chr.13: 72734972-72734972 on Build GRCh38
948352 c.261-1G>T p.? Chr.13: 72734959-72734959 on Build GRCh38
7082924 c.303A>G p.E101E Chr.13: 72735002-72735002 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs992331333 Chr.13: 72735276-72735276 on Build GRCh38
rs771229685 Chr.13: 72735041-72735041 on Build GRCh38
rs765064551 Chr.13: 72734938-72734938 on Build GRCh38
rs369686134 Chr.13: 72735032-72735032 on Build GRCh38
rs1049696993 Chr.13: 72735148-72735148 on Build GRCh38
rs959749541 Chr.13: 72735255-72735255 on Build GRCh38
rs9543106 Chr.13: 72735209-72735209 on Build GRCh38
rs759376889 Chr.13: 72735055-72735055 on Build GRCh38
rs373466936 Chr.13: 72735061-72735061 on Build GRCh38
rs73215007 Chr.13: 72734893-72734893 on Build GRCh38
rs1025184249 Chr.13: 72735312-72735312 on Build GRCh38
rs757844570 Chr.13: 72734972-72734972 on Build GRCh38
rs960943369 Chr.13: 72735274-72735274 on Build GRCh38
rs752721172 Chr.13: 72734953-72734953 on Build GRCh38
rs775743285 Chr.13: 72735156-72735156 on Build GRCh38
rs889010307 Chr.13: 72735199-72735199 on Build GRCh38
rs998640969 Chr.13: 72735228-72735228 on Build GRCh38
rs561701002 Chr.13: 72735287-72735287 on Build GRCh38
rs193254224 Chr.13: 72735159-72735159 on Build GRCh38
rs185339074 Chr.13: 72735016-72735016 on Build GRCh38
rs773361499 Chr.13: 72735027-72735027 on Build GRCh38
rs187154562 Chr.13: 72734882-72734882 on Build GRCh38
rs141534178 Chr.13: 72735252-72735252 on Build GRCh38
rs146117511 Chr.13: 72735081-72735081 on Build GRCh38
rs891035382 Chr.13: 72735152-72735152 on Build GRCh38
rs777277947 Chr.13: 72734974-72734974 on Build GRCh38
rs532346924 Chr.13: 72735224-72735224 on Build GRCh38
rs1026713470 Chr.13: 72735243-72735243 on Build GRCh38
rs1021432562 Chr.13: 72735213-72735213 on Build GRCh38
rs766717033 Chr.13: 72734909-72734909 on Build GRCh38
rs188204614 Chr.13: 72735089-72735089 on Build GRCh38
rs748744513 Chr.13: 72735022-72735022 on Build GRCh38
rs751072822 Chr.13: 72734998-72734998 on Build GRCh38
rs1036852875 Chr.13: 72735144-72735144 on Build GRCh38
rs747116998 Chr.13: 72735030-72735030 on Build GRCh38
rs926913877 Chr.13: 72735254-72735254 on Build GRCh38
rs148778395 Chr.13: 72735251-72735251 on Build GRCh38
rs1007929044 Chr.13: 72735171-72735171 on Build GRCh38
rs58536774 Chr.13: 72735140-72735140 on Build GRCh38
rs777046557 Chr.13: 72735047-72735047 on Build GRCh38
rs756862571 Chr.13: 72734999-72734999 on Build GRCh38
rs535173543 Chr.13: 72735093-72735093 on Build GRCh38
rs369345288 Chr.13: 72734921-72734921 on Build GRCh38
rs780844901 Chr.13: 72735007-72735007 on Build GRCh38
rs948244456 Chr.13: 72735082-72735084 on Build GRCh38
rs945455527 Chr.13: 72735129-72735129 on Build GRCh38
rs772472667 Chr.13: 72735026-72735026 on Build GRCh38
rs78285044 Chr.13: 72734929-72734929 on Build GRCh38
rs992883370 Chr.13: 72735309-72735309 on Build GRCh38
rs1007371408 Chr.13: 72734931-72734931 on Build GRCh38
rs996667672 Chr.13: 72735145-72735145 on Build GRCh38
rs913913127 Chr.13: 72735294-72735294 on Build GRCh38
rs1043783760 Chr.13: 72735137-72735137 on Build GRCh38
rs952337434 Chr.13: 72735245-72735245 on Build GRCh38
rs755407256 Chr.13: 72735018-72735018 on Build GRCh38
rs961031343 Chr.13: 72735217-72735217 on Build GRCh38
rs376746925 Chr.13: 72735019-72735019 on Build GRCh38
rs143102101 Chr.13: 72734891-72734891 on Build GRCh38

View Full Product Details