Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 72743516-72744062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BORA
Entrez Gene ID
79866
Gene Name
bora, aurora kinase A activator
Gene Aliases
C13orf34
UniGene
Hs.643464
Gene Chromosome Location
Chr.13: 72727739-72756198 on Build GRCh38
Transcripts
NM_001286746.1 NM_001286747.1 NM_024808.3 XM_006719868.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1300283 c.444G>C p.E148D Chr.13: 72743592-72743592 on Build GRCh38
4166266 c.392C>G p.T131S Chr.13: 72743540-72743540 on Build GRCh38
5005774 c.454+7_454+8insGAGT p.? Chr.13: 72743609-72743610 on Build GRCh38
6139655 c.417T>A p.V139V Chr.13: 72743565-72743565 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs925818965 Chr.13: 72743587-72743587 on Build GRCh38
rs762512528 Chr.13: 72743592-72743592 on Build GRCh38
rs374630673 Chr.13: 72743541-72743541 on Build GRCh38
rs1034441561 Chr.13: 72743901-72743901 on Build GRCh38
rs949762729 Chr.13: 72743626-72743627 on Build GRCh38
rs899342981 Chr.13: 72743725-72743730 on Build GRCh38
rs764810727 Chr.13: 72743570-72743592 on Build GRCh38
rs566010311 Chr.13: 72743952-72743952 on Build GRCh38
rs765979426 Chr.13: 72743553-72743553 on Build GRCh38
rs774257505 Chr.13: 72743652-72743652 on Build GRCh38
rs527874501 Chr.13: 72743631-72743631 on Build GRCh38
rs1024082734 Chr.13: 72743894-72743894 on Build GRCh38
rs781366346 Chr.13: 72743611-72743611 on Build GRCh38
rs113751531 Chr.13: 72743851-72743851 on Build GRCh38
rs111789858 Chr.13: 72743625-72743625 on Build GRCh38
rs550602801 Chr.13: 72743900-72743900 on Build GRCh38
rs771106426 Chr.13: 72744036-72744036 on Build GRCh38
rs761761052 Chr.13: 72743884-72743884 on Build GRCh38
rs901031610 Chr.13: 72743840-72743840 on Build GRCh38
rs938298430 Chr.13: 72743720-72743724 on Build GRCh38
rs539788456 Chr.13: 72743944-72743944 on Build GRCh38
rs924066714 Chr.13: 72743975-72743975 on Build GRCh38
rs1041562121 Chr.13: 72744023-72744023 on Build GRCh38
rs755726759 Chr.13: 72743545-72743545 on Build GRCh38
rs749078503 Chr.13: 72743632-72743632 on Build GRCh38
rs768506032 Chr.13: 72743648-72743648 on Build GRCh38
rs189415826 Chr.13: 72743998-72743998 on Build GRCh38
rs374551550 Chr.13: 72743745-72743745 on Build GRCh38
rs189819729 Chr.13: 72743573-72743573 on Build GRCh38
rs370727431 Chr.13: 72743608-72743609 on Build GRCh38
rs911237818 Chr.13: 72744007-72744007 on Build GRCh38
rs1041117463 Chr.13: 72743828-72743828 on Build GRCh38
rs1008860223 Chr.13: 72743542-72743542 on Build GRCh38
rs988568412 Chr.13: 72743855-72743857 on Build GRCh38
rs558072203 Chr.13: 72743958-72743958 on Build GRCh38
rs750148563 Chr.13: 72743580-72743583 on Build GRCh38
rs965059123 Chr.13: 72743829-72743829 on Build GRCh38
rs916991190 Chr.13: 72743943-72743943 on Build GRCh38
rs559087662 Chr.13: 72743709-72743710 on Build GRCh38
rs572770857 Chr.13: 72744000-72744003 on Build GRCh38
rs34245891 Chr.13: 72743710-72743711 on Build GRCh38
rs1047844667 Chr.13: 72743787-72743787 on Build GRCh38
rs745963019 Chr.13: 72743618-72743618 on Build GRCh38
rs1044100445 Chr.13: 72743673-72743674 on Build GRCh38
rs567552435 Chr.13: 72744035-72744035 on Build GRCh38
rs949512192 Chr.13: 72743668-72743668 on Build GRCh38
rs924181178 Chr.13: 72743737-72743737 on Build GRCh38
rs1007405329 Chr.13: 72743818-72743818 on Build GRCh38
rs555509386 Chr.13: 72744004-72744004 on Build GRCh38
rs879197855 Chr.13: 72743704-72743704 on Build GRCh38
rs926947721 Chr.13: 72743692-72743692 on Build GRCh38
rs958373301 Chr.13: 72743910-72743910 on Build GRCh38
rs199764576 Chr.13: 72743728-72743728 on Build GRCh38
rs1025557675 Chr.13: 72743850-72743850 on Build GRCh38
rs533455567 Chr.13: 72743598-72743598 on Build GRCh38
rs185789667 Chr.13: 72743995-72743995 on Build GRCh38
rs753058025 Chr.13: 72743554-72743554 on Build GRCh38
rs773525713 Chr.13: 72743968-72743968 on Build GRCh38
rs760032340 Chr.13: 72743965-72743965 on Build GRCh38
rs568821738 Chr.13: 72743933-72743933 on Build GRCh38
rs867890173 Chr.13: 72743601-72743601 on Build GRCh38
rs754491332 Chr.13: 72743925-72743927 on Build GRCh38
rs1054197705 Chr.13: 72743808-72743808 on Build GRCh38
rs927009184 Chr.13: 72743922-72743922 on Build GRCh38
rs763116076 Chr.13: 72743984-72743984 on Build GRCh38
rs889765522 Chr.13: 72743817-72743817 on Build GRCh38
rs758738171 Chr.13: 72743559-72743559 on Build GRCh38
rs993009333 Chr.13: 72743934-72743934 on Build GRCh38
rs546712610 Chr.13: 72743722-72743722 on Build GRCh38
rs10648315 Chr.13: 72743602-72743603 on Build GRCh38
rs181867414 Chr.13: 72743599-72743599 on Build GRCh38
rs964697889 Chr.13: 72743548-72743548 on Build GRCh38
rs568042443 Chr.13: 72743848-72743848 on Build GRCh38
rs926942345 Chr.13: 72743560-72743560 on Build GRCh38
rs942704312 Chr.13: 72744021-72744021 on Build GRCh38

View Full Product Details