Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 106543839-106544364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARGLU1
Entrez Gene ID
55082
Gene Name
arginine and glutamate rich 1
Gene Aliases
-
UniGene
Hs.508644
Gene Chromosome Location
Chr.13: 106543314-106568166 on Build GRCh38
Transcripts
NM_018011.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4860590 c.283A>G p.I95V Chr.13: 106544142-106544142 on Build GRCh38
5784679 c.685G>C p.E229Q Chr.13: 106544133-106544133 on Build GRCh38
4865145 c.299G>T p.R100I Chr.13: 106544126-106544126 on Build GRCh38
5788834 c.674G>C p.R225T Chr.13: 106544144-106544144 on Build GRCh38
5818800 c.816G>A p.Q272Q Chr.13: 106544002-106544002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778747468 Chr.13: 106544178-106544178 on Build GRCh38
rs200630944 Chr.13: 106543996-106543996 on Build GRCh38
rs1033949989 Chr.13: 106543999-106543999 on Build GRCh38
rs970389800 Chr.13: 106544338-106544338 on Build GRCh38
rs780004952 Chr.13: 106544043-106544043 on Build GRCh38
rs1023998810 Chr.13: 106543971-106543971 on Build GRCh38
rs1035097543 Chr.13: 106544250-106544250 on Build GRCh38
rs542189823 Chr.13: 106543922-106543922 on Build GRCh38
rs775508514 Chr.13: 106543992-106543992 on Build GRCh38
rs769936315 Chr.13: 106544211-106544211 on Build GRCh38
rs900903435 Chr.13: 106544055-106544055 on Build GRCh38
rs939776257 Chr.13: 106543974-106543974 on Build GRCh38
rs758374254 Chr.13: 106544233-106544236 on Build GRCh38
rs763777376 Chr.13: 106543955-106543955 on Build GRCh38
rs555744192 Chr.13: 106543926-106543926 on Build GRCh38
rs898974689 Chr.13: 106543902-106543903 on Build GRCh38
rs35335049 Chr.13: 106543884-106543885 on Build GRCh38
rs752184474 Chr.13: 106544134-106544134 on Build GRCh38
rs773581159 Chr.13: 106544212-106544212 on Build GRCh38
rs752048143 Chr.13: 106544243-106544243 on Build GRCh38
rs890081703 Chr.13: 106544215-106544215 on Build GRCh38
rs770744880 Chr.13: 106543949-106543949 on Build GRCh38
rs751669783 Chr.13: 106544325-106544325 on Build GRCh38
rs771931263 Chr.13: 106544187-106544187 on Build GRCh38
rs369814396 Chr.13: 106544017-106544017 on Build GRCh38
rs745786558 Chr.13: 106543979-106543979 on Build GRCh38
rs190323937 Chr.13: 106544271-106544271 on Build GRCh38
rs750805855 Chr.13: 106544058-106544058 on Build GRCh38
rs765607811 Chr.13: 106544038-106544038 on Build GRCh38
rs777481840 Chr.13: 106544171-106544171 on Build GRCh38
rs978173073 Chr.13: 106543882-106543882 on Build GRCh38
rs768720047 Chr.13: 106544195-106544195 on Build GRCh38
rs762547248 Chr.13: 106543984-106543985 on Build GRCh38
rs747008965 Chr.13: 106544192-106544192 on Build GRCh38
rs767026676 Chr.13: 106544242-106544242 on Build GRCh38
rs924465398 Chr.13: 106543884-106543884 on Build GRCh38
rs920517591 Chr.13: 106544325-106544325 on Build GRCh38
rs756898950 Chr.13: 106544175-106544175 on Build GRCh38
rs774127440 Chr.13: 106544181-106544182 on Build GRCh38
rs772046103 Chr.13: 106543984-106543984 on Build GRCh38
rs768809885 Chr.13: 106544014-106544014 on Build GRCh38
rs555102520 Chr.13: 106544095-106544095 on Build GRCh38
rs974595340 Chr.13: 106544257-106544257 on Build GRCh38
rs565620741 Chr.13: 106544214-106544214 on Build GRCh38
rs191391862 Chr.13: 106543918-106543918 on Build GRCh38
rs544618903 Chr.13: 106544250-106544251 on Build GRCh38
rs911792752 Chr.13: 106543868-106543868 on Build GRCh38
rs1021029013 Chr.13: 106544131-106544131 on Build GRCh38
rs35001662 Chr.13: 106544246-106544247 on Build GRCh38
rs150839693 Chr.13: 106544248-106544248 on Build GRCh38
rs762076788 Chr.13: 106544209-106544209 on Build GRCh38
rs776618160 Chr.13: 106544197-106544197 on Build GRCh38
rs745681801 Chr.13: 106544185-106544185 on Build GRCh38
rs542476053 Chr.13: 106543900-106543901 on Build GRCh38
rs553428007 Chr.13: 106543896-106543896 on Build GRCh38
rs116511299 Chr.13: 106543915-106543915 on Build GRCh38
rs992661437 Chr.13: 106544280-106544280 on Build GRCh38
rs373211319 Chr.13: 106544188-106544188 on Build GRCh38
rs915708058 Chr.13: 106544307-106544307 on Build GRCh38
rs183057392 Chr.13: 106544217-106544217 on Build GRCh38
rs776901750 Chr.13: 106544020-106544020 on Build GRCh38
rs766176801 Chr.13: 106544179-106544182 on Build GRCh38
rs930478114 Chr.13: 106543903-106543908 on Build GRCh38
rs776295310 Chr.13: 106544196-106544196 on Build GRCh38
rs967221281 Chr.13: 106544157-106544157 on Build GRCh38
rs187030869 Chr.13: 106544246-106544246 on Build GRCh38
rs755696822 Chr.13: 106544158-106544158 on Build GRCh38
rs763509996 Chr.13: 106544084-106544084 on Build GRCh38
rs761358571 Chr.13: 106544223-106544223 on Build GRCh38
rs753522396 Chr.13: 106544174-106544174 on Build GRCh38
rs948637292 Chr.13: 106543942-106543942 on Build GRCh38
rs576636775 Chr.13: 106543927-106543927 on Build GRCh38

View Full Product Details