Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 106567772-106568268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARGLU1
Entrez Gene ID
55082
Gene Name
arginine and glutamate rich 1
Gene Aliases
-
UniGene
Hs.508644
Gene Chromosome Location
Chr.13: 106543314-106568166 on Build GRCh38
Transcripts
NM_018011.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1747331 c.13C>T p.R5W Chr.13: 106567907-106567907 on Build GRCh38
5778448 c.29C>G p.S10C Chr.13: 106567891-106567891 on Build GRCh38
4045815 c.43A>G p.T15A Chr.13: 106567877-106567877 on Build GRCh38
6248297 c.72C>A p.R24R Chr.13: 106567848-106567848 on Build GRCh38
7174647 c.29C>T p.S10F Chr.13: 106567891-106567891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754834266 Chr.13: 106567951-106567952 on Build GRCh38
rs548532517 Chr.13: 106568138-106568138 on Build GRCh38
rs552926481 Chr.13: 106568232-106568232 on Build GRCh38
rs1031640751 Chr.13: 106568111-106568111 on Build GRCh38
rs897463437 Chr.13: 106567909-106567909 on Build GRCh38
rs201575538 Chr.13: 106567879-106567879 on Build GRCh38
rs373124293 Chr.13: 106567947-106567947 on Build GRCh38
rs1006076088 Chr.13: 106568235-106568235 on Build GRCh38
rs767116951 Chr.13: 106567963-106567963 on Build GRCh38
rs982066568 Chr.13: 106568148-106568148 on Build GRCh38
rs541918282 Chr.13: 106567936-106567936 on Build GRCh38
rs770799372 Chr.13: 106567815-106567815 on Build GRCh38
rs1021899457 Chr.13: 106568096-106568096 on Build GRCh38
rs774377998 Chr.13: 106567817-106567817 on Build GRCh38
rs941952682 Chr.13: 106568163-106568163 on Build GRCh38
rs992911670 Chr.13: 106568123-106568123 on Build GRCh38
rs969392610 Chr.13: 106568105-106568105 on Build GRCh38
rs564623729 Chr.13: 106568010-106568010 on Build GRCh38
rs747205659 Chr.13: 106567946-106567946 on Build GRCh38
rs777577622 Chr.13: 106567920-106567920 on Build GRCh38
rs762095590 Chr.13: 106567952-106567952 on Build GRCh38
rs763591381 Chr.13: 106567959-106567959 on Build GRCh38
rs764064860 Chr.13: 106568167-106568167 on Build GRCh38
rs932967736 Chr.13: 106568190-106568190 on Build GRCh38
rs780912770 Chr.13: 106567911-106567911 on Build GRCh38
rs1028923322 Chr.13: 106568230-106568230 on Build GRCh38
rs533129113 Chr.13: 106568103-106568103 on Build GRCh38
rs752370349 Chr.13: 106567914-106567914 on Build GRCh38
rs765663261 Chr.13: 106567882-106567882 on Build GRCh38
rs908567560 Chr.13: 106568146-106568146 on Build GRCh38
rs373387006 Chr.13: 106567811-106567811 on Build GRCh38
rs573491268 Chr.13: 106567872-106567872 on Build GRCh38
rs959990234 Chr.13: 106568115-106568115 on Build GRCh38
rs755775967 Chr.13: 106567915-106567915 on Build GRCh38
rs759617578 Chr.13: 106567821-106567821 on Build GRCh38
rs546981591 Chr.13: 106568109-106568109 on Build GRCh38
rs1021058023 Chr.13: 106568097-106568097 on Build GRCh38
rs1008126470 Chr.13: 106568063-106568063 on Build GRCh38
rs909017851 Chr.13: 106568154-106568154 on Build GRCh38
rs553899979 Chr.13: 106568166-106568166 on Build GRCh38
rs1015595150 Chr.13: 106568239-106568239 on Build GRCh38
rs539026537 Chr.13: 106568231-106568231 on Build GRCh38
rs984112836 Chr.13: 106568145-106568145 on Build GRCh38
rs577040146 Chr.13: 106568181-106568181 on Build GRCh38
rs554990505 Chr.13: 106567927-106567927 on Build GRCh38
rs760846320 Chr.13: 106567873-106567873 on Build GRCh38
rs534911262 Chr.13: 106568116-106568116 on Build GRCh38
rs561923949 Chr.13: 106567902-106567902 on Build GRCh38
rs889319566 Chr.13: 106568004-106568004 on Build GRCh38
rs764497745 Chr.13: 106567877-106567877 on Build GRCh38
rs1010529560 Chr.13: 106568085-106568085 on Build GRCh38
rs568247157 Chr.13: 106568147-106568147 on Build GRCh38
rs765752130 Chr.13: 106567954-106567954 on Build GRCh38
rs752174832 Chr.13: 106567964-106567964 on Build GRCh38
rs565427919 Chr.13: 106568117-106568117 on Build GRCh38
rs752882738 Chr.13: 106568137-106568137 on Build GRCh38
rs181906968 Chr.13: 106568150-106568150 on Build GRCh38
rs755755080 Chr.13: 106567965-106567965 on Build GRCh38
rs997001881 Chr.13: 106568209-106568209 on Build GRCh38
rs566792442 Chr.13: 106568114-106568114 on Build GRCh38
rs902210042 Chr.13: 106568180-106568180 on Build GRCh38
rs777472804 Chr.13: 106567797-106567797 on Build GRCh38
rs775594566 Chr.13: 106567945-106567945 on Build GRCh38
rs745863402 Chr.13: 106567941-106567941 on Build GRCh38
rs1042042441 Chr.13: 106568173-106568173 on Build GRCh38
rs751053879 Chr.13: 106567896-106567896 on Build GRCh38
rs1041248615 Chr.13: 106568188-106568188 on Build GRCh38
rs1054914896 Chr.13: 106567999-106567999 on Build GRCh38
rs576878421 Chr.13: 106568189-106568189 on Build GRCh38
rs772225897 Chr.13: 106567852-106567852 on Build GRCh38
rs550624344 Chr.13: 106567948-106567948 on Build GRCh38
rs779153915 Chr.13: 106567855-106567857 on Build GRCh38
rs1051538468 Chr.13: 106568195-106568195 on Build GRCh38
rs749072172 Chr.13: 106567926-106567926 on Build GRCh38
rs952926899 Chr.13: 106568128-106568128 on Build GRCh38
rs915983110 Chr.13: 106568125-106568125 on Build GRCh38
rs773847676 Chr.13: 106567955-106567955 on Build GRCh38

View Full Product Details