Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 109754164-109754673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRS2
Entrez Gene ID
8660
Gene Name
insulin receptor substrate 2
Gene Aliases
IRS-2
UniGene
Hs.442344
Gene Chromosome Location
Chr.13: 109753837-109786567 on Build GRCh38
Transcripts
NM_003749.2

SNP Detail

SNP ID Chromosome Location
rs184775640 Chr.13: 109754448-109754448 on Build GRCh38
rs1020707653 Chr.13: 109754464-109754464 on Build GRCh38
rs114764400 Chr.13: 109754470-109754470 on Build GRCh38
rs913205504 Chr.13: 109754213-109754213 on Build GRCh38
rs41275080 Chr.13: 109754484-109754484 on Build GRCh38
rs371101194 Chr.13: 109754443-109754443 on Build GRCh38
rs892338627 Chr.13: 109754257-109754257 on Build GRCh38
rs536353518 Chr.13: 109754540-109754540 on Build GRCh38
rs71811421 Chr.13: 109754523-109754524 on Build GRCh38
rs1043492750 Chr.13: 109754302-109754302 on Build GRCh38
rs991705499 Chr.13: 109754360-109754360 on Build GRCh38
rs768193214 Chr.13: 109754406-109754406 on Build GRCh38
rs181042644 Chr.13: 109754589-109754589 on Build GRCh38
rs547656706 Chr.13: 109754505-109754505 on Build GRCh38
rs113246790 Chr.13: 109754523-109754523 on Build GRCh38
rs1046773778 Chr.13: 109754564-109754564 on Build GRCh38
rs567321084 Chr.13: 109754510-109754510 on Build GRCh38
rs534125055 Chr.13: 109754593-109754593 on Build GRCh38
rs923307959 Chr.13: 109754435-109754435 on Build GRCh38
rs769557402 Chr.13: 109754264-109754264 on Build GRCh38
rs1026484063 Chr.13: 109754361-109754361 on Build GRCh38
rs867464401 Chr.13: 109754242-109754242 on Build GRCh38
rs150855052 Chr.13: 109754592-109754592 on Build GRCh38
rs141399784 Chr.13: 109754569-109754569 on Build GRCh38
rs1013809502 Chr.13: 109754582-109754582 on Build GRCh38
rs1001641034 Chr.13: 109754430-109754430 on Build GRCh38
rs912479555 Chr.13: 109754496-109754496 on Build GRCh38
rs577795007 Chr.13: 109754235-109754235 on Build GRCh38
rs558015607 Chr.13: 109754590-109754590 on Build GRCh38
rs573844738 Chr.13: 109754306-109754306 on Build GRCh38
rs540462461 Chr.13: 109754247-109754247 on Build GRCh38
rs1001895796 Chr.13: 109754336-109754336 on Build GRCh38
rs950868436 Chr.13: 109754368-109754368 on Build GRCh38
rs775498762 Chr.13: 109754349-109754349 on Build GRCh38
rs985248857 Chr.13: 109754381-109754381 on Build GRCh38
rs181502906 Chr.13: 109754289-109754289 on Build GRCh38
rs376874772 Chr.13: 109754405-109754405 on Build GRCh38
rs551333920 Chr.13: 109754446-109754446 on Build GRCh38
rs1033790309 Chr.13: 109754340-109754340 on Build GRCh38
rs1054791995 Chr.13: 109754545-109754545 on Build GRCh38
rs1034427268 Chr.13: 109754437-109754437 on Build GRCh38
rs762862116 Chr.13: 109754352-109754352 on Build GRCh38
rs35685841 Chr.13: 109754522-109754523 on Build GRCh38
rs1016365399 Chr.13: 109754382-109754382 on Build GRCh38
rs866524766 Chr.13: 109754551-109754551 on Build GRCh38
rs933905833 Chr.13: 109754444-109754444 on Build GRCh38
rs549475514 Chr.13: 109754543-109754543 on Build GRCh38
rs963901053 Chr.13: 109754499-109754499 on Build GRCh38
rs542609695 Chr.13: 109754392-109754392 on Build GRCh38
rs896728995 Chr.13: 109754350-109754350 on Build GRCh38
rs945242538 Chr.13: 109754449-109754449 on Build GRCh38
rs531478607 Chr.13: 109754462-109754462 on Build GRCh38
rs1047188074 Chr.13: 109754373-109754373 on Build GRCh38
rs933938351 Chr.13: 109754225-109754225 on Build GRCh38
rs773896022 Chr.13: 109754414-109754414 on Build GRCh38
rs148299648 Chr.13: 109754465-109754465 on Build GRCh38

View Full Product Details