Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23356668-23357167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFS
Entrez Gene ID
10278
Gene Name
embryonal Fyn-associated substrate
Gene Aliases
CAS3, CASS3, EFS1, EFS2, HEFS, SIN
UniGene
Hs.24587
Gene Chromosome Location
Chr.14: 23356400-23365633 on Build GRCh38
Transcripts
NM_001277174.1 NM_005864.3 NM_032459.2 XM_005267256.1

SNP Detail

SNP ID Chromosome Location
rs981925342 Chr.14: 23356889-23356889 on Build GRCh38
rs545079993 Chr.14: 23357136-23357136 on Build GRCh38
rs971462453 Chr.14: 23356880-23356880 on Build GRCh38
rs901763045 Chr.14: 23357107-23357107 on Build GRCh38
rs952118648 Chr.14: 23356751-23356751 on Build GRCh38
rs572663870 Chr.14: 23357142-23357142 on Build GRCh38
rs957588816 Chr.14: 23356822-23356822 on Build GRCh38
rs201085012 Chr.14: 23356886-23356886 on Build GRCh38
rs1040255783 Chr.14: 23356718-23356718 on Build GRCh38
rs929847456 Chr.14: 23357045-23357045 on Build GRCh38
rs890776631 Chr.14: 23357093-23357093 on Build GRCh38
rs1000583279 Chr.14: 23357132-23357132 on Build GRCh38
rs573000324 Chr.14: 23357120-23357120 on Build GRCh38
rs1054180786 Chr.14: 23356989-23356989 on Build GRCh38
rs971085788 Chr.14: 23356805-23356805 on Build GRCh38
rs117880380 Chr.14: 23357063-23357063 on Build GRCh38
rs2231816 Chr.14: 23357114-23357114 on Build GRCh38
rs1008355304 Chr.14: 23356747-23356747 on Build GRCh38
rs185187816 Chr.14: 23356983-23356983 on Build GRCh38
rs935766811 Chr.14: 23356908-23356908 on Build GRCh38
rs760292762 Chr.14: 23356788-23356788 on Build GRCh38
rs180745255 Chr.14: 23356877-23356877 on Build GRCh38
rs962208887 Chr.14: 23356777-23356777 on Build GRCh38
rs1038188808 Chr.14: 23356966-23356966 on Build GRCh38
rs544555997 Chr.14: 23356878-23356878 on Build GRCh38
rs913063147 Chr.14: 23357049-23357049 on Build GRCh38
rs937152914 Chr.14: 23356952-23356952 on Build GRCh38
rs979751766 Chr.14: 23356839-23356839 on Build GRCh38
rs918134978 Chr.14: 23356789-23356789 on Build GRCh38
rs965226648 Chr.14: 23356757-23356757 on Build GRCh38
rs1006731454 Chr.14: 23357138-23357138 on Build GRCh38
rs112184652 Chr.14: 23356799-23356799 on Build GRCh38
rs890807046 Chr.14: 23356743-23356743 on Build GRCh38
rs555026368 Chr.14: 23356901-23356901 on Build GRCh38
rs772941965 Chr.14: 23356694-23356694 on Build GRCh38
rs566961397 Chr.14: 23356742-23356742 on Build GRCh38
rs996323440 Chr.14: 23356794-23356794 on Build GRCh38
rs184715647 Chr.14: 23356719-23356719 on Build GRCh38
rs1015526975 Chr.14: 23356762-23356762 on Build GRCh38
rs946439112 Chr.14: 23357067-23357067 on Build GRCh38
rs927431117 Chr.14: 23356950-23356950 on Build GRCh38
rs764192158 Chr.14: 23357117-23357117 on Build GRCh38
rs188601644 Chr.14: 23356720-23356720 on Build GRCh38
rs919772625 Chr.14: 23356975-23356975 on Build GRCh38
rs1038282099 Chr.14: 23356705-23356705 on Build GRCh38
rs887757418 Chr.14: 23357127-23357127 on Build GRCh38
rs552434831 Chr.14: 23356817-23356817 on Build GRCh38
rs1006414094 Chr.14: 23356755-23356755 on Build GRCh38
rs989029489 Chr.14: 23356849-23356849 on Build GRCh38
rs1040304831 Chr.14: 23357074-23357074 on Build GRCh38
rs532588371 Chr.14: 23356774-23356774 on Build GRCh38

View Full Product Details