Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 30589218-30589746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G2E3
Entrez Gene ID
55632
Gene Name
G2/M-phase specific E3 ubiquitin protein ligase
Gene Aliases
KIAA1333, PHF7B
UniGene
Hs.509008
Gene Chromosome Location
Chr.14: 30559123-30620064 on Build GRCh38
Transcripts
NM_001308097.1 NM_017769.4 XM_011536925.2 XM_011536926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5887420 c.135-4C>T p.? Chr.14: 30589378-30589378 on Build GRCh38
1238950 c.237+1G>A p.? Chr.14: 30589485-30589485 on Build GRCh38
69058 c.166_170delAAAGA p.K56fs*18 Chr.14: 30589413-30589417 on Build GRCh38
3495621 c.175G>A p.E59K Chr.14: 30589422-30589422 on Build GRCh38
955189 c.202G>A p.D68N Chr.14: 30589449-30589449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763015379 Chr.14: 30589375-30589375 on Build GRCh38
rs561565981 Chr.14: 30589532-30589532 on Build GRCh38
rs750866172 Chr.14: 30589511-30589511 on Build GRCh38
rs761666975 Chr.14: 30589365-30589365 on Build GRCh38
rs1057323654 Chr.14: 30589637-30589637 on Build GRCh38
rs572547938 Chr.14: 30589290-30589290 on Build GRCh38
rs1021753255 Chr.14: 30589664-30589664 on Build GRCh38
rs140166042 Chr.14: 30589311-30589311 on Build GRCh38
rs771447296 Chr.14: 30589336-30589336 on Build GRCh38
rs754920776 Chr.14: 30589343-30589344 on Build GRCh38
rs780625181 Chr.14: 30589484-30589485 on Build GRCh38
rs376052707 Chr.14: 30589633-30589635 on Build GRCh38
rs754110380 Chr.14: 30589503-30589503 on Build GRCh38
rs919807197 Chr.14: 30589473-30589473 on Build GRCh38
rs767430353 Chr.14: 30589396-30589396 on Build GRCh38
rs542746877 Chr.14: 30589352-30589352 on Build GRCh38
rs1020381279 Chr.14: 30589285-30589285 on Build GRCh38
rs996399073 Chr.14: 30589707-30589707 on Build GRCh38
rs548258405 Chr.14: 30589550-30589550 on Build GRCh38
rs768338052 Chr.14: 30589361-30589361 on Build GRCh38
rs567577014 Chr.14: 30589714-30589714 on Build GRCh38
rs1038158147 Chr.14: 30589654-30589654 on Build GRCh38
rs779624617 Chr.14: 30589353-30589353 on Build GRCh38
rs779284683 Chr.14: 30589506-30589506 on Build GRCh38
rs773233955 Chr.14: 30589371-30589371 on Build GRCh38
rs746400140 Chr.14: 30589334-30589334 on Build GRCh38
rs747775633 Chr.14: 30589428-30589428 on Build GRCh38
rs904762721 Chr.14: 30589706-30589706 on Build GRCh38
rs757571738 Chr.14: 30589505-30589505 on Build GRCh38
rs907686957 Chr.14: 30589608-30589608 on Build GRCh38
rs1010789081 Chr.14: 30589651-30589651 on Build GRCh38
rs899241608 Chr.14: 30589657-30589660 on Build GRCh38
rs375678714 Chr.14: 30589354-30589354 on Build GRCh38
rs958060345 Chr.14: 30589339-30589339 on Build GRCh38
rs536663228 Chr.14: 30589324-30589324 on Build GRCh38
rs940539743 Chr.14: 30589622-30589622 on Build GRCh38
rs749404673 Chr.14: 30589707-30589710 on Build GRCh38
rs747658299 Chr.14: 30589502-30589502 on Build GRCh38
rs1044060848 Chr.14: 30589507-30589507 on Build GRCh38
rs954786162 Chr.14: 30589261-30589261 on Build GRCh38
rs780479943 Chr.14: 30589524-30589524 on Build GRCh38
rs891956469 Chr.14: 30589644-30589644 on Build GRCh38
rs878912217 Chr.14: 30589328-30589328 on Build GRCh38
rs764346943 Chr.14: 30589493-30589493 on Build GRCh38
rs746520611 Chr.14: 30589528-30589528 on Build GRCh38
rs769759019 Chr.14: 30589369-30589369 on Build GRCh38
rs760733961 Chr.14: 30589442-30589442 on Build GRCh38
rs758961930 Chr.14: 30589515-30589515 on Build GRCh38

View Full Product Details