Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 30593282-30593787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G2E3
Entrez Gene ID
55632
Gene Name
G2/M-phase specific E3 ubiquitin protein ligase
Gene Aliases
KIAA1333, PHF7B
UniGene
Hs.509008
Gene Chromosome Location
Chr.14: 30559123-30620064 on Build GRCh38
Transcripts
NM_001308097.1 NM_017769.4 XM_011536925.2 XM_011536926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1252913 c.370T>C p.C124R Chr.14: 30593481-30593481 on Build GRCh38
1684379 c.517_518delGA p.D173fs*15 Chr.14: 30593628-30593629 on Build GRCh38
6487161 c.389T>C p.V130A Chr.14: 30593500-30593500 on Build GRCh38
1661932 c.406A>C p.N136H Chr.14: 30593517-30593517 on Build GRCh38
4550690 c.506G>A p.W169* Chr.14: 30593617-30593617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370503701 Chr.14: 30593642-30593642 on Build GRCh38
rs772585677 Chr.14: 30593650-30593650 on Build GRCh38
rs1030639978 Chr.14: 30593501-30593501 on Build GRCh38
rs372568601 Chr.14: 30593661-30593661 on Build GRCh38
rs528910545 Chr.14: 30593549-30593549 on Build GRCh38
rs776098583 Chr.14: 30593651-30593651 on Build GRCh38
rs376704586 Chr.14: 30593468-30593468 on Build GRCh38
rs971055000 Chr.14: 30593445-30593445 on Build GRCh38
rs749471272 Chr.14: 30593614-30593614 on Build GRCh38
rs753270402 Chr.14: 30593671-30593671 on Build GRCh38
rs1012571403 Chr.14: 30593399-30593399 on Build GRCh38
rs937052630 Chr.14: 30593673-30593673 on Build GRCh38
rs771456742 Chr.14: 30593668-30593671 on Build GRCh38
rs763118158 Chr.14: 30593428-30593428 on Build GRCh38
rs899038409 Chr.14: 30593362-30593362 on Build GRCh38
rs756870349 Chr.14: 30593488-30593488 on Build GRCh38
rs908568494 Chr.14: 30593723-30593723 on Build GRCh38
rs943826006 Chr.14: 30593691-30593691 on Build GRCh38
rs748903245 Chr.14: 30593518-30593523 on Build GRCh38
rs895449177 Chr.14: 30593711-30593714 on Build GRCh38
rs909746810 Chr.14: 30593314-30593314 on Build GRCh38
rs762735432 Chr.14: 30593582-30593582 on Build GRCh38
rs759957467 Chr.14: 30593649-30593649 on Build GRCh38
rs774593357 Chr.14: 30593438-30593438 on Build GRCh38
rs376658710 Chr.14: 30593686-30593686 on Build GRCh38
rs750131939 Chr.14: 30593526-30593526 on Build GRCh38
rs1038488717 Chr.14: 30593759-30593759 on Build GRCh38
rs763541897 Chr.14: 30593455-30593455 on Build GRCh38
rs191852930 Chr.14: 30593453-30593453 on Build GRCh38
rs934248603 Chr.14: 30593685-30593685 on Build GRCh38
rs761449206 Chr.14: 30593677-30593677 on Build GRCh38
rs201545564 Chr.14: 30593533-30593533 on Build GRCh38
rs200693428 Chr.14: 30593569-30593569 on Build GRCh38
rs912272878 Chr.14: 30593440-30593440 on Build GRCh38
rs764751462 Chr.14: 30593427-30593427 on Build GRCh38
rs777584106 Chr.14: 30593446-30593447 on Build GRCh38
rs139585540 Chr.14: 30593613-30593613 on Build GRCh38
rs758234085 Chr.14: 30593523-30593523 on Build GRCh38
rs769828044 Chr.14: 30593593-30593593 on Build GRCh38
rs1014330195 Chr.14: 30593712-30593712 on Build GRCh38
rs941424371 Chr.14: 30593724-30593724 on Build GRCh38
rs533648221 Chr.14: 30593665-30593665 on Build GRCh38
rs771262088 Chr.14: 30593436-30593436 on Build GRCh38
rs528218363 Chr.14: 30593454-30593454 on Build GRCh38
rs768106656 Chr.14: 30593446-30593446 on Build GRCh38
rs545708497 Chr.14: 30593343-30593343 on Build GRCh38
rs1055557915 Chr.14: 30593698-30593698 on Build GRCh38
rs34009891 Chr.14: 30593471-30593472 on Build GRCh38
rs541427833 Chr.14: 30593494-30593494 on Build GRCh38
rs1016492794 Chr.14: 30593335-30593335 on Build GRCh38
rs771046804 Chr.14: 30593635-30593635 on Build GRCh38
rs956441557 Chr.14: 30593505-30593505 on Build GRCh38
rs778682128 Chr.14: 30593493-30593493 on Build GRCh38
rs376502139 Chr.14: 30593612-30593612 on Build GRCh38

View Full Product Details