Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 30880361-30880761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COCH
Entrez Gene ID
1690
Gene Name
cochlin
Gene Aliases
COCH-5B2, COCH5B2, DFNA9
UniGene
Hs.21016
Gene Chromosome Location
Chr.14: 30874496-30890618 on Build GRCh38
Transcripts
NM_001135058.1 NM_004086.2 XM_017021071.1 XM_017021072.1 NR_038356.1

Gene Symbol
LOC100506071
Entrez Gene ID
100506071
Gene Name
uncharacterized LOC100506071
Gene Aliases
-
UniGene
Hs.729195
Gene Chromosome Location
Chr.14: 30876179-30889808 on Build GRCh38
Transcripts
NM_001135058.1 NM_004086.2 XM_017021071.1 XM_017021072.1 NR_038356.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2034214 c.622C>A p.Q208K Chr.14: 30880727-30880727 on Build GRCh38
955211 c.539G>A p.R180Q Chr.14: 30880644-30880644 on Build GRCh38
1201740 c.442C>T p.R148* Chr.14: 30880457-30880457 on Build GRCh38
7310872 c.592G>T p.G198* Chr.14: 30880697-30880697 on Build GRCh38
6278504 c.443G>A p.R148Q Chr.14: 30880458-30880458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755127637 Chr.14: 30880561-30880561 on Build GRCh38
rs369962252 Chr.14: 30880408-30880408 on Build GRCh38
rs763763889 Chr.14: 30880623-30880623 on Build GRCh38
rs375789836 Chr.14: 30880539-30880539 on Build GRCh38
rs960489145 Chr.14: 30880567-30880567 on Build GRCh38
rs745372732 Chr.14: 30880584-30880584 on Build GRCh38
rs751651997 Chr.14: 30880554-30880554 on Build GRCh38
rs141421714 Chr.14: 30880676-30880676 on Build GRCh38
rs759246142 Chr.14: 30880489-30880489 on Build GRCh38
rs146395267 Chr.14: 30880660-30880660 on Build GRCh38
rs752593278 Chr.14: 30880404-30880404 on Build GRCh38
rs757286862 Chr.14: 30880538-30880538 on Build GRCh38
rs559481815 Chr.14: 30880643-30880643 on Build GRCh38
rs771252412 Chr.14: 30880624-30880624 on Build GRCh38
rs1004024834 Chr.14: 30880511-30880511 on Build GRCh38
rs763942042 Chr.14: 30880409-30880409 on Build GRCh38
rs369882771 Chr.14: 30880644-30880644 on Build GRCh38
rs752834753 Chr.14: 30880652-30880652 on Build GRCh38
rs200549732 Chr.14: 30880715-30880715 on Build GRCh38
rs771685804 Chr.14: 30880697-30880697 on Build GRCh38
rs34622961 Chr.14: 30880465-30880466 on Build GRCh38
rs756414925 Chr.14: 30880572-30880572 on Build GRCh38
rs775356889 Chr.14: 30880526-30880526 on Build GRCh38
rs774069829 Chr.14: 30880477-30880477 on Build GRCh38
rs780155141 Chr.14: 30880435-30880435 on Build GRCh38
rs577009320 Chr.14: 30880558-30880558 on Build GRCh38
rs775303808 Chr.14: 30880605-30880605 on Build GRCh38
rs1012826993 Chr.14: 30880412-30880412 on Build GRCh38
rs768368801 Chr.14: 30880734-30880734 on Build GRCh38
rs773935807 Chr.14: 30880583-30880583 on Build GRCh38
rs756422562 Chr.14: 30880671-30880671 on Build GRCh38
rs779593424 Chr.14: 30880688-30880688 on Build GRCh38
rs755973091 Chr.14: 30880697-30880697 on Build GRCh38
rs541167867 Chr.14: 30880632-30880632 on Build GRCh38
rs543399762 Chr.14: 30880458-30880458 on Build GRCh38
rs753694963 Chr.14: 30880537-30880537 on Build GRCh38
rs962753370 Chr.14: 30880540-30880540 on Build GRCh38
rs765164378 Chr.14: 30880639-30880639 on Build GRCh38
rs773617215 Chr.14: 30880474-30880474 on Build GRCh38
rs1027550048 Chr.14: 30880505-30880505 on Build GRCh38
rs757768757 Chr.14: 30880687-30880687 on Build GRCh38
rs987057488 Chr.14: 30880707-30880707 on Build GRCh38
rs764041213 Chr.14: 30880530-30880530 on Build GRCh38
rs751600633 Chr.14: 30880641-30880641 on Build GRCh38
rs745321921 Chr.14: 30880693-30880693 on Build GRCh38
rs770299353 Chr.14: 30880464-30880464 on Build GRCh38
rs767288400 Chr.14: 30880499-30880499 on Build GRCh38
rs913569022 Chr.14: 30880611-30880611 on Build GRCh38
rs748500996 Chr.14: 30880566-30880566 on Build GRCh38
rs771622071 Chr.14: 30880582-30880582 on Build GRCh38
rs755252656 Chr.14: 30880457-30880457 on Build GRCh38
rs576687183 Chr.14: 30880434-30880434 on Build GRCh38
rs201955674 Chr.14: 30880576-30880576 on Build GRCh38
rs747162634 Chr.14: 30880428-30880428 on Build GRCh38
rs536937953 Chr.14: 30880394-30880394 on Build GRCh38
rs548387738 Chr.14: 30880720-30880720 on Build GRCh38
rs750476421 Chr.14: 30880433-30880433 on Build GRCh38
rs749858367 Chr.14: 30880475-30880475 on Build GRCh38
rs369035387 Chr.14: 30880387-30880387 on Build GRCh38
rs867764324 Chr.14: 30880431-30880431 on Build GRCh38
rs760423119 Chr.14: 30880615-30880615 on Build GRCh38
rs771810191 Chr.14: 30880601-30880601 on Build GRCh38
rs760418137 Chr.14: 30880528-30880528 on Build GRCh38
rs781427063 Chr.14: 30880564-30880564 on Build GRCh38
rs28400037 Chr.14: 30880388-30880388 on Build GRCh38
rs747229581 Chr.14: 30880441-30880441 on Build GRCh38
rs754241461 Chr.14: 30880680-30880680 on Build GRCh38

View Full Product Details