Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 34712843-34713352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFL2
Entrez Gene ID
1073
Gene Name
cofilin 2
Gene Aliases
NEM7
UniGene
Hs.180141
Gene Chromosome Location
Chr.14: 34710382-34714823 on Build GRCh38
Transcripts
NM_001243645.1 NM_021914.7 NM_138638.4 NR_028130.1 NR_028131.1 XM_011536363.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6214190 c.339C>A p.S113R Chr.14: 34713109-34713109 on Build GRCh38
2195847 c.264A>G p.T88T Chr.14: 34713301-34713301 on Build GRCh38
1742999 c.470T>A p.V157E Chr.14: 34712896-34712896 on Build GRCh38
6140648 c.461G>C p.G154A Chr.14: 34712905-34712905 on Build GRCh38
6042775 c.443C>T p.T148I Chr.14: 34712923-34712923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758639720 Chr.14: 34712946-34712946 on Build GRCh38
rs750722327 Chr.14: 34713198-34713198 on Build GRCh38
rs879148457 Chr.14: 34713297-34713297 on Build GRCh38
rs777207427 Chr.14: 34713291-34713291 on Build GRCh38
rs769277560 Chr.14: 34712975-34712975 on Build GRCh38
rs1052474137 Chr.14: 34713233-34713233 on Build GRCh38
rs779077709 Chr.14: 34712930-34712930 on Build GRCh38
rs35219728 Chr.14: 34713201-34713203 on Build GRCh38
rs772029781 Chr.14: 34713185-34713185 on Build GRCh38
rs529065569 Chr.14: 34713017-34713017 on Build GRCh38
rs757526740 Chr.14: 34712926-34712926 on Build GRCh38
rs976211060 Chr.14: 34713319-34713319 on Build GRCh38
rs1008143116 Chr.14: 34712929-34712929 on Build GRCh38
rs3835290 Chr.14: 34713209-34713211 on Build GRCh38
rs773564793 Chr.14: 34713296-34713300 on Build GRCh38
rs780623732 Chr.14: 34712958-34712958 on Build GRCh38
rs778057528 Chr.14: 34713215-34713215 on Build GRCh38
rs35515423 Chr.14: 34713143-34713143 on Build GRCh38
rs767724863 Chr.14: 34712950-34712950 on Build GRCh38
rs960366274 Chr.14: 34713088-34713088 on Build GRCh38
rs769373531 Chr.14: 34713068-34713068 on Build GRCh38
rs532379436 Chr.14: 34713013-34713016 on Build GRCh38
rs745669852 Chr.14: 34713141-34713142 on Build GRCh38
rs891173756 Chr.14: 34713238-34713238 on Build GRCh38
rs193170329 Chr.14: 34712999-34712999 on Build GRCh38
rs905045395 Chr.14: 34713064-34713064 on Build GRCh38
rs775291364 Chr.14: 34713155-34713155 on Build GRCh38
rs865927835 Chr.14: 34713095-34713095 on Build GRCh38
rs747406746 Chr.14: 34712969-34712969 on Build GRCh38
rs761839341 Chr.14: 34713024-34713024 on Build GRCh38
rs751907026 Chr.14: 34713232-34713232 on Build GRCh38
rs142164698 Chr.14: 34713298-34713298 on Build GRCh38
rs773337503 Chr.14: 34713219-34713219 on Build GRCh38
rs763138871 Chr.14: 34713228-34713228 on Build GRCh38
rs757986839 Chr.14: 34713143-34713144 on Build GRCh38
rs757178985 Chr.14: 34713008-34713012 on Build GRCh38
rs1014929507 Chr.14: 34713253-34713253 on Build GRCh38
rs780902770 Chr.14: 34712979-34712979 on Build GRCh38
rs200623045 Chr.14: 34713203-34713203 on Build GRCh38
rs748964090 Chr.14: 34713123-34713123 on Build GRCh38
rs759449417 Chr.14: 34713011-34713011 on Build GRCh38
rs767422749 Chr.14: 34713013-34713013 on Build GRCh38
rs756901142 Chr.14: 34713309-34713309 on Build GRCh38
rs765464943 Chr.14: 34713032-34713032 on Build GRCh38
rs916242055 Chr.14: 34713190-34713193 on Build GRCh38
rs765233939 Chr.14: 34713209-34713209 on Build GRCh38
rs774192992 Chr.14: 34713005-34713005 on Build GRCh38
rs748646442 Chr.14: 34713231-34713233 on Build GRCh38
rs1026763385 Chr.14: 34713171-34713171 on Build GRCh38
rs992200941 Chr.14: 34713183-34713183 on Build GRCh38
rs775357047 Chr.14: 34713186-34713186 on Build GRCh38
rs201619073 Chr.14: 34712871-34712871 on Build GRCh38
rs1018868261 Chr.14: 34712985-34712985 on Build GRCh38
rs781754042 Chr.14: 34713051-34713051 on Build GRCh38
rs777491432 Chr.14: 34713109-34713109 on Build GRCh38
rs755347719 Chr.14: 34713248-34713248 on Build GRCh38
rs932113690 Chr.14: 34712916-34712916 on Build GRCh38
rs766598005 Chr.14: 34713229-34713229 on Build GRCh38
rs768153736 Chr.14: 34713254-34713254 on Build GRCh38
rs761892592 Chr.14: 34713206-34713206 on Build GRCh38
rs769604813 Chr.14: 34712985-34712985 on Build GRCh38
rs766364990 Chr.14: 34712874-34712874 on Build GRCh38
rs747636094 Chr.14: 34712981-34712981 on Build GRCh38
rs772693480 Chr.14: 34712992-34712992 on Build GRCh38
rs547962200 Chr.14: 34712890-34712890 on Build GRCh38
rs982661020 Chr.14: 34713193-34713193 on Build GRCh38
rs770605729 Chr.14: 34713132-34713132 on Build GRCh38
rs747686516 Chr.14: 34713055-34713055 on Build GRCh38
rs138917817 Chr.14: 34712938-34712938 on Build GRCh38
rs755966265 Chr.14: 34712931-34712931 on Build GRCh38
rs200185906 Chr.14: 34713034-34713034 on Build GRCh38
rs770568677 Chr.14: 34713002-34713002 on Build GRCh38
rs145938782 Chr.14: 34712925-34712925 on Build GRCh38
rs751399023 Chr.14: 34712885-34712885 on Build GRCh38
rs935560061 Chr.14: 34712986-34712986 on Build GRCh38
rs758794470 Chr.14: 34713045-34713045 on Build GRCh38
rs774063031 Chr.14: 34713158-34713158 on Build GRCh38
rs995325576 Chr.14: 34713147-34713147 on Build GRCh38
rs756687915 Chr.14: 34713284-34713284 on Build GRCh38
rs944117415 Chr.14: 34713298-34713298 on Build GRCh38
rs753788777 Chr.14: 34712925-34712926 on Build GRCh38
rs1023778741 Chr.14: 34713052-34713052 on Build GRCh38
rs751934703 Chr.14: 34713049-34713049 on Build GRCh38
rs376185980 Chr.14: 34713022-34713022 on Build GRCh38
rs587780312 Chr.14: 34712909-34712909 on Build GRCh38
rs753192192 Chr.14: 34713277-34713277 on Build GRCh38
rs759298325 Chr.14: 34713181-34713181 on Build GRCh38
rs776043224 Chr.14: 34713225-34713227 on Build GRCh38
rs770223195 Chr.14: 34713243-34713288 on Build GRCh38
rs746731371 Chr.14: 34713142-34713143 on Build GRCh38
rs780345278 Chr.14: 34713047-34713047 on Build GRCh38

View Full Product Details