Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 34713121-34713643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFL2
Entrez Gene ID
1073
Gene Name
cofilin 2
Gene Aliases
NEM7
UniGene
Hs.180141
Gene Chromosome Location
Chr.14: 34710382-34714823 on Build GRCh38
Transcripts
NM_001243645.1 NM_021914.7 NM_138638.4 NR_028130.1 NR_028131.1 XM_011536363.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7297847 c.155C>T p.A52V Chr.14: 34713410-34713410 on Build GRCh38
7105198 c.193G>C p.E65Q Chr.14: 34713372-34713372 on Build GRCh38
955445 c.311+1G>A p.? Chr.14: 34713253-34713253 on Build GRCh38
2195851 c.186T>C p.D62D Chr.14: 34713379-34713379 on Build GRCh38
6339242 c.267C>T p.Y89Y Chr.14: 34713298-34713298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751972348 Chr.14: 34713485-34713487 on Build GRCh38
rs753321299 Chr.14: 34713514-34713514 on Build GRCh38
rs774697837 Chr.14: 34713493-34713495 on Build GRCh38
rs879148457 Chr.14: 34713297-34713297 on Build GRCh38
rs750722327 Chr.14: 34713198-34713198 on Build GRCh38
rs397515451 Chr.14: 34713546-34713546 on Build GRCh38
rs777207427 Chr.14: 34713291-34713291 on Build GRCh38
rs746780416 Chr.14: 34713552-34713552 on Build GRCh38
rs1052474137 Chr.14: 34713233-34713233 on Build GRCh38
rs35219728 Chr.14: 34713201-34713203 on Build GRCh38
rs772029781 Chr.14: 34713185-34713185 on Build GRCh38
rs752753334 Chr.14: 34713610-34713612 on Build GRCh38
rs976211060 Chr.14: 34713319-34713319 on Build GRCh38
rs932039956 Chr.14: 34713394-34713394 on Build GRCh38
rs3835290 Chr.14: 34713209-34713211 on Build GRCh38
rs773564793 Chr.14: 34713296-34713300 on Build GRCh38
rs778057528 Chr.14: 34713215-34713215 on Build GRCh38
rs35515423 Chr.14: 34713143-34713143 on Build GRCh38
rs768512313 Chr.14: 34713559-34713559 on Build GRCh38
rs891173756 Chr.14: 34713238-34713238 on Build GRCh38
rs763411916 Chr.14: 34713481-34713483 on Build GRCh38
rs1959149 Chr.14: 34713349-34713349 on Build GRCh38
rs775291364 Chr.14: 34713155-34713155 on Build GRCh38
rs537415856 Chr.14: 34713538-34713538 on Build GRCh38
rs1014692310 Chr.14: 34713507-34713507 on Build GRCh38
rs758040099 Chr.14: 34713548-34713548 on Build GRCh38
rs925438451 Chr.14: 34713418-34713418 on Build GRCh38
rs774659360 Chr.14: 34713434-34713434 on Build GRCh38
rs751907026 Chr.14: 34713232-34713232 on Build GRCh38
rs142164698 Chr.14: 34713298-34713298 on Build GRCh38
rs773337503 Chr.14: 34713219-34713219 on Build GRCh38
rs763138871 Chr.14: 34713228-34713228 on Build GRCh38
rs757986839 Chr.14: 34713143-34713144 on Build GRCh38
rs80358250 Chr.14: 34713462-34713462 on Build GRCh38
rs775410349 Chr.14: 34713387-34713387 on Build GRCh38
rs1014929507 Chr.14: 34713253-34713253 on Build GRCh38
rs200623045 Chr.14: 34713203-34713203 on Build GRCh38
rs749347212 Chr.14: 34713586-34713586 on Build GRCh38
rs778590067 Chr.14: 34713354-34713354 on Build GRCh38
rs772401948 Chr.14: 34713608-34713608 on Build GRCh38
rs756744491 Chr.14: 34713515-34713515 on Build GRCh38
rs756901142 Chr.14: 34713309-34713309 on Build GRCh38
rs759900342 Chr.14: 34713598-34713598 on Build GRCh38
rs763054045 Chr.14: 34713405-34713405 on Build GRCh38
rs916242055 Chr.14: 34713190-34713193 on Build GRCh38
rs765233939 Chr.14: 34713209-34713209 on Build GRCh38
rs145958933 Chr.14: 34713334-34713334 on Build GRCh38
rs747015278 Chr.14: 34713392-34713392 on Build GRCh38
rs748646442 Chr.14: 34713231-34713233 on Build GRCh38
rs1004680147 Chr.14: 34713492-34713492 on Build GRCh38
rs1026763385 Chr.14: 34713171-34713171 on Build GRCh38
rs992200941 Chr.14: 34713183-34713183 on Build GRCh38
rs775357047 Chr.14: 34713186-34713186 on Build GRCh38
rs757341071 Chr.14: 34713472-34713472 on Build GRCh38
rs149700171 Chr.14: 34713544-34713544 on Build GRCh38
rs75685384 Chr.14: 34713367-34713367 on Build GRCh38
rs755347719 Chr.14: 34713248-34713248 on Build GRCh38
rs766598005 Chr.14: 34713229-34713229 on Build GRCh38
rs899812033 Chr.14: 34713377-34713377 on Build GRCh38
rs768153736 Chr.14: 34713254-34713254 on Build GRCh38
rs761892592 Chr.14: 34713206-34713206 on Build GRCh38
rs185307646 Chr.14: 34713593-34713593 on Build GRCh38
rs922033257 Chr.14: 34713385-34713385 on Build GRCh38
rs776588232 Chr.14: 34713575-34713575 on Build GRCh38
rs982661020 Chr.14: 34713193-34713193 on Build GRCh38
rs766538409 Chr.14: 34713411-34713411 on Build GRCh38
rs797045460 Chr.14: 34713491-34713491 on Build GRCh38
rs759986387 Chr.14: 34713442-34713442 on Build GRCh38
rs759890309 Chr.14: 34713583-34713584 on Build GRCh38
rs969050808 Chr.14: 34713596-34713596 on Build GRCh38
rs372420724 Chr.14: 34713468-34713468 on Build GRCh38
rs771794609 Chr.14: 34713386-34713386 on Build GRCh38
rs774571512 Chr.14: 34713595-34713595 on Build GRCh38
rs779212509 Chr.14: 34713594-34713594 on Build GRCh38
rs774063031 Chr.14: 34713158-34713158 on Build GRCh38
rs995325576 Chr.14: 34713147-34713147 on Build GRCh38
rs200536303 Chr.14: 34713396-34713396 on Build GRCh38
rs756687915 Chr.14: 34713284-34713284 on Build GRCh38
rs139506764 Chr.14: 34713397-34713397 on Build GRCh38
rs148966691 Chr.14: 34713599-34713599 on Build GRCh38
rs764632685 Chr.14: 34713513-34713513 on Build GRCh38
rs944117415 Chr.14: 34713298-34713298 on Build GRCh38
rs542840152 Chr.14: 34713550-34713550 on Build GRCh38
rs1044719001 Chr.14: 34713590-34713590 on Build GRCh38
rs753192192 Chr.14: 34713277-34713277 on Build GRCh38
rs756734750 Chr.14: 34713476-34713476 on Build GRCh38
rs759298325 Chr.14: 34713181-34713181 on Build GRCh38
rs770223195 Chr.14: 34713243-34713288 on Build GRCh38
rs776043224 Chr.14: 34713225-34713227 on Build GRCh38

View Full Product Details