Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 34714450-34714966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFL2
Entrez Gene ID
1073
Gene Name
cofilin 2
Gene Aliases
NEM7
UniGene
Hs.180141
Gene Chromosome Location
Chr.14: 34710382-34714823 on Build GRCh38
Transcripts
NM_001243645.1 NM_138638.4 NR_028130.1 NR_028131.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4808438 c.3+8G>A p.? Chr.14: 34714530-34714530 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1015680915 Chr.14: 34714785-34714785 on Build GRCh38
rs961039790 Chr.14: 34714789-34714789 on Build GRCh38
rs17523485 Chr.14: 34714781-34714781 on Build GRCh38
rs1001908174 Chr.14: 34714893-34714893 on Build GRCh38
rs770312711 Chr.14: 34714601-34714601 on Build GRCh38
rs775806566 Chr.14: 34714497-34714497 on Build GRCh38
rs760962793 Chr.14: 34714503-34714503 on Build GRCh38
rs35137894 Chr.14: 34714911-34714911 on Build GRCh38
rs879280273 Chr.14: 34714708-34714708 on Build GRCh38
rs932382131 Chr.14: 34714827-34714827 on Build GRCh38
rs754654792 Chr.14: 34714840-34714841 on Build GRCh38
rs17523478 Chr.14: 34714837-34714838 on Build GRCh38
rs748426356 Chr.14: 34714598-34714598 on Build GRCh38
rs199792497 Chr.14: 34714911-34714911 on Build GRCh38
rs1046815914 Chr.14: 34714846-34714846 on Build GRCh38
rs926492909 Chr.14: 34714806-34714806 on Build GRCh38
rs777814896 Chr.14: 34714552-34714552 on Build GRCh38
rs541465259 Chr.14: 34714829-34714829 on Build GRCh38
rs755924600 Chr.14: 34714646-34714646 on Build GRCh38
rs575088363 Chr.14: 34714839-34714839 on Build GRCh38
rs756117892 Chr.14: 34714549-34714549 on Build GRCh38
rs571272233 Chr.14: 34714867-34714867 on Build GRCh38
rs906714105 Chr.14: 34714717-34714717 on Build GRCh38
rs780237178 Chr.14: 34714669-34714669 on Build GRCh38
rs781102093 Chr.14: 34714543-34714543 on Build GRCh38
rs879777401 Chr.14: 34714844-34714844 on Build GRCh38
rs1002241476 Chr.14: 34714853-34714853 on Build GRCh38
rs1053827949 Chr.14: 34714674-34714674 on Build GRCh38
rs779873355 Chr.14: 34714837-34714837 on Build GRCh38
rs765942244 Chr.14: 34714611-34714611 on Build GRCh38
rs773710471 Chr.14: 34714780-34714780 on Build GRCh38
rs556331060 Chr.14: 34714625-34714625 on Build GRCh38
rs368371830 Chr.14: 34714720-34714720 on Build GRCh38
rs752569476 Chr.14: 34714639-34714639 on Build GRCh38
rs567576604 Chr.14: 34714573-34714573 on Build GRCh38
rs773723587 Chr.14: 34714616-34714616 on Build GRCh38
rs752416089 Chr.14: 34714545-34714545 on Build GRCh38
rs1041181541 Chr.14: 34714837-34714837 on Build GRCh38
rs759285434 Chr.14: 34714621-34714621 on Build GRCh38
rs369681050 Chr.14: 34714531-34714531 on Build GRCh38
rs970887168 Chr.14: 34714475-34714475 on Build GRCh38
rs533579807 Chr.14: 34714487-34714487 on Build GRCh38
rs906651525 Chr.14: 34714848-34714848 on Build GRCh38
rs961490814 Chr.14: 34714738-34714738 on Build GRCh38
rs922439117 Chr.14: 34714594-34714594 on Build GRCh38
rs922398488 Chr.14: 34714799-34714799 on Build GRCh38
rs760552595 Chr.14: 34714843-34714843 on Build GRCh38
rs1053471228 Chr.14: 34714662-34714662 on Build GRCh38
rs773955266 Chr.14: 34714803-34714803 on Build GRCh38
rs901459821 Chr.14: 34714481-34714481 on Build GRCh38
rs936483456 Chr.14: 34714647-34714647 on Build GRCh38
rs970086131 Chr.14: 34714864-34714864 on Build GRCh38
rs1024337363 Chr.14: 34714746-34714746 on Build GRCh38
rs1033811966 Chr.14: 34714872-34714872 on Build GRCh38
rs997139091 Chr.14: 34714701-34714701 on Build GRCh38
rs765710594 Chr.14: 34714525-34714525 on Build GRCh38
rs376942478 Chr.14: 34714528-34714528 on Build GRCh38
rs1019833263 Chr.14: 34714478-34714478 on Build GRCh38
rs773657183 Chr.14: 34714605-34714605 on Build GRCh38
rs772455253 Chr.14: 34714492-34714492 on Build GRCh38
rs567482083 Chr.14: 34714795-34714795 on Build GRCh38
rs952951507 Chr.14: 34714527-34714527 on Build GRCh38
rs754585455 Chr.14: 34714533-34714533 on Build GRCh38
rs559662522 Chr.14: 34714482-34714482 on Build GRCh38
rs757172205 Chr.14: 34714600-34714602 on Build GRCh38
rs1025030706 Chr.14: 34714715-34714715 on Build GRCh38
rs777102210 Chr.14: 34714588-34714588 on Build GRCh38
rs536028256 Chr.14: 34714609-34714609 on Build GRCh38
rs985326502 Chr.14: 34714830-34714830 on Build GRCh38
rs528415846 Chr.14: 34714742-34714742 on Build GRCh38
rs1015761263 Chr.14: 34714730-34714730 on Build GRCh38
rs925775824 Chr.14: 34714836-34714836 on Build GRCh38
rs748564505 Chr.14: 34714709-34714709 on Build GRCh38
rs888578729 Chr.14: 34714921-34714921 on Build GRCh38
rs551361036 Chr.14: 34714838-34714838 on Build GRCh38
rs547355423 Chr.14: 34714504-34714504 on Build GRCh38
rs117211470 Chr.14: 34714682-34714682 on Build GRCh38
rs917233368 Chr.14: 34714595-34714595 on Build GRCh38
rs764141850 Chr.14: 34714663-34714663 on Build GRCh38
rs757253725 Chr.14: 34714668-34714668 on Build GRCh38
rs1005687804 Chr.14: 34714728-34714728 on Build GRCh38
rs753764659 Chr.14: 34714666-34714666 on Build GRCh38
rs780426319 Chr.14: 34714569-34714569 on Build GRCh38
rs776853901 Chr.14: 34714515-34714515 on Build GRCh38
rs771408387 Chr.14: 34714793-34714793 on Build GRCh38
rs765101534 Chr.14: 34714690-34714707 on Build GRCh38
rs957874639 Chr.14: 34714812-34714812 on Build GRCh38
rs376321909 Chr.14: 34714686-34714686 on Build GRCh38
rs545536413 Chr.14: 34714884-34714884 on Build GRCh38
rs1049573612 Chr.14: 34714608-34714608 on Build GRCh38
rs764045634 Chr.14: 34714851-34714851 on Build GRCh38
rs762440101 Chr.14: 34714518-34714518 on Build GRCh38
rs747322046 Chr.14: 34714572-34714572 on Build GRCh38
rs1033800597 Chr.14: 34714772-34714772 on Build GRCh38
rs949883387 Chr.14: 34714623-34714623 on Build GRCh38
rs750167030 Chr.14: 34714765-34714766 on Build GRCh38
rs747036710 Chr.14: 34714670-34714670 on Build GRCh38
rs745969445 Chr.14: 34714565-34714565 on Build GRCh38
rs556478891 Chr.14: 34714836-34714837 on Build GRCh38
rs1025567097 Chr.14: 34714474-34714474 on Build GRCh38
rs768747176 Chr.14: 34714576-34714576 on Build GRCh38
rs971276894 Chr.14: 34714715-34714734 on Build GRCh38
rs559132787 Chr.14: 34714753-34714753 on Build GRCh38

View Full Product Details