Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54842677-54843179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCH1
Entrez Gene ID
2643
Gene Name
GTP cyclohydrolase 1
Gene Aliases
DYT14, DYT5, DYT5a, GCH, GTP-CH-1, GTPCH1, HPABH4B
UniGene
Hs.86724
Gene Chromosome Location
Chr.14: 54842005-54902824 on Build GRCh38
Transcripts
NM_000161.2 NM_001024024.1 NM_001024070.1 NM_001024071.1 XM_017021218.1

SNP Detail

SNP ID Chromosome Location
rs56130647 Chr.14: 54842787-54842787 on Build GRCh38
rs770558029 Chr.14: 54843002-54843002 on Build GRCh38
rs925809602 Chr.14: 54842877-54842877 on Build GRCh38
rs939635665 Chr.14: 54843087-54843087 on Build GRCh38
rs566705845 Chr.14: 54842934-54842934 on Build GRCh38
rs941682193 Chr.14: 54843140-54843141 on Build GRCh38
rs868303976 Chr.14: 54843044-54843044 on Build GRCh38
rs773107365 Chr.14: 54843076-54843076 on Build GRCh38
rs890735774 Chr.14: 54842714-54842714 on Build GRCh38
rs189352198 Chr.14: 54843028-54843028 on Build GRCh38
rs549379993 Chr.14: 54842975-54842975 on Build GRCh38
rs376668776 Chr.14: 54843031-54843031 on Build GRCh38
rs144676716 Chr.14: 54842960-54842961 on Build GRCh38
rs758614226 Chr.14: 54843022-54843022 on Build GRCh38
rs867220379 Chr.14: 54842984-54842984 on Build GRCh38
rs941757524 Chr.14: 54842935-54842935 on Build GRCh38
rs964613934 Chr.14: 54842799-54842799 on Build GRCh38
rs772782531 Chr.14: 54842999-54842999 on Build GRCh38
rs879179089 Chr.14: 54843134-54843134 on Build GRCh38
rs930893834 Chr.14: 54843116-54843116 on Build GRCh38
rs10151500 Chr.14: 54842875-54842875 on Build GRCh38
rs568895596 Chr.14: 54843018-54843018 on Build GRCh38
rs76009965 Chr.14: 54842834-54842834 on Build GRCh38
rs770712450 Chr.14: 54843034-54843034 on Build GRCh38
rs967298634 Chr.14: 54842872-54842872 on Build GRCh38
rs143961472 Chr.14: 54842973-54842973 on Build GRCh38
rs1055904097 Chr.14: 54842960-54842960 on Build GRCh38
rs748793695 Chr.14: 54843051-54843051 on Build GRCh38
rs778955876 Chr.14: 54842817-54842817 on Build GRCh38
rs1036568285 Chr.14: 54842753-54842753 on Build GRCh38
rs995386735 Chr.14: 54842959-54842959 on Build GRCh38
rs886050544 Chr.14: 54842734-54842734 on Build GRCh38
rs908913666 Chr.14: 54843124-54843125 on Build GRCh38
rs777628224 Chr.14: 54843036-54843036 on Build GRCh38
rs930222058 Chr.14: 54843117-54843117 on Build GRCh38
rs183455779 Chr.14: 54843003-54843003 on Build GRCh38
rs982793541 Chr.14: 54842903-54842903 on Build GRCh38
rs769144102 Chr.14: 54842998-54842998 on Build GRCh38
rs1023705665 Chr.14: 54843065-54843065 on Build GRCh38
rs1047348151 Chr.14: 54842931-54842931 on Build GRCh38
rs768865732 Chr.14: 54843083-54843083 on Build GRCh38
rs899327360 Chr.14: 54842756-54842756 on Build GRCh38
rs145762799 Chr.14: 54843123-54843125 on Build GRCh38
rs983094016 Chr.14: 54843123-54843128 on Build GRCh38
rs955141614 Chr.14: 54843042-54843042 on Build GRCh38
rs554045962 Chr.14: 54842788-54842788 on Build GRCh38
rs993157713 Chr.14: 54842759-54842759 on Build GRCh38
rs140865143 Chr.14: 54843067-54843067 on Build GRCh38
rs150937402 Chr.14: 54843039-54843039 on Build GRCh38
rs551629849 Chr.14: 54842924-54842924 on Build GRCh38
rs764569623 Chr.14: 54843007-54843007 on Build GRCh38
rs543818323 Chr.14: 54842887-54842887 on Build GRCh38
rs776957978 Chr.14: 54843015-54843015 on Build GRCh38
rs978110731 Chr.14: 54842856-54842856 on Build GRCh38
rs572286120 Chr.14: 54842796-54842796 on Build GRCh38
rs1014282544 Chr.14: 54842983-54842983 on Build GRCh38
rs773983177 Chr.14: 54843042-54843042 on Build GRCh38
rs1036983132 Chr.14: 54842947-54842947 on Build GRCh38
rs918838734 Chr.14: 54843116-54843117 on Build GRCh38
rs899341447 Chr.14: 54842957-54842957 on Build GRCh38
rs748081442 Chr.14: 54842913-54842913 on Build GRCh38
rs957397528 Chr.14: 54842881-54842881 on Build GRCh38
rs920823865 Chr.14: 54843109-54843109 on Build GRCh38
rs56126158 Chr.14: 54842895-54842895 on Build GRCh38
rs1024244931 Chr.14: 54842865-54842865 on Build GRCh38
rs533048548 Chr.14: 54842912-54842912 on Build GRCh38
rs180979582 Chr.14: 54843001-54843001 on Build GRCh38
rs749552821 Chr.14: 54842750-54842750 on Build GRCh38
rs777063292 Chr.14: 54843086-54843086 on Build GRCh38
rs868459642 Chr.14: 54842946-54842946 on Build GRCh38
rs1005125171 Chr.14: 54842734-54842734 on Build GRCh38
rs370056236 Chr.14: 54843021-54843021 on Build GRCh38
rs1038747682 Chr.14: 54842955-54842955 on Build GRCh38
rs113211390 Chr.14: 54842841-54842841 on Build GRCh38
rs765167079 Chr.14: 54843017-54843017 on Build GRCh38
rs991004026 Chr.14: 54842894-54842894 on Build GRCh38
rs967152964 Chr.14: 54842843-54842843 on Build GRCh38
rs971340420 Chr.14: 54843079-54843079 on Build GRCh38
rs762415774 Chr.14: 54843016-54843016 on Build GRCh38
rs556250070 Chr.14: 54843032-54843032 on Build GRCh38
rs748137436 Chr.14: 54842989-54842989 on Build GRCh38
rs760998949 Chr.14: 54843004-54843004 on Build GRCh38
rs769757099 Chr.14: 54843066-54843066 on Build GRCh38
rs756091241 Chr.14: 54843035-54843035 on Build GRCh38
rs554432105 Chr.14: 54843094-54843094 on Build GRCh38
rs763425111 Chr.14: 54842760-54842760 on Build GRCh38
rs578152963 Chr.14: 54842786-54842786 on Build GRCh38

View Full Product Details