Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58004402-58004908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf37
Entrez Gene ID
145407
Gene Name
chromosome 14 open reading frame 37
Gene Aliases
c14_5376
UniGene
Hs.26766
Gene Chromosome Location
Chr.14: 57993536-58152305 on Build GRCh38
Transcripts
NM_001001872.3 XM_017021029.1 XM_017021030.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
553353 c.2273G>A p.S758N Chr.14: 58004788-58004788 on Build GRCh38
4536279 c.2285G>A p.R762Q Chr.14: 58004776-58004776 on Build GRCh38
4999998 c.2302G>A p.D768N Chr.14: 58004759-58004759 on Build GRCh38
4452907 c.2312A>T p.E771V Chr.14: 58004749-58004749 on Build GRCh38
3814974 c.2304C>T p.D768D Chr.14: 58004757-58004757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024440794 Chr.14: 58004620-58004620 on Build GRCh38
rs553729686 Chr.14: 58004871-58004871 on Build GRCh38
rs373676952 Chr.14: 58004754-58004754 on Build GRCh38
rs745935676 Chr.14: 58004569-58004569 on Build GRCh38
rs542386100 Chr.14: 58004442-58004442 on Build GRCh38
rs17807721 Chr.14: 58004631-58004631 on Build GRCh38
rs997734553 Chr.14: 58004475-58004475 on Build GRCh38
rs778823764 Chr.14: 58004735-58004735 on Build GRCh38
rs1013160042 Chr.14: 58004815-58004815 on Build GRCh38
rs969224500 Chr.14: 58004865-58004865 on Build GRCh38
rs779925956 Chr.14: 58004756-58004756 on Build GRCh38
rs546457781 Chr.14: 58004524-58004524 on Build GRCh38
rs1029578615 Chr.14: 58004475-58004476 on Build GRCh38
rs1050751982 Chr.14: 58004672-58004672 on Build GRCh38
rs181029578 Chr.14: 58004589-58004589 on Build GRCh38
rs771823293 Chr.14: 58004666-58004666 on Build GRCh38
rs994695036 Chr.14: 58004551-58004551 on Build GRCh38
rs998018386 Chr.14: 58004670-58004670 on Build GRCh38
rs989691065 Chr.14: 58004884-58004884 on Build GRCh38
rs563590028 Chr.14: 58004518-58004518 on Build GRCh38
rs932097528 Chr.14: 58004484-58004484 on Build GRCh38
rs754007607 Chr.14: 58004837-58004837 on Build GRCh38
rs370534913 Chr.14: 58004819-58004819 on Build GRCh38
rs991881785 Chr.14: 58004624-58004624 on Build GRCh38
rs775799797 Chr.14: 58004807-58004807 on Build GRCh38
rs1050506536 Chr.14: 58004687-58004687 on Build GRCh38
rs542934434 Chr.14: 58004846-58004846 on Build GRCh38
rs767995153 Chr.14: 58004760-58004760 on Build GRCh38
rs552623375 Chr.14: 58004665-58004665 on Build GRCh38
rs764225506 Chr.14: 58004834-58004834 on Build GRCh38
rs369463188 Chr.14: 58004660-58004661 on Build GRCh38
rs543362459 Chr.14: 58004505-58004505 on Build GRCh38
rs911955618 Chr.14: 58004714-58004714 on Build GRCh38
rs759355933 Chr.14: 58004776-58004776 on Build GRCh38
rs947868912 Chr.14: 58004630-58004630 on Build GRCh38
rs188065502 Chr.14: 58004556-58004556 on Build GRCh38
rs143126848 Chr.14: 58004872-58004872 on Build GRCh38
rs2295139 Chr.14: 58004637-58004637 on Build GRCh38
rs138638785 Chr.14: 58004786-58004786 on Build GRCh38
rs374667150 Chr.14: 58004704-58004704 on Build GRCh38
rs764278914 Chr.14: 58004719-58004719 on Build GRCh38
rs574050983 Chr.14: 58004474-58004474 on Build GRCh38
rs745711371 Chr.14: 58004736-58004736 on Build GRCh38
rs371375281 Chr.14: 58004694-58004694 on Build GRCh38
rs765617039 Chr.14: 58004847-58004847 on Build GRCh38
rs759882651 Chr.14: 58004792-58004792 on Build GRCh38
rs767927332 Chr.14: 58004793-58004793 on Build GRCh38
rs371953215 Chr.14: 58004690-58004690 on Build GRCh38
rs971123809 Chr.14: 58004629-58004629 on Build GRCh38
rs368780412 Chr.14: 58004696-58004696 on Build GRCh38
rs547864334 Chr.14: 58004814-58004814 on Build GRCh38
rs915339276 Chr.14: 58004458-58004458 on Build GRCh38
rs756078789 Chr.14: 58004708-58004708 on Build GRCh38
rs575290232 Chr.14: 58004476-58004476 on Build GRCh38
rs563284972 Chr.14: 58004459-58004459 on Build GRCh38
rs769484368 Chr.14: 58004777-58004777 on Build GRCh38
rs901919725 Chr.14: 58004472-58004472 on Build GRCh38
rs757580653 Chr.14: 58004733-58004733 on Build GRCh38
rs2256270 Chr.14: 58004571-58004571 on Build GRCh38
rs376684860 Chr.14: 58004773-58004773 on Build GRCh38
rs758065391 Chr.14: 58004852-58004852 on Build GRCh38
rs373959654 Chr.14: 58004851-58004851 on Build GRCh38
rs545492091 Chr.14: 58004504-58004504 on Build GRCh38
rs935371175 Chr.14: 58004646-58004646 on Build GRCh38
rs746972575 Chr.14: 58004759-58004759 on Build GRCh38
rs879018452 Chr.14: 58004661-58004661 on Build GRCh38
rs528392021 Chr.14: 58004523-58004523 on Build GRCh38
rs1000581403 Chr.14: 58004493-58004505 on Build GRCh38
rs145902415 Chr.14: 58004737-58004737 on Build GRCh38
rs80150192 Chr.14: 58004661-58004661 on Build GRCh38
rs377508439 Chr.14: 58004705-58004705 on Build GRCh38
rs998508093 Chr.14: 58004860-58004860 on Build GRCh38
rs17094192 Chr.14: 58004727-58004727 on Build GRCh38

View Full Product Details