Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58305944-58306445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID4A
Entrez Gene ID
5926
Gene Name
AT-rich interaction domain 4A
Gene Aliases
RBBP-1, RBBP1, RBP-1, RBP1
UniGene
Hs.161000
Gene Chromosome Location
Chr.14: 58298385-58373733 on Build GRCh38
Transcripts
NM_002892.3 NM_023000.2 NM_023001.2 XM_005267964.2 XM_005267965.2 XM_017021560.1 XM_017021561.1 XM_017021562.1 XM_017021563.1 XM_017021564.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6140830 c.216T>C p.D72D Chr.14: 58306054-58306054 on Build GRCh38
6076048 c.227A>G p.Q76R Chr.14: 58306065-58306065 on Build GRCh38
5568784 c.212C>T p.S71F Chr.14: 58306050-58306050 on Build GRCh38
6448771 c.253G>A p.D85N Chr.14: 58306091-58306091 on Build GRCh38
6604682 c.271G>A p.V91M Chr.14: 58306109-58306109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752222578 Chr.14: 58306323-58306324 on Build GRCh38
rs1026874278 Chr.14: 58306292-58306292 on Build GRCh38
rs566079585 Chr.14: 58306286-58306286 on Build GRCh38
rs945673107 Chr.14: 58306177-58306177 on Build GRCh38
rs754924074 Chr.14: 58305998-58305998 on Build GRCh38
rs1007834127 Chr.14: 58306262-58306262 on Build GRCh38
rs780349272 Chr.14: 58306110-58306110 on Build GRCh38
rs747377526 Chr.14: 58306115-58306115 on Build GRCh38
rs577090615 Chr.14: 58306421-58306421 on Build GRCh38
rs774835788 Chr.14: 58305976-58305979 on Build GRCh38
rs777084405 Chr.14: 58306121-58306121 on Build GRCh38
rs1053620489 Chr.14: 58306152-58306152 on Build GRCh38
rs780934127 Chr.14: 58306309-58306309 on Build GRCh38
rs772489797 Chr.14: 58306108-58306108 on Build GRCh38
rs890432485 Chr.14: 58306242-58306242 on Build GRCh38
rs769192149 Chr.14: 58306117-58306117 on Build GRCh38
rs938405116 Chr.14: 58306197-58306197 on Build GRCh38
rs906917841 Chr.14: 58306192-58306192 on Build GRCh38
rs752292659 Chr.14: 58306034-58306034 on Build GRCh38
rs371294379 Chr.14: 58306196-58306196 on Build GRCh38
rs781250349 Chr.14: 58306002-58306002 on Build GRCh38
rs748833176 Chr.14: 58306058-58306058 on Build GRCh38
rs766024843 Chr.14: 58305984-58305984 on Build GRCh38
rs982521635 Chr.14: 58306390-58306390 on Build GRCh38
rs777588429 Chr.14: 58306050-58306050 on Build GRCh38
rs1056072028 Chr.14: 58306209-58306209 on Build GRCh38
rs762822983 Chr.14: 58305983-58305983 on Build GRCh38
rs995452199 Chr.14: 58306323-58306323 on Build GRCh38
rs951212358 Chr.14: 58306382-58306383 on Build GRCh38
rs751369809 Chr.14: 58305994-58305994 on Build GRCh38
rs989737329 Chr.14: 58306142-58306142 on Build GRCh38
rs914238514 Chr.14: 58306159-58306159 on Build GRCh38
rs1041490876 Chr.14: 58306191-58306191 on Build GRCh38
rs1019663603 Chr.14: 58306365-58306365 on Build GRCh38
rs565170385 Chr.14: 58306338-58306338 on Build GRCh38
rs1017894390 Chr.14: 58306269-58306269 on Build GRCh38
rs369720691 Chr.14: 58306162-58306162 on Build GRCh38
rs963614562 Chr.14: 58306273-58306273 on Build GRCh38
rs755643697 Chr.14: 58306049-58306049 on Build GRCh38
rs761906548 Chr.14: 58306153-58306153 on Build GRCh38
rs975517415 Chr.14: 58306008-58306008 on Build GRCh38
rs142407317 Chr.14: 58306026-58306026 on Build GRCh38
rs949314737 Chr.14: 58306006-58306006 on Build GRCh38
rs375549627 Chr.14: 58306226-58306226 on Build GRCh38
rs575829324 Chr.14: 58306236-58306236 on Build GRCh38
rs887126568 Chr.14: 58306298-58306298 on Build GRCh38
rs543254532 Chr.14: 58306255-58306255 on Build GRCh38

View Full Product Details