Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 67384341-67384888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2S1
Entrez Gene ID
1965
Gene Name
eukaryotic translation initiation factor 2 subunit alpha
Gene Aliases
EIF-2, EIF-2A, EIF-2alpha, EIF2, EIF2A
UniGene
Hs.151777
Gene Chromosome Location
Chr.14: 67360317-67386516 on Build GRCh38
Transcripts
NM_004094.4 XM_017020925.1

Gene Symbol
GPHN
Entrez Gene ID
10243
Gene Name
gephyrin
Gene Aliases
GEPH, GPH, GPHRYN, HKPX1, MOCODC
UniGene
Hs.208765
Gene Chromosome Location
Chr.14: 66507407-67735831 on Build GRCh38
Transcripts
NM_004094.4 XM_017020925.1

SNP Detail

SNP ID Chromosome Location
rs929492174 Chr.14: 67384504-67384504 on Build GRCh38
rs945743960 Chr.14: 67384696-67384696 on Build GRCh38
rs559859294 Chr.14: 67384673-67384673 on Build GRCh38
rs1001873098 Chr.14: 67384612-67384612 on Build GRCh38
rs397766076 Chr.14: 67384380-67384381 on Build GRCh38
rs34318358 Chr.14: 67384367-67384368 on Build GRCh38
rs558439482 Chr.14: 67384680-67384680 on Build GRCh38
rs749449805 Chr.14: 67384672-67384672 on Build GRCh38
rs146973725 Chr.14: 67384639-67384639 on Build GRCh38
rs966838457 Chr.14: 67384466-67384466 on Build GRCh38
rs74058416 Chr.14: 67384531-67384531 on Build GRCh38
rs1051856466 Chr.14: 67384418-67384418 on Build GRCh38
rs75955766 Chr.14: 67384632-67384632 on Build GRCh38
rs983747172 Chr.14: 67384568-67384568 on Build GRCh38
rs1001040761 Chr.14: 67384481-67384481 on Build GRCh38
rs995977928 Chr.14: 67384370-67384370 on Build GRCh38
rs900261869 Chr.14: 67384367-67384367 on Build GRCh38
rs904862877 Chr.14: 67384593-67384593 on Build GRCh38
rs183671945 Chr.14: 67384434-67384434 on Build GRCh38
rs896224299 Chr.14: 67384638-67384638 on Build GRCh38
rs532134486 Chr.14: 67384445-67384445 on Build GRCh38
rs1032498643 Chr.14: 67384804-67384804 on Build GRCh38
rs1012724593 Chr.14: 67384845-67384845 on Build GRCh38
rs1020762629 Chr.14: 67384649-67384649 on Build GRCh38
rs979295059 Chr.14: 67384422-67384424 on Build GRCh38
rs923379440 Chr.14: 67384498-67384498 on Build GRCh38
rs1025072792 Chr.14: 67384862-67384862 on Build GRCh38
rs141325817 Chr.14: 67384555-67384555 on Build GRCh38
rs895341382 Chr.14: 67384838-67384838 on Build GRCh38
rs965113840 Chr.14: 67384695-67384695 on Build GRCh38
rs1042628801 Chr.14: 67384706-67384706 on Build GRCh38
rs976103209 Chr.14: 67384725-67384727 on Build GRCh38
rs370644660 Chr.14: 67384716-67384716 on Build GRCh38
rs373197062 Chr.14: 67384784-67384784 on Build GRCh38
rs781644450 Chr.14: 67384459-67384459 on Build GRCh38
rs921799538 Chr.14: 67384667-67384667 on Build GRCh38
rs960862363 Chr.14: 67384367-67384367 on Build GRCh38
rs987984728 Chr.14: 67384368-67384368 on Build GRCh38
rs967827292 Chr.14: 67384652-67384652 on Build GRCh38
rs1032552429 Chr.14: 67384524-67384524 on Build GRCh38
rs923030352 Chr.14: 67384777-67384777 on Build GRCh38
rs770107358 Chr.14: 67384582-67384582 on Build GRCh38
rs547862762 Chr.14: 67384584-67384584 on Build GRCh38
rs75744302 Chr.14: 67384622-67384622 on Build GRCh38
rs923372368 Chr.14: 67384815-67384815 on Build GRCh38
rs780287322 Chr.14: 67384817-67384817 on Build GRCh38
rs868435072 Chr.14: 67384640-67384640 on Build GRCh38
rs942271664 Chr.14: 67384551-67384552 on Build GRCh38
rs902766340 Chr.14: 67384708-67384708 on Build GRCh38
rs147836861 Chr.14: 67384465-67384465 on Build GRCh38
rs970774531 Chr.14: 67384556-67384556 on Build GRCh38
rs1034734951 Chr.14: 67384366-67384367 on Build GRCh38
rs990949751 Chr.14: 67384553-67384553 on Build GRCh38

View Full Product Details