Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73502538-73503075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT1
Entrez Gene ID
641371
Gene Name
acyl-CoA thioesterase 1
Gene Aliases
ACH2, CTE-1, LACH2
UniGene
Hs.446685
Gene Chromosome Location
Chr.14: 73493021-73543799 on Build GRCh38
Transcripts
XM_017021590.1 XM_017021591.1 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_017021289.1

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
-
UniGene
Hs.660774
Gene Chromosome Location
Chr.14: 73478484-73634418 on Build GRCh38
Transcripts
XM_017021590.1 XM_017021591.1 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_017021289.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7189685 c.2087A>C p.E696A Chr.14: 73502913-73502913 on Build GRCh38
4689804 c.1858A>C p.K620Q Chr.14: 73503001-73503001 on Build GRCh38
5959894 c.1953C>T p.H651H Chr.14: 73502906-73502906 on Build GRCh38
5971243 c.1919C>T p.A640V Chr.14: 73502940-73502940 on Build GRCh38
5596833 c.1941G>A p.G647G Chr.14: 73502918-73502918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895344506 Chr.14: 73502571-73502571 on Build GRCh38
rs866633093 Chr.14: 73502620-73502620 on Build GRCh38
rs749447091 Chr.14: 73503043-73503043 on Build GRCh38
rs919862904 Chr.14: 73502668-73502668 on Build GRCh38
rs1051422169 Chr.14: 73502763-73502763 on Build GRCh38
rs922789545 Chr.14: 73502928-73502928 on Build GRCh38
rs770849626 Chr.14: 73502879-73502879 on Build GRCh38
rs763183716 Chr.14: 73502601-73502601 on Build GRCh38
rs571789320 Chr.14: 73502789-73502789 on Build GRCh38
rs536726874 Chr.14: 73502894-73502894 on Build GRCh38
rs752302136 Chr.14: 73502953-73502953 on Build GRCh38
rs756122377 Chr.14: 73503037-73503037 on Build GRCh38
rs1008304357 Chr.14: 73502902-73502902 on Build GRCh38
rs928331114 Chr.14: 73502730-73502730 on Build GRCh38
rs756133895 Chr.14: 73502820-73502820 on Build GRCh38
rs764412315 Chr.14: 73502650-73502650 on Build GRCh38
rs756519907 Chr.14: 73503049-73503052 on Build GRCh38
rs745827012 Chr.14: 73503007-73503007 on Build GRCh38
rs1043208002 Chr.14: 73502795-73502795 on Build GRCh38
rs948828598 Chr.14: 73502600-73502600 on Build GRCh38
rs61987096 Chr.14: 73502664-73502664 on Build GRCh38
rs768045009 Chr.14: 73502863-73502864 on Build GRCh38
rs751706230 Chr.14: 73502651-73502651 on Build GRCh38
rs746859092 Chr.14: 73502878-73502878 on Build GRCh38
rs551149994 Chr.14: 73502871-73502871 on Build GRCh38
rs752321894 Chr.14: 73502867-73502867 on Build GRCh38
rs750874906 Chr.14: 73502855-73502855 on Build GRCh38
rs781283201 Chr.14: 73502888-73502888 on Build GRCh38
rs757968898 Chr.14: 73502979-73502979 on Build GRCh38
rs1043934820 Chr.14: 73502860-73502860 on Build GRCh38
rs745764069 Chr.14: 73502892-73502892 on Build GRCh38
rs775676644 Chr.14: 73502901-73502901 on Build GRCh38
rs754459009 Chr.14: 73502861-73502861 on Build GRCh38
rs147719236 Chr.14: 73502864-73502865 on Build GRCh38
rs768075421 Chr.14: 73502847-73502847 on Build GRCh38
rs111526585 Chr.14: 73502673-73502673 on Build GRCh38
rs371823233 Chr.14: 73502921-73502921 on Build GRCh38
rs753011821 Chr.14: 73502907-73502908 on Build GRCh38
rs11626122 Chr.14: 73502906-73502906 on Build GRCh38
rs184765328 Chr.14: 73503041-73503041 on Build GRCh38
rs978081970 Chr.14: 73502634-73502634 on Build GRCh38
rs372061895 Chr.14: 73502984-73502984 on Build GRCh38
rs761202138 Chr.14: 73502938-73502938 on Build GRCh38
rs370564586 Chr.14: 73502957-73502957 on Build GRCh38
rs1031492100 Chr.14: 73502875-73502875 on Build GRCh38
rs762917216 Chr.14: 73502905-73502905 on Build GRCh38
rs904266294 Chr.14: 73503021-73503021 on Build GRCh38
rs555141796 Chr.14: 73502769-73502769 on Build GRCh38
rs111562922 Chr.14: 73502962-73502962 on Build GRCh38
rs142135484 Chr.14: 73502923-73502923 on Build GRCh38
rs529716151 Chr.14: 73502635-73502635 on Build GRCh38
rs1022533119 Chr.14: 73502608-73502608 on Build GRCh38
rs147647682 Chr.14: 73502915-73502915 on Build GRCh38
rs968003298 Chr.14: 73502610-73502610 on Build GRCh38
rs934230619 Chr.14: 73502944-73502944 on Build GRCh38
rs1012493399 Chr.14: 73502572-73502572 on Build GRCh38
rs569387576 Chr.14: 73502876-73502876 on Build GRCh38
rs1007303718 Chr.14: 73502790-73502790 on Build GRCh38
rs369712820 Chr.14: 73503046-73503046 on Build GRCh38
rs532751176 Chr.14: 73502798-73502798 on Build GRCh38
rs781088389 Chr.14: 73503003-73503003 on Build GRCh38
rs757135108 Chr.14: 73502998-73502998 on Build GRCh38
rs769788840 Chr.14: 73502893-73502893 on Build GRCh38
rs778436873 Chr.14: 73502866-73502866 on Build GRCh38
rs201974868 Chr.14: 73502920-73502920 on Build GRCh38
rs1050467088 Chr.14: 73502971-73502971 on Build GRCh38
rs562244964 Chr.14: 73502609-73502609 on Build GRCh38
rs376616331 Chr.14: 73502877-73502877 on Build GRCh38
rs553523284 Chr.14: 73502990-73502990 on Build GRCh38
rs187705000 Chr.14: 73502638-73502638 on Build GRCh38
rs750460581 Chr.14: 73502735-73502735 on Build GRCh38
rs942560517 Chr.14: 73502658-73502658 on Build GRCh38
rs766767211 Chr.14: 73502939-73502939 on Build GRCh38
rs193230151 Chr.14: 73502705-73502705 on Build GRCh38
rs558821646 Chr.14: 73502989-73502989 on Build GRCh38
rs984645869 Chr.14: 73502612-73502612 on Build GRCh38
rs943248710 Chr.14: 73503004-73503004 on Build GRCh38

View Full Product Details