Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77858975-77859472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCK1
Entrez Gene ID
57143
Gene Name
aarF domain containing kinase 1
Gene Aliases
-
UniGene
Hs.413208
Gene Chromosome Location
Chr.14: 77800058-77933954 on Build GRCh38
Transcripts
NM_001142545.1 NM_020421.3 XM_005267885.3 XM_005267886.3 XM_011536987.2 XM_011536988.2 XM_017021474.1 XM_017021475.1 XM_017021476.1 XM_017021477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6594763 c.366T>C p.A122A Chr.14: 77859222-77859222 on Build GRCh38
4407767 c.263C>A p.A88D Chr.14: 77859119-77859119 on Build GRCh38
5705776 c.260G>A p.C87Y Chr.14: 77859116-77859116 on Build GRCh38
6278509 c.423+5C>A p.? Chr.14: 77859284-77859284 on Build GRCh38
3793890 c.285G>A p.K95K Chr.14: 77859141-77859141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544400304 Chr.14: 77859229-77859229 on Build GRCh38
rs562565501 Chr.14: 77859283-77859283 on Build GRCh38
rs747527460 Chr.14: 77859225-77859225 on Build GRCh38
rs766848926 Chr.14: 77859125-77859125 on Build GRCh38
rs968820232 Chr.14: 77859036-77859036 on Build GRCh38
rs754193430 Chr.14: 77859145-77859145 on Build GRCh38
rs778973947 Chr.14: 77859201-77859201 on Build GRCh38
rs772034265 Chr.14: 77859217-77859217 on Build GRCh38
rs749507611 Chr.14: 77859099-77859099 on Build GRCh38
rs774139527 Chr.14: 77859026-77859026 on Build GRCh38
rs1048220334 Chr.14: 77859233-77859233 on Build GRCh38
rs143170023 Chr.14: 77859177-77859177 on Build GRCh38
rs576777492 Chr.14: 77859336-77859336 on Build GRCh38
rs77725622 Chr.14: 77859285-77859285 on Build GRCh38
rs759187791 Chr.14: 77859282-77859293 on Build GRCh38
rs765034942 Chr.14: 77859047-77859047 on Build GRCh38
rs147514523 Chr.14: 77859185-77859185 on Build GRCh38
rs767008084 Chr.14: 77859253-77859253 on Build GRCh38
rs987638749 Chr.14: 77859379-77859379 on Build GRCh38
rs200591022 Chr.14: 77859296-77859296 on Build GRCh38
rs931029925 Chr.14: 77859247-77859247 on Build GRCh38
rs761758645 Chr.14: 77859109-77859109 on Build GRCh38
rs912157457 Chr.14: 77859396-77859396 on Build GRCh38
rs969980282 Chr.14: 77859186-77859186 on Build GRCh38
rs752386872 Chr.14: 77859051-77859051 on Build GRCh38
rs753673888 Chr.14: 77859271-77859271 on Build GRCh38
rs780948276 Chr.14: 77859096-77859096 on Build GRCh38
rs754708436 Chr.14: 77859274-77859274 on Build GRCh38
rs762711232 Chr.14: 77859154-77859154 on Build GRCh38
rs767097940 Chr.14: 77859317-77859317 on Build GRCh38
rs746291372 Chr.14: 77859297-77859297 on Build GRCh38
rs772676200 Chr.14: 77859110-77859110 on Build GRCh38
rs376231729 Chr.14: 77859302-77859302 on Build GRCh38
rs1010971400 Chr.14: 77859361-77859361 on Build GRCh38
rs751108036 Chr.14: 77859172-77859172 on Build GRCh38
rs749868972 Chr.14: 77859255-77859255 on Build GRCh38
rs35409693 Chr.14: 77859411-77859412 on Build GRCh38
rs748309206 Chr.14: 77859211-77859211 on Build GRCh38
rs778016662 Chr.14: 77859289-77859289 on Build GRCh38
rs773925350 Chr.14: 77859218-77859218 on Build GRCh38
rs373018018 Chr.14: 77859244-77859244 on Build GRCh38
rs201149093 Chr.14: 77859230-77859230 on Build GRCh38
rs568153737 Chr.14: 77859248-77859248 on Build GRCh38
rs374671690 Chr.14: 77859124-77859124 on Build GRCh38
rs28498552 Chr.14: 77859332-77859332 on Build GRCh38
rs1037228013 Chr.14: 77859238-77859238 on Build GRCh38
rs369771535 Chr.14: 77859198-77859198 on Build GRCh38
rs763601911 Chr.14: 77859156-77859156 on Build GRCh38
rs762512719 Chr.14: 77859081-77859081 on Build GRCh38
rs779828368 Chr.14: 77859194-77859194 on Build GRCh38
rs750345682 Chr.14: 77859183-77859183 on Build GRCh38
rs771515697 Chr.14: 77859377-77859377 on Build GRCh38
rs774117602 Chr.14: 77859300-77859300 on Build GRCh38
rs768692475 Chr.14: 77859100-77859100 on Build GRCh38
rs763844933 Chr.14: 77859242-77859242 on Build GRCh38
rs978805861 Chr.14: 77859335-77859335 on Build GRCh38
rs757237522 Chr.14: 77859086-77859086 on Build GRCh38
rs372576994 Chr.14: 77859085-77859085 on Build GRCh38
rs12887455 Chr.14: 77859362-77859362 on Build GRCh38
rs7149884 Chr.14: 77859355-77859355 on Build GRCh38
rs142770287 Chr.14: 77859259-77859259 on Build GRCh38
rs139353105 Chr.14: 77859097-77859097 on Build GRCh38
rs140370982 Chr.14: 77859228-77859228 on Build GRCh38
rs761092410 Chr.14: 77859030-77859030 on Build GRCh38
rs889923682 Chr.14: 77859265-77859265 on Build GRCh38
rs780996595 Chr.14: 77859098-77859098 on Build GRCh38
rs748318021 Chr.14: 77859284-77859284 on Build GRCh38
rs185196719 Chr.14: 77859446-77859446 on Build GRCh38
rs969901879 Chr.14: 77859412-77859412 on Build GRCh38
rs955893963 Chr.14: 77859160-77859160 on Build GRCh38
rs913331049 Chr.14: 77859009-77859009 on Build GRCh38
rs532827034 Chr.14: 77859128-77859128 on Build GRCh38
rs200082195 Chr.14: 77859190-77859190 on Build GRCh38
rs770040664 Chr.14: 77859298-77859298 on Build GRCh38
rs372253385 Chr.14: 77859062-77859062 on Build GRCh38
rs922054942 Chr.14: 77859059-77859059 on Build GRCh38
rs766265393 Chr.14: 77859260-77859260 on Build GRCh38
rs149221192 Chr.14: 77859231-77859231 on Build GRCh38
rs1036057735 Chr.14: 77859276-77859276 on Build GRCh38
rs373310949 Chr.14: 77859167-77859167 on Build GRCh38
rs777345477 Chr.14: 77859076-77859076 on Build GRCh38
rs539690246 Chr.14: 77859348-77859348 on Build GRCh38
rs747041005 Chr.14: 77859295-77859295 on Build GRCh38
rs376615066 Chr.14: 77859191-77859191 on Build GRCh38
rs773835223 Chr.14: 77859114-77859114 on Build GRCh38
rs552612645 Chr.14: 77859197-77859197 on Build GRCh38
rs576143697 Chr.14: 77859360-77859360 on Build GRCh38

View Full Product Details