Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91273442-91273986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC88C
Entrez Gene ID
440193
Gene Name
coiled-coil domain containing 88C
Gene Aliases
DAPLE, HKRP2, KIAA1509, SCA40
UniGene
Hs.525536
Gene Chromosome Location
Chr.14: 91271323-91418498 on Build GRCh38
Transcripts
NM_001080414.3 XM_011536796.2 XM_017021336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6622613 c.804G>A p.P268P Chr.14: 91273480-91273480 on Build GRCh38
5148339 c.5191C>T p.P1731S Chr.14: 91273521-91273521 on Build GRCh38
2251209 c.791C>T p.S264L Chr.14: 91273493-91273493 on Build GRCh38
2251217 c.5132G>T p.G1711V Chr.14: 91273580-91273580 on Build GRCh38
2251218 c.704G>T p.G235V Chr.14: 91273580-91273580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113441106 Chr.14: 91273947-91273947 on Build GRCh38
rs77154172 Chr.14: 91273625-91273625 on Build GRCh38
rs747213669 Chr.14: 91273471-91273471 on Build GRCh38
rs368545903 Chr.14: 91273636-91273636 on Build GRCh38
rs369927918 Chr.14: 91273540-91273540 on Build GRCh38
rs868026711 Chr.14: 91273961-91273961 on Build GRCh38
rs761239537 Chr.14: 91273489-91273489 on Build GRCh38
rs978648901 Chr.14: 91273781-91273781 on Build GRCh38
rs764772042 Chr.14: 91273638-91273638 on Build GRCh38
rs565301599 Chr.14: 91273835-91273835 on Build GRCh38
rs202069805 Chr.14: 91273504-91273504 on Build GRCh38
rs551561240 Chr.14: 91273795-91273795 on Build GRCh38
rs201467365 Chr.14: 91273535-91273535 on Build GRCh38
rs746019915 Chr.14: 91273602-91273602 on Build GRCh38
rs766410191 Chr.14: 91273667-91273668 on Build GRCh38
rs905988380 Chr.14: 91273820-91273820 on Build GRCh38
rs777866370 Chr.14: 91273699-91273699 on Build GRCh38
rs759992639 Chr.14: 91273567-91273567 on Build GRCh38
rs766982420 Chr.14: 91273490-91273490 on Build GRCh38
rs184130248 Chr.14: 91273850-91273850 on Build GRCh38
rs745827807 Chr.14: 91273725-91273725 on Build GRCh38
rs940902796 Chr.14: 91273890-91273890 on Build GRCh38
rs767541596 Chr.14: 91273676-91273676 on Build GRCh38
rs895512913 Chr.14: 91273876-91273876 on Build GRCh38
rs925392159 Chr.14: 91273722-91273722 on Build GRCh38
rs772831151 Chr.14: 91273549-91273549 on Build GRCh38
rs533593750 Chr.14: 91273910-91273910 on Build GRCh38
rs992578901 Chr.14: 91273822-91273822 on Build GRCh38
rs757330043 Chr.14: 91273675-91273675 on Build GRCh38
rs374780136 Chr.14: 91273505-91273505 on Build GRCh38
rs369187490 Chr.14: 91273515-91273515 on Build GRCh38
rs774327623 Chr.14: 91273487-91273487 on Build GRCh38
rs760843998 Chr.14: 91273551-91273553 on Build GRCh38
rs200543687 Chr.14: 91273470-91273470 on Build GRCh38
rs199730476 Chr.14: 91273600-91273600 on Build GRCh38
rs752001413 Chr.14: 91273666-91273666 on Build GRCh38
rs183319966 Chr.14: 91273551-91273551 on Build GRCh38
rs963411509 Chr.14: 91273571-91273571 on Build GRCh38
rs749104245 Chr.14: 91273769-91273769 on Build GRCh38
rs371067583 Chr.14: 91273681-91273681 on Build GRCh38
rs758118736 Chr.14: 91273693-91273693 on Build GRCh38
rs769800189 Chr.14: 91273735-91273735 on Build GRCh38
rs770508496 Chr.14: 91273634-91273634 on Build GRCh38
rs755390490 Chr.14: 91273495-91273495 on Build GRCh38
rs899438095 Chr.14: 91273904-91273904 on Build GRCh38
rs200409556 Chr.14: 91273596-91273596 on Build GRCh38
rs766112055 Chr.14: 91273493-91273493 on Build GRCh38
rs762800401 Chr.14: 91273663-91273664 on Build GRCh38
rs139128801 Chr.14: 91273759-91273759 on Build GRCh38
rs944010898 Chr.14: 91273749-91273749 on Build GRCh38
rs768365233 Chr.14: 91273610-91273610 on Build GRCh38
rs1020236928 Chr.14: 91273765-91273765 on Build GRCh38
rs781402428 Chr.14: 91273601-91273601 on Build GRCh38
rs753528597 Chr.14: 91273565-91273565 on Build GRCh38
rs746877587 Chr.14: 91273702-91273702 on Build GRCh38
rs866025460 Chr.14: 91273833-91273833 on Build GRCh38
rs750663602 Chr.14: 91273679-91273679 on Build GRCh38
rs771181468 Chr.14: 91273514-91273514 on Build GRCh38
rs543258629 Chr.14: 91273780-91273780 on Build GRCh38
rs764178205 Chr.14: 91273591-91273592 on Build GRCh38
rs759225705 Chr.14: 91273637-91273637 on Build GRCh38
rs7145583 Chr.14: 91273529-91273529 on Build GRCh38
rs193276761 Chr.14: 91273632-91273632 on Build GRCh38
rs368661534 Chr.14: 91273500-91273500 on Build GRCh38
rs965245351 Chr.14: 91273746-91273746 on Build GRCh38
rs74474131 Chr.14: 91273689-91273689 on Build GRCh38
rs377167904 Chr.14: 91273573-91273573 on Build GRCh38
rs772301698 Chr.14: 91273477-91273478 on Build GRCh38
rs768378092 Chr.14: 91273523-91273523 on Build GRCh38
rs954161998 Chr.14: 91273724-91273724 on Build GRCh38
rs904432160 Chr.14: 91273917-91273917 on Build GRCh38
rs762705123 Chr.14: 91273481-91273481 on Build GRCh38
rs374091647 Chr.14: 91273776-91273776 on Build GRCh38
rs1036924191 Chr.14: 91273897-91273897 on Build GRCh38
rs374066841 Chr.14: 91273492-91273492 on Build GRCh38
rs571896627 Chr.14: 91273787-91273787 on Build GRCh38
rs761768016 Chr.14: 91273531-91273531 on Build GRCh38
rs1000079188 Chr.14: 91273925-91273925 on Build GRCh38
rs1037271673 Chr.14: 91273753-91273753 on Build GRCh38
rs771600264 Chr.14: 91273852-91273852 on Build GRCh38
rs763221917 Chr.14: 91273656-91273656 on Build GRCh38
rs776631908 Chr.14: 91273657-91273658 on Build GRCh38
rs775727347 Chr.14: 91273497-91273498 on Build GRCh38
rs531867565 Chr.14: 91273794-91273794 on Build GRCh38
rs780111091 Chr.14: 91273687-91273687 on Build GRCh38
rs370005900 Chr.14: 91273584-91273584 on Build GRCh38
rs1056870961 Chr.14: 91273658-91273658 on Build GRCh38
rs758992342 Chr.14: 91273585-91273585 on Build GRCh38
rs1019413085 Chr.14: 91273736-91273736 on Build GRCh38
rs146391250 Chr.14: 91273599-91273599 on Build GRCh38
rs200602954 Chr.14: 91273646-91273646 on Build GRCh38
rs1036654091 Chr.14: 91273688-91273688 on Build GRCh38
rs571920001 Chr.14: 91273657-91273657 on Build GRCh38
rs551793070 Chr.14: 91273516-91273516 on Build GRCh38
rs574962577 Chr.14: 91273503-91273503 on Build GRCh38
rs766205214 Chr.14: 91273562-91273562 on Build GRCh38
rs778723151 Chr.14: 91273619-91273619 on Build GRCh38
rs373485982 Chr.14: 91273622-91273622 on Build GRCh38
rs766317492 Chr.14: 91273713-91273713 on Build GRCh38
rs113563246 Chr.14: 91273729-91273729 on Build GRCh38
rs775312368 Chr.14: 91273480-91273480 on Build GRCh38
rs201393695 Chr.14: 91273626-91273626 on Build GRCh38

View Full Product Details