Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91283206-91283722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC88C
Entrez Gene ID
440193
Gene Name
coiled-coil domain containing 88C
Gene Aliases
DAPLE, HKRP2, KIAA1509, SCA40
UniGene
Hs.525536
Gene Chromosome Location
Chr.14: 91271323-91418498 on Build GRCh38
Transcripts
NM_001080414.3 XM_011536796.2 XM_017021335.1 XM_017021336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6293625 c.4524G>A p.M1508I Chr.14: 91283435-91283435 on Build GRCh38
5743674 c.181C>T p.R61W Chr.14: 91283350-91283350 on Build GRCh38
3815501 c.4613A>C p.H1538P Chr.14: 91283346-91283346 on Build GRCh38
3886505 c.182G>A p.R61Q Chr.14: 91283349-91283349 on Build GRCh38
1740087 c.4520C>T p.A1507V Chr.14: 91283439-91283439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008457693 Chr.14: 91283549-91283549 on Build GRCh38
rs763495585 Chr.14: 91283361-91283361 on Build GRCh38
rs975722700 Chr.14: 91283324-91283324 on Build GRCh38
rs758450000 Chr.14: 91283391-91283391 on Build GRCh38
rs926521187 Chr.14: 91283406-91283406 on Build GRCh38
rs778915447 Chr.14: 91283328-91283328 on Build GRCh38
rs920125235 Chr.14: 91283486-91283486 on Build GRCh38
rs745347761 Chr.14: 91283500-91283500 on Build GRCh38
rs766593910 Chr.14: 91283455-91283455 on Build GRCh38
rs1001977948 Chr.14: 91283671-91283671 on Build GRCh38
rs1045252573 Chr.14: 91283252-91283252 on Build GRCh38
rs761956102 Chr.14: 91283432-91283432 on Build GRCh38
rs753090408 Chr.14: 91283285-91283285 on Build GRCh38
rs751704870 Chr.14: 91283370-91283370 on Build GRCh38
rs750542948 Chr.14: 91283376-91283376 on Build GRCh38
rs2277509 Chr.14: 91283251-91283251 on Build GRCh38
rs745458331 Chr.14: 91283407-91283407 on Build GRCh38
rs201414940 Chr.14: 91283380-91283380 on Build GRCh38
rs903511128 Chr.14: 91283594-91283595 on Build GRCh38
rs955282665 Chr.14: 91283637-91283637 on Build GRCh38
rs1021876421 Chr.14: 91283298-91283298 on Build GRCh38
rs368026448 Chr.14: 91283356-91283356 on Build GRCh38
rs371149133 Chr.14: 91283288-91283288 on Build GRCh38
rs550665064 Chr.14: 91283634-91283634 on Build GRCh38
rs369071126 Chr.14: 91283420-91283420 on Build GRCh38
rs769395513 Chr.14: 91283545-91283545 on Build GRCh38
rs8014683 Chr.14: 91283297-91283297 on Build GRCh38
rs760856363 Chr.14: 91283447-91283447 on Build GRCh38
rs1021930244 Chr.14: 91283561-91283561 on Build GRCh38
rs939016107 Chr.14: 91283237-91283237 on Build GRCh38
rs573909474 Chr.14: 91283553-91283553 on Build GRCh38
rs374694567 Chr.14: 91283385-91283385 on Build GRCh38
rs1053533907 Chr.14: 91283550-91283550 on Build GRCh38
rs763296075 Chr.14: 91283415-91283415 on Build GRCh38
rs886844121 Chr.14: 91283246-91283246 on Build GRCh38
rs1015411185 Chr.14: 91283676-91283676 on Build GRCh38
rs377091200 Chr.14: 91283687-91283687 on Build GRCh38
rs905344036 Chr.14: 91283260-91283260 on Build GRCh38
rs372277532 Chr.14: 91283374-91283374 on Build GRCh38
rs562720610 Chr.14: 91283596-91283596 on Build GRCh38
rs267604088 Chr.14: 91283375-91283375 on Build GRCh38
rs780195795 Chr.14: 91283678-91283678 on Build GRCh38
rs1003315627 Chr.14: 91283286-91283286 on Build GRCh38
rs1021711496 Chr.14: 91283638-91283638 on Build GRCh38
rs748061020 Chr.14: 91283337-91283337 on Build GRCh38
rs749164277 Chr.14: 91283320-91283320 on Build GRCh38
rs1042147548 Chr.14: 91283686-91283686 on Build GRCh38
rs752609625 Chr.14: 91283463-91283463 on Build GRCh38
rs767457119 Chr.14: 91283446-91283446 on Build GRCh38
rs955381657 Chr.14: 91283340-91283340 on Build GRCh38
rs559097659 Chr.14: 91283688-91283688 on Build GRCh38
rs777852000 Chr.14: 91283395-91283395 on Build GRCh38
rs542531485 Chr.14: 91283560-91283560 on Build GRCh38
rs780613012 Chr.14: 91283493-91283493 on Build GRCh38
rs369301673 Chr.14: 91283482-91283482 on Build GRCh38
rs764550186 Chr.14: 91283364-91283364 on Build GRCh38
rs781265445 Chr.14: 91283309-91283309 on Build GRCh38
rs776085267 Chr.14: 91283358-91283358 on Build GRCh38
rs752041919 Chr.14: 91283341-91283341 on Build GRCh38
rs778526185 Chr.14: 91283383-91283383 on Build GRCh38
rs769462542 Chr.14: 91283408-91283408 on Build GRCh38
rs1056603965 Chr.14: 91283437-91283437 on Build GRCh38
rs375420508 Chr.14: 91283353-91283353 on Build GRCh38
rs3950029 Chr.14: 91283421-91283421 on Build GRCh38
rs891397660 Chr.14: 91283540-91283540 on Build GRCh38
rs967956130 Chr.14: 91283316-91283316 on Build GRCh38
rs200878782 Chr.14: 91283349-91283349 on Build GRCh38
rs531364925 Chr.14: 91283600-91283600 on Build GRCh38
rs952140720 Chr.14: 91283693-91283693 on Build GRCh38
rs141037411 Chr.14: 91283677-91283677 on Build GRCh38
rs746579410 Chr.14: 91283397-91283397 on Build GRCh38
rs754671206 Chr.14: 91283322-91283322 on Build GRCh38
rs757507757 Chr.14: 91283299-91283299 on Build GRCh38
rs1008218713 Chr.14: 91283310-91283310 on Build GRCh38
rs780905563 Chr.14: 91283405-91283405 on Build GRCh38
rs1056237410 Chr.14: 91283238-91283238 on Build GRCh38
rs374283893 Chr.14: 91283294-91283294 on Build GRCh38
rs777868444 Chr.14: 91283479-91283479 on Build GRCh38
rs374862964 Chr.14: 91283464-91283464 on Build GRCh38
rs770292577 Chr.14: 91283398-91283398 on Build GRCh38
rs534378366 Chr.14: 91283663-91283663 on Build GRCh38
rs777439207 Chr.14: 91283314-91283314 on Build GRCh38
rs775868329 Chr.14: 91283409-91283409 on Build GRCh38
rs920620878 Chr.14: 91283690-91283690 on Build GRCh38
rs1050034163 Chr.14: 91283511-91283511 on Build GRCh38
rs151102993 Chr.14: 91283338-91283338 on Build GRCh38
rs755780759 Chr.14: 91283290-91283290 on Build GRCh38
rs992049827 Chr.14: 91283684-91283684 on Build GRCh38
rs765476380 Chr.14: 91283354-91283354 on Build GRCh38
rs967876505 Chr.14: 91283655-91283655 on Build GRCh38
rs770687824 Chr.14: 91283350-91283350 on Build GRCh38
rs776137082 Chr.14: 91283352-91283352 on Build GRCh38
rs891156860 Chr.14: 91283303-91283303 on Build GRCh38

View Full Product Details