Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92059013-92059540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN3
Entrez Gene ID
4287
Gene Name
ataxin 3
Gene Aliases
AT3, ATX3, JOS, MJD, MJD1, SCA3
UniGene
Hs.532632
Gene Chromosome Location
Chr.14: 92058552-92106621 on Build GRCh38
Transcripts
NM_001127696.1 NM_001127697.2 NM_001164774.1 NM_001164776.1 NM_001164777.1 NM_001164778.1 NM_001164779.1 NM_001164780.1 NM_001164781.1 NM_001164782.1 NM_004993.5 NM_030660.4 NR_028453.1 NR_028454.1 NR_028455.1 NR_028456.1 NR_028457.1 NR_028458.1 NR_028459.1 NR_028460.1 NR_028461.1 NR_028462.1 NR_028463.1 NR_028464.1 NR_028465.1 NR_028466.1 NR_028467.1 NR_028468.1 NR_028469.1 NR_028470.1 NR_031765.1

SNP Detail

SNP ID Chromosome Location
rs541880574 Chr.14: 92059179-92059179 on Build GRCh38
rs773379500 Chr.14: 92059412-92059412 on Build GRCh38
rs10151945 Chr.14: 92059269-92059269 on Build GRCh38
rs919597255 Chr.14: 92059505-92059505 on Build GRCh38
rs758956940 Chr.14: 92059137-92059137 on Build GRCh38
rs1011882164 Chr.14: 92059303-92059303 on Build GRCh38
rs890996851 Chr.14: 92059261-92059261 on Build GRCh38
rs939459070 Chr.14: 92059041-92059041 on Build GRCh38
rs751097712 Chr.14: 92059252-92059252 on Build GRCh38
rs760852869 Chr.14: 92059489-92059489 on Build GRCh38
rs532290325 Chr.14: 92059333-92059333 on Build GRCh38
rs991510077 Chr.14: 92059145-92059145 on Build GRCh38
rs765591204 Chr.14: 92059075-92059075 on Build GRCh38
rs116994678 Chr.14: 92059399-92059399 on Build GRCh38
rs940299181 Chr.14: 92059432-92059432 on Build GRCh38
rs924465521 Chr.14: 92059312-92059312 on Build GRCh38
rs891693461 Chr.14: 92059274-92059274 on Build GRCh38
rs749214501 Chr.14: 92059371-92059371 on Build GRCh38
rs192385527 Chr.14: 92059315-92059315 on Build GRCh38
rs960895221 Chr.14: 92059403-92059403 on Build GRCh38
rs908682915 Chr.14: 92059408-92059408 on Build GRCh38
rs983294998 Chr.14: 92059110-92059110 on Build GRCh38
rs992324297 Chr.14: 92059404-92059404 on Build GRCh38
rs939707405 Chr.14: 92059093-92059093 on Build GRCh38
rs554732515 Chr.14: 92059238-92059238 on Build GRCh38
rs781437447 Chr.14: 92059295-92059303 on Build GRCh38
rs992855906 Chr.14: 92059063-92059063 on Build GRCh38
rs74244591 Chr.14: 92059218-92059218 on Build GRCh38
rs8022758 Chr.14: 92059127-92059127 on Build GRCh38
rs974696530 Chr.14: 92059503-92059503 on Build GRCh38
rs976935141 Chr.14: 92059050-92059050 on Build GRCh38
rs369630957 Chr.14: 92059270-92059273 on Build GRCh38
rs9652396 Chr.14: 92059273-92059273 on Build GRCh38
rs769626047 Chr.14: 92059323-92059323 on Build GRCh38
rs144799309 Chr.14: 92059107-92059107 on Build GRCh38
rs909954199 Chr.14: 92059123-92059123 on Build GRCh38
rs892907106 Chr.14: 92059386-92059386 on Build GRCh38
rs932317645 Chr.14: 92059517-92059517 on Build GRCh38

View Full Product Details