Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92061329-92061876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN3
Entrez Gene ID
4287
Gene Name
ataxin 3
Gene Aliases
AT3, ATX3, JOS, MJD, MJD1, SCA3
UniGene
Hs.532632
Gene Chromosome Location
Chr.14: 92058552-92106621 on Build GRCh38
Transcripts
NM_001127696.1 NM_001127697.2 NM_001164774.1 NM_001164776.1 NM_001164777.1 NM_001164778.1 NM_001164779.1 NM_001164780.1 NM_001164781.1 NM_001164782.1 NM_004993.5 NM_030660.4 NR_028453.1 NR_028454.1 NR_028455.1 NR_028456.1 NR_028457.1 NR_028458.1 NR_028459.1 NR_028460.1 NR_028461.1 NR_028462.1 NR_028463.1 NR_028464.1 NR_028465.1 NR_028466.1 NR_028467.1 NR_028468.1 NR_028469.1 NR_028470.1 NR_031765.1

SNP Detail

SNP ID Chromosome Location
rs577684465 Chr.14: 92061804-92061804 on Build GRCh38
rs56196346 Chr.14: 92061848-92061848 on Build GRCh38
rs911226054 Chr.14: 92061406-92061406 on Build GRCh38
rs1007473208 Chr.14: 92061774-92061774 on Build GRCh38
rs981790407 Chr.14: 92061420-92061420 on Build GRCh38
rs763317999 Chr.14: 92061665-92061665 on Build GRCh38
rs1015207948 Chr.14: 92061370-92061370 on Build GRCh38
rs947566830 Chr.14: 92061556-92061556 on Build GRCh38
rs11628764 Chr.14: 92061633-92061633 on Build GRCh38
rs546433513 Chr.14: 92061390-92061390 on Build GRCh38
rs190326858 Chr.14: 92061648-92061648 on Build GRCh38
rs993330134 Chr.14: 92061362-92061362 on Build GRCh38
rs940645317 Chr.14: 92061706-92061706 on Build GRCh38
rs936245295 Chr.14: 92061571-92061571 on Build GRCh38
rs890449980 Chr.14: 92061697-92061697 on Build GRCh38
rs960286246 Chr.14: 92061477-92061477 on Build GRCh38
rs188739788 Chr.14: 92061441-92061441 on Build GRCh38
rs1023369788 Chr.14: 92061389-92061390 on Build GRCh38
rs993629366 Chr.14: 92061663-92061663 on Build GRCh38
rs1027453634 Chr.14: 92061712-92061712 on Build GRCh38
rs976762294 Chr.14: 92061849-92061849 on Build GRCh38
rs922039176 Chr.14: 92061620-92061620 on Build GRCh38
rs1017469781 Chr.14: 92061823-92061823 on Build GRCh38
rs913414395 Chr.14: 92061524-92061524 on Build GRCh38
rs894185704 Chr.14: 92061617-92061617 on Build GRCh38
rs1045916278 Chr.14: 92061635-92061635 on Build GRCh38
rs769942768 Chr.14: 92061608-92061608 on Build GRCh38
rs768003087 Chr.14: 92061795-92061798 on Build GRCh38
rs922697284 Chr.14: 92061490-92061490 on Build GRCh38
rs951978650 Chr.14: 92061388-92061388 on Build GRCh38
rs926081364 Chr.14: 92061567-92061567 on Build GRCh38
rs1036261281 Chr.14: 92061708-92061708 on Build GRCh38
rs1023448773 Chr.14: 92061781-92061781 on Build GRCh38
rs186208016 Chr.14: 92061601-92061601 on Build GRCh38
rs573022747 Chr.14: 92061380-92061380 on Build GRCh38
rs532483134 Chr.14: 92061379-92061379 on Build GRCh38
rs371606868 Chr.14: 92061409-92061409 on Build GRCh38
rs375246638 Chr.14: 92061468-92061468 on Build GRCh38
rs764515154 Chr.14: 92061772-92061772 on Build GRCh38
rs1041928634 Chr.14: 92061546-92061546 on Build GRCh38
rs111461096 Chr.14: 92061494-92061494 on Build GRCh38

View Full Product Details