Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92063482-92063991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN3
Entrez Gene ID
4287
Gene Name
ataxin 3
Gene Aliases
AT3, ATX3, JOS, MJD, MJD1, SCA3
UniGene
Hs.532632
Gene Chromosome Location
Chr.14: 92058552-92106621 on Build GRCh38
Transcripts
NM_001127696.1 NM_001127697.2 NM_001164774.1 NM_001164776.1 NM_001164777.1 NM_001164778.1 NM_001164779.1 NM_001164780.1 NM_001164781.1 NM_001164782.1 NM_004993.5 NM_030660.4 NR_028453.1 NR_028454.1 NR_028455.1 NR_028456.1 NR_028457.1 NR_028458.1 NR_028459.1 NR_028460.1 NR_028461.1 NR_028462.1 NR_028463.1 NR_028464.1 NR_028465.1 NR_028466.1 NR_028467.1 NR_028468.1 NR_028469.1 NR_028470.1 NR_031765.1

SNP Detail

SNP ID Chromosome Location
rs55644237 Chr.14: 92063655-92063655 on Build GRCh38
rs1020917607 Chr.14: 92063788-92063788 on Build GRCh38
rs886641665 Chr.14: 92063683-92063683 on Build GRCh38
rs377668917 Chr.14: 92063801-92063801 on Build GRCh38
rs912325566 Chr.14: 92063875-92063875 on Build GRCh38
rs17127901 Chr.14: 92063834-92063834 on Build GRCh38
rs1044806469 Chr.14: 92063654-92063654 on Build GRCh38
rs777583007 Chr.14: 92063942-92063942 on Build GRCh38
rs977870565 Chr.14: 92063785-92063785 on Build GRCh38
rs879766263 Chr.14: 92063519-92063519 on Build GRCh38
rs896540864 Chr.14: 92063732-92063732 on Build GRCh38
rs529420712 Chr.14: 92063647-92063647 on Build GRCh38
rs752468952 Chr.14: 92063920-92063920 on Build GRCh38
rs965117753 Chr.14: 92063905-92063905 on Build GRCh38
rs1016492119 Chr.14: 92063693-92063693 on Build GRCh38
rs1032176949 Chr.14: 92063827-92063827 on Build GRCh38
rs878916285 Chr.14: 92063933-92063933 on Build GRCh38
rs947973660 Chr.14: 92063631-92063631 on Build GRCh38
rs17847274 Chr.14: 92063694-92063694 on Build GRCh38
rs1006330316 Chr.14: 92063685-92063685 on Build GRCh38
rs936482592 Chr.14: 92063532-92063532 on Build GRCh38
rs750893431 Chr.14: 92063799-92063800 on Build GRCh38
rs967038557 Chr.14: 92063811-92063811 on Build GRCh38
rs1047802392 Chr.14: 92063672-92063672 on Build GRCh38
rs529386181 Chr.14: 92063635-92063635 on Build GRCh38
rs560592057 Chr.14: 92063643-92063643 on Build GRCh38
rs910369 Chr.14: 92063938-92063938 on Build GRCh38
rs56396823 Chr.14: 92063606-92063606 on Build GRCh38
rs1055199489 Chr.14: 92063727-92063727 on Build GRCh38
rs969720921 Chr.14: 92063881-92063881 on Build GRCh38
rs900768680 Chr.14: 92063702-92063702 on Build GRCh38
rs180896627 Chr.14: 92063669-92063669 on Build GRCh38
rs954363063 Chr.14: 92063765-92063765 on Build GRCh38
rs140741824 Chr.14: 92063925-92063925 on Build GRCh38
rs916391569 Chr.14: 92063855-92063855 on Build GRCh38
rs78530327 Chr.14: 92063815-92063815 on Build GRCh38
rs985174078 Chr.14: 92063586-92063586 on Build GRCh38
rs932438693 Chr.14: 92063585-92063585 on Build GRCh38
rs754727174 Chr.14: 92063671-92063671 on Build GRCh38
rs989427695 Chr.14: 92063555-92063555 on Build GRCh38
rs1030744909 Chr.14: 92063808-92063808 on Build GRCh38
rs745890873 Chr.14: 92063961-92063961 on Build GRCh38
rs1030418274 Chr.14: 92063762-92063762 on Build GRCh38
rs938687030 Chr.14: 92063930-92063930 on Build GRCh38
rs985205164 Chr.14: 92063841-92063841 on Build GRCh38
rs996414658 Chr.14: 92063710-92063710 on Build GRCh38

View Full Product Details