Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95190495-95191002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLMN
Entrez Gene ID
79789
Gene Name
calmin
Gene Aliases
-
UniGene
Hs.301478
Gene Chromosome Location
Chr.14: 95181939-95319908 on Build GRCh38
Transcripts
NM_024734.3 XM_011537158.1 XM_017021646.1

SNP Detail

SNP ID Chromosome Location
rs985385351 Chr.14: 95190952-95190952 on Build GRCh38
rs902368198 Chr.14: 95190672-95190672 on Build GRCh38
rs919875708 Chr.14: 95190628-95190628 on Build GRCh38
rs573211401 Chr.14: 95190712-95190712 on Build GRCh38
rs565265187 Chr.14: 95190814-95190814 on Build GRCh38
rs553014914 Chr.14: 95190668-95190668 on Build GRCh38
rs536124176 Chr.14: 95190547-95190547 on Build GRCh38
rs115349753 Chr.14: 95190782-95190782 on Build GRCh38
rs914945770 Chr.14: 95190925-95190926 on Build GRCh38
rs146467023 Chr.14: 95190872-95190872 on Build GRCh38
rs188838283 Chr.14: 95190517-95190517 on Build GRCh38
rs1040478267 Chr.14: 95190726-95190726 on Build GRCh38
rs1049631596 Chr.14: 95190654-95190654 on Build GRCh38
rs545098613 Chr.14: 95190733-95190733 on Build GRCh38
rs193035161 Chr.14: 95190944-95190944 on Build GRCh38
rs774258037 Chr.14: 95190547-95190547 on Build GRCh38
rs961409399 Chr.14: 95190915-95190915 on Build GRCh38
rs952488229 Chr.14: 95190941-95190941 on Build GRCh38
rs556425952 Chr.14: 95190609-95190609 on Build GRCh38
rs1025806912 Chr.14: 95190849-95190849 on Build GRCh38
rs964374942 Chr.14: 95190915-95190916 on Build GRCh38
rs1024754444 Chr.14: 95190912-95190912 on Build GRCh38
rs554782476 Chr.14: 95190744-95190744 on Build GRCh38
rs767493473 Chr.14: 95190587-95190589 on Build GRCh38
rs931215288 Chr.14: 95190651-95190651 on Build GRCh38
rs962838069 Chr.14: 95190546-95190546 on Build GRCh38
rs1025857488 Chr.14: 95190552-95190552 on Build GRCh38
rs943049253 Chr.14: 95190604-95190604 on Build GRCh38
rs56119341 Chr.14: 95190579-95190579 on Build GRCh38
rs919759784 Chr.14: 95190925-95190925 on Build GRCh38
rs1001081772 Chr.14: 95190797-95190797 on Build GRCh38
rs939269070 Chr.14: 95190740-95190740 on Build GRCh38
rs1033310298 Chr.14: 95190551-95190551 on Build GRCh38
rs752532592 Chr.14: 95190784-95190784 on Build GRCh38
rs140680442 Chr.14: 95190669-95190669 on Build GRCh38
rs774228005 Chr.14: 95190922-95190922 on Build GRCh38
rs749465913 Chr.14: 95190773-95190773 on Build GRCh38
rs955634541 Chr.14: 95190569-95190569 on Build GRCh38
rs73333229 Chr.14: 95190539-95190539 on Build GRCh38
rs539692451 Chr.14: 95190877-95190877 on Build GRCh38
rs1034318918 Chr.14: 95190802-95190802 on Build GRCh38
rs1017033548 Chr.14: 95190544-95190544 on Build GRCh38
rs558591178 Chr.14: 95190783-95190783 on Build GRCh38
rs1018946399 Chr.14: 95190905-95190905 on Build GRCh38
rs369117848 Chr.14: 95190878-95190878 on Build GRCh38
rs572160733 Chr.14: 95190593-95190593 on Build GRCh38
rs911534034 Chr.14: 95190582-95190582 on Build GRCh38
rs144399247 Chr.14: 95190749-95190749 on Build GRCh38
rs994277423 Chr.14: 95190815-95190815 on Build GRCh38
rs145820165 Chr.14: 95190608-95190608 on Build GRCh38
rs552171604 Chr.14: 95190584-95190584 on Build GRCh38
rs987049084 Chr.14: 95190899-95190899 on Build GRCh38
rs544187168 Chr.14: 95190833-95190833 on Build GRCh38
rs1009725909 Chr.14: 95190756-95190756 on Build GRCh38
rs1016982462 Chr.14: 95190790-95190790 on Build GRCh38

View Full Product Details