Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95190865-95191389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLMN
Entrez Gene ID
79789
Gene Name
calmin
Gene Aliases
-
UniGene
Hs.301478
Gene Chromosome Location
Chr.14: 95181939-95319908 on Build GRCh38
Transcripts
NM_024734.3 XM_011537158.1 XM_017021646.1

SNP Detail

SNP ID Chromosome Location
rs919759784 Chr.14: 95190925-95190925 on Build GRCh38
rs985385351 Chr.14: 95190952-95190952 on Build GRCh38
rs1055405164 Chr.14: 95191066-95191066 on Build GRCh38
rs994226822 Chr.14: 95191060-95191060 on Build GRCh38
rs952541273 Chr.14: 95191232-95191232 on Build GRCh38
rs575887510 Chr.14: 95191030-95191030 on Build GRCh38
rs947992002 Chr.14: 95191049-95191049 on Build GRCh38
rs1013285983 Chr.14: 95191077-95191077 on Build GRCh38
rs992400358 Chr.14: 95191316-95191316 on Build GRCh38
rs749174871 Chr.14: 95191127-95191128 on Build GRCh38
rs774228005 Chr.14: 95190922-95190922 on Build GRCh38
rs542083352 Chr.14: 95191015-95191015 on Build GRCh38
rs566624759 Chr.14: 95191284-95191284 on Build GRCh38
rs943405506 Chr.14: 95191002-95191002 on Build GRCh38
rs373561764 Chr.14: 95191204-95191204 on Build GRCh38
rs914945770 Chr.14: 95190925-95190926 on Build GRCh38
rs140401571 Chr.14: 95191353-95191353 on Build GRCh38
rs1021902649 Chr.14: 95191243-95191243 on Build GRCh38
rs560281277 Chr.14: 95191022-95191022 on Build GRCh38
rs1018946399 Chr.14: 95190905-95190905 on Build GRCh38
rs148186555 Chr.14: 95191254-95191254 on Build GRCh38
rs1042421524 Chr.14: 95191033-95191033 on Build GRCh38
rs112773983 Chr.14: 95191026-95191026 on Build GRCh38
rs193035161 Chr.14: 95190944-95190944 on Build GRCh38
rs952488229 Chr.14: 95190941-95190941 on Build GRCh38
rs961409399 Chr.14: 95190915-95190915 on Build GRCh38
rs911105161 Chr.14: 95190998-95190998 on Build GRCh38
rs960521568 Chr.14: 95191289-95191289 on Build GRCh38
rs1024754444 Chr.14: 95190912-95190912 on Build GRCh38
rs964374942 Chr.14: 95190915-95190916 on Build GRCh38
rs1025059063 Chr.14: 95191083-95191083 on Build GRCh38
rs987049084 Chr.14: 95190899-95190899 on Build GRCh38
rs558936101 Chr.14: 95191362-95191362 on Build GRCh38
rs916441339 Chr.14: 95191324-95191324 on Build GRCh38
rs1018476882 Chr.14: 95191194-95191194 on Build GRCh38
rs773254295 Chr.14: 95191031-95191031 on Build GRCh38
rs140864411 Chr.14: 95191052-95191052 on Build GRCh38
rs377279072 Chr.14: 95191255-95191255 on Build GRCh38
rs765797806 Chr.14: 95191120-95191120 on Build GRCh38
rs143029831 Chr.14: 95191059-95191059 on Build GRCh38
rs1047609576 Chr.14: 95191100-95191100 on Build GRCh38
rs568487537 Chr.14: 95191134-95191134 on Build GRCh38
rs1026703463 Chr.14: 95191217-95191217 on Build GRCh38
rs916477573 Chr.14: 95191034-95191034 on Build GRCh38
rs923687784 Chr.14: 95191275-95191275 on Build GRCh38
rs41286554 Chr.14: 95191070-95191070 on Build GRCh38

View Full Product Details