Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96289577-96290082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG2B
Entrez Gene ID
55102
Gene Name
autophagy related 2B
Gene Aliases
C14orf103
UniGene
Hs.168241
Gene Chromosome Location
Chr.14: 96281258-96363341 on Build GRCh38
Transcripts
XM_006720187.2 XM_017021397.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3499568 c.5907C>T p.I1969I Chr.14: 96289755-96289755 on Build GRCh38
7356813 c.5938C>T p.H1980Y Chr.14: 96289724-96289724 on Build GRCh38
959362 c.5869A>G p.T1957A Chr.14: 96289793-96289793 on Build GRCh38
4053348 c.5947G>A p.A1983T Chr.14: 96289715-96289715 on Build GRCh38
959363 c.5857G>A p.A1953T Chr.14: 96289805-96289805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748454114 Chr.14: 96289620-96289620 on Build GRCh38
rs773343738 Chr.14: 96289853-96289854 on Build GRCh38
rs373233271 Chr.14: 96289844-96289845 on Build GRCh38
rs141284028 Chr.14: 96289697-96289697 on Build GRCh38
rs1039065888 Chr.14: 96290010-96290010 on Build GRCh38
rs1022731263 Chr.14: 96289885-96289885 on Build GRCh38
rs757463182 Chr.14: 96289775-96289775 on Build GRCh38
rs991221908 Chr.14: 96289864-96289864 on Build GRCh38
rs758833442 Chr.14: 96289754-96289754 on Build GRCh38
rs771044543 Chr.14: 96289665-96289665 on Build GRCh38
rs1039119606 Chr.14: 96289686-96289686 on Build GRCh38
rs144991946 Chr.14: 96289721-96289721 on Build GRCh38
rs368626903 Chr.14: 96289641-96289641 on Build GRCh38
rs755534743 Chr.14: 96289616-96289616 on Build GRCh38
rs767198930 Chr.14: 96289936-96289936 on Build GRCh38
rs934559546 Chr.14: 96289955-96289955 on Build GRCh38
rs963014241 Chr.14: 96289898-96289898 on Build GRCh38
rs753035063 Chr.14: 96289743-96289743 on Build GRCh38
rs753330534 Chr.14: 96289723-96289723 on Build GRCh38
rs760210708 Chr.14: 96290033-96290033 on Build GRCh38
rs973479735 Chr.14: 96289909-96289909 on Build GRCh38
rs892911100 Chr.14: 96289687-96289687 on Build GRCh38
rs1044102182 Chr.14: 96289704-96289704 on Build GRCh38
rs778870296 Chr.14: 96289841-96289841 on Build GRCh38
rs768504093 Chr.14: 96289802-96289802 on Build GRCh38
rs912044729 Chr.14: 96289978-96289978 on Build GRCh38
rs189894249 Chr.14: 96289927-96289927 on Build GRCh38
rs765725852 Chr.14: 96289733-96289733 on Build GRCh38
rs747197195 Chr.14: 96289650-96289650 on Build GRCh38
rs774148982 Chr.14: 96289819-96289819 on Build GRCh38
rs558564578 Chr.14: 96289868-96289868 on Build GRCh38
rs146775079 Chr.14: 96289730-96289730 on Build GRCh38
rs35258365 Chr.14: 96289757-96289757 on Build GRCh38
rs921321897 Chr.14: 96289880-96289880 on Build GRCh38
rs267604117 Chr.14: 96289755-96289755 on Build GRCh38
rs556980719 Chr.14: 96289767-96289767 on Build GRCh38
rs540748828 Chr.14: 96289647-96289647 on Build GRCh38
rs754447193 Chr.14: 96289614-96289614 on Build GRCh38
rs114552965 Chr.14: 96289988-96289988 on Build GRCh38
rs760382963 Chr.14: 96289720-96289720 on Build GRCh38
rs560141417 Chr.14: 96289618-96289618 on Build GRCh38
rs768575219 Chr.14: 96289681-96289681 on Build GRCh38
rs185816070 Chr.14: 96289678-96289678 on Build GRCh38
rs201507342 Chr.14: 96289711-96289711 on Build GRCh38
rs565686611 Chr.14: 96289860-96289860 on Build GRCh38
rs144845407 Chr.14: 96289983-96289983 on Build GRCh38
rs35575967 Chr.14: 96289848-96289849 on Build GRCh38
rs774034138 Chr.14: 96289683-96289683 on Build GRCh38
rs552242263 Chr.14: 96289674-96289674 on Build GRCh38
rs781256255 Chr.14: 96289779-96289779 on Build GRCh38
rs1044540927 Chr.14: 96290004-96290004 on Build GRCh38
rs149862692 Chr.14: 96289977-96289977 on Build GRCh38
rs77065927 Chr.14: 96289613-96289613 on Build GRCh38
rs549682225 Chr.14: 96289716-96289716 on Build GRCh38
rs375669692 Chr.14: 96289995-96289995 on Build GRCh38
rs758570961 Chr.14: 96289622-96289622 on Build GRCh38
rs533925935 Chr.14: 96289861-96289861 on Build GRCh38
rs929541357 Chr.14: 96289973-96289973 on Build GRCh38
rs745893854 Chr.14: 96289791-96289791 on Build GRCh38
rs912495958 Chr.14: 96289950-96289950 on Build GRCh38
rs954190663 Chr.14: 96289889-96289889 on Build GRCh38
rs376093834 Chr.14: 96289632-96289632 on Build GRCh38
rs1015759609 Chr.14: 96289840-96289840 on Build GRCh38
rs914397909 Chr.14: 96289982-96289982 on Build GRCh38
rs866337318 Chr.14: 96289705-96289705 on Build GRCh38
rs370468248 Chr.14: 96289619-96289619 on Build GRCh38
rs527274217 Chr.14: 96289668-96289668 on Build GRCh38

View Full Product Details