Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99169988-99170492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL11B
Entrez Gene ID
64919
Gene Name
B-cell CLL/lymphoma 11B
Gene Aliases
ATL1, ATL1-alpha, ATL1-beta, ATL1-delta, ATL1-gamma, CTIP-2, CTIP2, RIT1, ZNF856B, hRIT1-alpha
UniGene
Hs.709690
Gene Chromosome Location
Chr.14: 99169287-99271713 on Build GRCh38
Transcripts
NM_001282237.1 NM_001282238.1 NM_022898.2 NM_138576.3

SNP Detail

SNP ID Chromosome Location
rs1016028799 Chr.14: 99170120-99170120 on Build GRCh38
rs779651873 Chr.14: 99170320-99170320 on Build GRCh38
rs993223693 Chr.14: 99170266-99170266 on Build GRCh38
rs72702620 Chr.14: 99170058-99170058 on Build GRCh38
rs574941330 Chr.14: 99170068-99170068 on Build GRCh38
rs1025316707 Chr.14: 99170185-99170185 on Build GRCh38
rs1026666754 Chr.14: 99170249-99170249 on Build GRCh38
rs868399177 Chr.14: 99170052-99170052 on Build GRCh38
rs1043228784 Chr.14: 99170309-99170309 on Build GRCh38
rs969163076 Chr.14: 99170195-99170195 on Build GRCh38
rs919165055 Chr.14: 99170260-99170260 on Build GRCh38
rs1032345555 Chr.14: 99170059-99170059 on Build GRCh38
rs932808776 Chr.14: 99170056-99170056 on Build GRCh38
rs531680931 Chr.14: 99170310-99170310 on Build GRCh38
rs953236069 Chr.14: 99170086-99170086 on Build GRCh38
rs963186277 Chr.14: 99170166-99170166 on Build GRCh38
rs1030383934 Chr.14: 99170448-99170448 on Build GRCh38
rs1053332328 Chr.14: 99170333-99170333 on Build GRCh38
rs943741557 Chr.14: 99170262-99170262 on Build GRCh38
rs998477501 Chr.14: 99170370-99170370 on Build GRCh38
rs980583656 Chr.14: 99170196-99170196 on Build GRCh38
rs1031660009 Chr.14: 99170085-99170085 on Build GRCh38
rs367775311 Chr.14: 99170205-99170205 on Build GRCh38
rs971921963 Chr.14: 99170206-99170206 on Build GRCh38
rs140410144 Chr.14: 99170116-99170116 on Build GRCh38
rs375744382 Chr.14: 99170210-99170210 on Build GRCh38
rs901461517 Chr.14: 99170062-99170062 on Build GRCh38
rs952444756 Chr.14: 99170259-99170259 on Build GRCh38
rs377670037 Chr.14: 99170344-99170344 on Build GRCh38
rs142778175 Chr.14: 99170295-99170295 on Build GRCh38
rs572185309 Chr.14: 99170148-99170148 on Build GRCh38
rs191005530 Chr.14: 99170353-99170353 on Build GRCh38
rs895840215 Chr.14: 99170101-99170101 on Build GRCh38
rs765813862 Chr.14: 99170219-99170219 on Build GRCh38
rs545826280 Chr.14: 99170262-99170262 on Build GRCh38
rs934805316 Chr.14: 99170325-99170325 on Build GRCh38
rs934753808 Chr.14: 99170349-99170349 on Build GRCh38
rs948758810 Chr.14: 99170297-99170297 on Build GRCh38
rs1043216052 Chr.14: 99170038-99170038 on Build GRCh38
rs997591919 Chr.14: 99170063-99170063 on Build GRCh38
rs549811281 Chr.14: 99170342-99170342 on Build GRCh38
rs147127382 Chr.14: 99170072-99170072 on Build GRCh38
rs1013503554 Chr.14: 99170139-99170139 on Build GRCh38

View Full Product Details