Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99174590-99175079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL11B
Entrez Gene ID
64919
Gene Name
B-cell CLL/lymphoma 11B
Gene Aliases
ATL1, ATL1-alpha, ATL1-beta, ATL1-delta, ATL1-gamma, CTIP-2, CTIP2, RIT1, ZNF856B, hRIT1-alpha
UniGene
Hs.709690
Gene Chromosome Location
Chr.14: 99169287-99271713 on Build GRCh38
Transcripts
NM_001282237.1 NM_001282238.1 NM_022898.2 NM_138576.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4053383 c.1768G>A p.E590K Chr.14: 99175068-99175068 on Build GRCh38
4053379 c.2099C>T p.A700V Chr.14: 99174737-99174737 on Build GRCh38
5521542 c.2108C>T p.P703L Chr.14: 99174728-99174728 on Build GRCh38
4053381 c.1891G>A p.G631R Chr.14: 99174945-99174945 on Build GRCh38
1718148 c.2025G>A p.P675P Chr.14: 99174811-99174811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754081029 Chr.14: 99174612-99174612 on Build GRCh38
rs983403866 Chr.14: 99174889-99174889 on Build GRCh38
rs752763028 Chr.14: 99174628-99174628 on Build GRCh38
rs760277241 Chr.14: 99174929-99174930 on Build GRCh38
rs753747298 Chr.14: 99174859-99174859 on Build GRCh38
rs369506800 Chr.14: 99174991-99174991 on Build GRCh38
rs761140637 Chr.14: 99174689-99174689 on Build GRCh38
rs200197315 Chr.14: 99174698-99174698 on Build GRCh38
rs946484398 Chr.14: 99174928-99174928 on Build GRCh38
rs755998847 Chr.14: 99174722-99174722 on Build GRCh38
rs561461377 Chr.14: 99174931-99174931 on Build GRCh38
rs752477920 Chr.14: 99174901-99174901 on Build GRCh38
rs549637836 Chr.14: 99174924-99174924 on Build GRCh38
rs1056645820 Chr.14: 99174932-99174932 on Build GRCh38
rs765002731 Chr.14: 99174871-99174871 on Build GRCh38
rs779752788 Chr.14: 99174672-99174672 on Build GRCh38
rs867661023 Chr.14: 99175061-99175061 on Build GRCh38
rs761923052 Chr.14: 99174988-99174988 on Build GRCh38
rs773857239 Chr.14: 99174682-99174682 on Build GRCh38
rs778489263 Chr.14: 99174730-99174730 on Build GRCh38
rs1024245996 Chr.14: 99175026-99175026 on Build GRCh38
rs902748229 Chr.14: 99174812-99174812 on Build GRCh38
rs765576982 Chr.14: 99174768-99174768 on Build GRCh38
rs963008719 Chr.14: 99174880-99174880 on Build GRCh38
rs756802485 Chr.14: 99174937-99174937 on Build GRCh38
rs754658390 Chr.14: 99174729-99174729 on Build GRCh38
rs762756917 Chr.14: 99175020-99175020 on Build GRCh38
rs531213868 Chr.14: 99174814-99174814 on Build GRCh38
rs777226398 Chr.14: 99174817-99174817 on Build GRCh38
rs754171079 Chr.14: 99174694-99174694 on Build GRCh38
rs139779656 Chr.14: 99174718-99174718 on Build GRCh38
rs754746674 Chr.14: 99174801-99174801 on Build GRCh38
rs1035143185 Chr.14: 99174840-99174840 on Build GRCh38
rs115625258 Chr.14: 99174737-99174737 on Build GRCh38
rs1023729728 Chr.14: 99174867-99174867 on Build GRCh38
rs375433500 Chr.14: 99174640-99174640 on Build GRCh38
rs765485153 Chr.14: 99174700-99174700 on Build GRCh38
rs765155091 Chr.14: 99175013-99175013 on Build GRCh38
rs757221112 Chr.14: 99174637-99174637 on Build GRCh38
rs200687140 Chr.14: 99174972-99174972 on Build GRCh38
rs564036489 Chr.14: 99174799-99174799 on Build GRCh38
rs145319589 Chr.14: 99174747-99174747 on Build GRCh38
rs916578228 Chr.14: 99174664-99174664 on Build GRCh38
rs781781969 Chr.14: 99174946-99174946 on Build GRCh38
rs746366763 Chr.14: 99174949-99174949 on Build GRCh38
rs761905083 Chr.14: 99174850-99174850 on Build GRCh38
rs867236646 Chr.14: 99174993-99174993 on Build GRCh38
rs779842515 Chr.14: 99174724-99174724 on Build GRCh38
rs761274858 Chr.14: 99174685-99174685 on Build GRCh38
rs968881427 Chr.14: 99174889-99174912 on Build GRCh38
rs775842333 Chr.14: 99174838-99174838 on Build GRCh38
rs755173953 Chr.14: 99174616-99174616 on Build GRCh38
rs371673923 Chr.14: 99174691-99174691 on Build GRCh38
rs756720526 Chr.14: 99175050-99175050 on Build GRCh38
rs974081519 Chr.14: 99174886-99174886 on Build GRCh38
rs1043387726 Chr.14: 99174929-99174929 on Build GRCh38
rs768249265 Chr.14: 99174933-99174947 on Build GRCh38
rs751603553 Chr.14: 99174796-99174796 on Build GRCh38
rs778574887 Chr.14: 99174802-99174802 on Build GRCh38
rs1010586637 Chr.14: 99174786-99174786 on Build GRCh38
rs769448442 Chr.14: 99174655-99174655 on Build GRCh38
rs767532473 Chr.14: 99174715-99174715 on Build GRCh38
rs780863859 Chr.14: 99175044-99175044 on Build GRCh38
rs865985916 Chr.14: 99174651-99174651 on Build GRCh38
rs759392301 Chr.14: 99174861-99174861 on Build GRCh38
rs556840516 Chr.14: 99174976-99174976 on Build GRCh38
rs374549866 Chr.14: 99175041-99175041 on Build GRCh38
rs745727457 Chr.14: 99174643-99174643 on Build GRCh38
rs776605814 Chr.14: 99175005-99175005 on Build GRCh38
rs779122255 Chr.14: 99174620-99174620 on Build GRCh38
rs775826732 Chr.14: 99174769-99174769 on Build GRCh38
rs764314197 Chr.14: 99174845-99174845 on Build GRCh38
rs759937138 Chr.14: 99174749-99174749 on Build GRCh38
rs746458540 Chr.14: 99174815-99174815 on Build GRCh38
rs746281008 Chr.14: 99174821-99174821 on Build GRCh38
rs780290076 Chr.14: 99174956-99174956 on Build GRCh38
rs770369592 Chr.14: 99174953-99174953 on Build GRCh38
rs775290403 Chr.14: 99174915-99174932 on Build GRCh38
rs757991781 Chr.14: 99174921-99174921 on Build GRCh38
rs752852673 Chr.14: 99174702-99174702 on Build GRCh38
rs911686691 Chr.14: 99174892-99174892 on Build GRCh38
rs763700243 Chr.14: 99175027-99175027 on Build GRCh38
rs200835381 Chr.14: 99174978-99174978 on Build GRCh38
rs771593562 Chr.14: 99174816-99174816 on Build GRCh38
rs764070866 Chr.14: 99174788-99174788 on Build GRCh38
rs990898783 Chr.14: 99174642-99174642 on Build GRCh38
rs375263416 Chr.14: 99174709-99174709 on Build GRCh38
rs749016527 Chr.14: 99174727-99174727 on Build GRCh38
rs757984083 Chr.14: 99174810-99174810 on Build GRCh38
rs756887977 Chr.14: 99175043-99175043 on Build GRCh38
rs576785327 Chr.14: 99174663-99174663 on Build GRCh38
rs868859021 Chr.14: 99174776-99174776 on Build GRCh38
rs868732382 Chr.14: 99174647-99174647 on Build GRCh38
rs753657450 Chr.14: 99174800-99174800 on Build GRCh38
rs748930743 Chr.14: 99174679-99174679 on Build GRCh38
rs547124145 Chr.14: 99175060-99175060 on Build GRCh38
rs918611301 Chr.14: 99174888-99174888 on Build GRCh38
rs752462647 Chr.14: 99175014-99175014 on Build GRCh38
rs759482444 Chr.14: 99175006-99175006 on Build GRCh38
rs377444580 Chr.14: 99175047-99175047 on Build GRCh38
rs747457528 Chr.14: 99174740-99174740 on Build GRCh38
rs772657790 Chr.14: 99174688-99174688 on Build GRCh38
rs944528801 Chr.14: 99174896-99174896 on Build GRCh38
rs372020796 Chr.14: 99174746-99174746 on Build GRCh38
rs770281394 Chr.14: 99174835-99174835 on Build GRCh38
rs747749567 Chr.14: 99174805-99174805 on Build GRCh38
rs747744449 Chr.14: 99174733-99174733 on Build GRCh38
rs763163522 Chr.14: 99174778-99174778 on Build GRCh38
rs12890136 Chr.14: 99175009-99175009 on Build GRCh38
rs774326484 Chr.14: 99174846-99174846 on Build GRCh38
rs766353606 Chr.14: 99174856-99174856 on Build GRCh38
rs763101782 Chr.14: 99174841-99174841 on Build GRCh38

View Full Product Details